Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102919727-102920220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM2
Entrez Gene ID
54805
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Gene Aliases
ACDP2, HOMG6, HOMGSMR
UniGene
Hs.643509
Gene Chromosome Location
Chr.10: 102918293-103087169 on Build GRCh38
Transcripts
NM_017649.4 NM_199076.2 NM_199077.2 XM_005269933.4 XM_006717908.2 XM_011539911.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011331 c.1264G>A p.D422N Chr.10: 102919744-102919744 on Build GRCh38
3414647 c.1429G>A p.G477S Chr.10: 102919909-102919909 on Build GRCh38
5929175 c.1466C>T p.S489F Chr.10: 102919946-102919946 on Build GRCh38
7251047 c.1344C>T p.D448D Chr.10: 102919824-102919824 on Build GRCh38
3433814 c.1389C>T p.A463A Chr.10: 102919869-102919869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752337072 Chr.10: 102920010-102920010 on Build GRCh38
rs866823473 Chr.10: 102919919-102919919 on Build GRCh38
rs773801491 Chr.10: 102919999-102919999 on Build GRCh38
rs775787922 Chr.10: 102919946-102919946 on Build GRCh38
rs1005861792 Chr.10: 102920188-102920188 on Build GRCh38
rs750803704 Chr.10: 102919761-102919761 on Build GRCh38
rs768841980 Chr.10: 102919953-102919953 on Build GRCh38
rs766784112 Chr.10: 102920008-102920008 on Build GRCh38
rs755806846 Chr.10: 102920117-102920117 on Build GRCh38
rs749515393 Chr.10: 102920119-102920119 on Build GRCh38
rs75961613 Chr.10: 102919924-102919924 on Build GRCh38
rs960390566 Chr.10: 102920194-102920194 on Build GRCh38
rs763560961 Chr.10: 102920041-102920041 on Build GRCh38
rs1049540437 Chr.10: 102920179-102920179 on Build GRCh38
rs915531187 Chr.10: 102919915-102919915 on Build GRCh38
rs751115904 Chr.10: 102920060-102920060 on Build GRCh38
rs865847626 Chr.10: 102919868-102919868 on Build GRCh38
rs780947911 Chr.10: 102920080-102920080 on Build GRCh38
rs560592481 Chr.10: 102919770-102919770 on Build GRCh38
rs996600000 Chr.10: 102920133-102920133 on Build GRCh38
rs746947940 Chr.10: 102919861-102919861 on Build GRCh38
rs928166628 Chr.10: 102919806-102919806 on Build GRCh38
rs764757429 Chr.10: 102919808-102919808 on Build GRCh38
rs769050615 Chr.10: 102920135-102920135 on Build GRCh38
rs745597919 Chr.10: 102919914-102919914 on Build GRCh38
rs772198222 Chr.10: 102920150-102920150 on Build GRCh38
rs774636915 Chr.10: 102919960-102919960 on Build GRCh38
rs371958180 Chr.10: 102919764-102919764 on Build GRCh38
rs781015921 Chr.10: 102919887-102919887 on Build GRCh38
rs888400828 Chr.10: 102920184-102920184 on Build GRCh38
rs552611833 Chr.10: 102920189-102920189 on Build GRCh38
rs745821665 Chr.10: 102920091-102920091 on Build GRCh38
rs998058765 Chr.10: 102920067-102920067 on Build GRCh38
rs757849792 Chr.10: 102919848-102919848 on Build GRCh38
rs146507239 Chr.10: 102920077-102920077 on Build GRCh38
rs1041170083 Chr.10: 102920018-102920018 on Build GRCh38
rs774738741 Chr.10: 102920139-102920139 on Build GRCh38
rs139440669 Chr.10: 102919845-102919845 on Build GRCh38
rs748138258 Chr.10: 102920143-102920143 on Build GRCh38
rs760881297 Chr.10: 102920007-102920007 on Build GRCh38
rs900905069 Chr.10: 102920037-102920037 on Build GRCh38
rs767894648 Chr.10: 102919755-102919755 on Build GRCh38
rs769658377 Chr.10: 102919927-102919927 on Build GRCh38
rs377103063 Chr.10: 102919948-102919948 on Build GRCh38
rs946949528 Chr.10: 102919941-102919941 on Build GRCh38
rs757179091 Chr.10: 102919863-102919863 on Build GRCh38
rs777325600 Chr.10: 102919857-102919857 on Build GRCh38
rs938137051 Chr.10: 102919815-102919815 on Build GRCh38
rs369542125 Chr.10: 102920058-102920058 on Build GRCh38
rs760447162 Chr.10: 102920116-102920117 on Build GRCh38
rs527653944 Chr.10: 102919851-102919851 on Build GRCh38
rs991079302 Chr.10: 102919886-102919886 on Build GRCh38
rs1048600759 Chr.10: 102919827-102919827 on Build GRCh38
rs144140948 Chr.10: 102919995-102919995 on Build GRCh38
rs752355514 Chr.10: 102919836-102919836 on Build GRCh38
rs766367564 Chr.10: 102920138-102920141 on Build GRCh38
rs369230465 Chr.10: 102919965-102919965 on Build GRCh38
rs758115800 Chr.10: 102920020-102920020 on Build GRCh38
rs1015623300 Chr.10: 102920192-102920192 on Build GRCh38
rs901017398 Chr.10: 102920124-102920124 on Build GRCh38
rs202014868 Chr.10: 102919773-102919773 on Build GRCh38

View Full Product Details