Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103383106-103383556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF5
Entrez Gene ID
6877
Gene Name
TATA-box binding protein associated factor 5
Gene Aliases
TAF(II)100, TAF2D, TAFII-100, TAFII100
UniGene
Hs.96103
Gene Chromosome Location
Chr.10: 103367953-103389065 on Build GRCh38
Transcripts
NM_006951.4 NM_139052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928906 c.1535-1G>C p.? Chr.10: 103383237-103383237 on Build GRCh38
76669 c.1543C>G p.L515V Chr.10: 103383246-103383246 on Build GRCh38
6796398 c.1659G>A p.P553P Chr.10: 103383362-103383362 on Build GRCh38
4882107 c.1559C>T p.S520L Chr.10: 103383262-103383262 on Build GRCh38
914328 c.1555G>T p.E519* Chr.10: 103383258-103383258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748924169 Chr.10: 103383385-103383385 on Build GRCh38
rs374375232 Chr.10: 103383406-103383406 on Build GRCh38
rs1052878857 Chr.10: 103383129-103383129 on Build GRCh38
rs373652183 Chr.10: 103383340-103383340 on Build GRCh38
rs143265231 Chr.10: 103383362-103383362 on Build GRCh38
rs760525086 Chr.10: 103383308-103383308 on Build GRCh38
rs569232810 Chr.10: 103383248-103383248 on Build GRCh38
rs758942294 Chr.10: 103383361-103383361 on Build GRCh38
rs759704884 Chr.10: 103383335-103383335 on Build GRCh38
rs778259468 Chr.10: 103383196-103383196 on Build GRCh38
rs779700559 Chr.10: 103383215-103383215 on Build GRCh38
rs201820525 Chr.10: 103383193-103383193 on Build GRCh38
rs768174498 Chr.10: 103383244-103383244 on Build GRCh38
rs760606812 Chr.10: 103383407-103383407 on Build GRCh38
rs201620768 Chr.10: 103383287-103383287 on Build GRCh38
rs374775425 Chr.10: 103383441-103383441 on Build GRCh38
rs370040243 Chr.10: 103383401-103383401 on Build GRCh38
rs891518046 Chr.10: 103383130-103383130 on Build GRCh38
rs371499142 Chr.10: 103383409-103383409 on Build GRCh38
rs769037968 Chr.10: 103383495-103383495 on Build GRCh38
rs747726628 Chr.10: 103383399-103383399 on Build GRCh38
rs769565807 Chr.10: 103383208-103383208 on Build GRCh38
rs773758118 Chr.10: 103383246-103383246 on Build GRCh38
rs781443624 Chr.10: 103383367-103383367 on Build GRCh38
rs977770345 Chr.10: 103383452-103383452 on Build GRCh38
rs766096431 Chr.10: 103383315-103383315 on Build GRCh38
rs772040036 Chr.10: 103383257-103383257 on Build GRCh38
rs148301664 Chr.10: 103383491-103383491 on Build GRCh38
rs771532311 Chr.10: 103383483-103383483 on Build GRCh38
rs368658296 Chr.10: 103383145-103383151 on Build GRCh38
rs754021084 Chr.10: 103383322-103383322 on Build GRCh38
rs888025690 Chr.10: 103383132-103383132 on Build GRCh38
rs748898525 Chr.10: 103383220-103383220 on Build GRCh38
rs377022068 Chr.10: 103383396-103383396 on Build GRCh38
rs992237787 Chr.10: 103383481-103383481 on Build GRCh38
rs200515297 Chr.10: 103383218-103383218 on Build GRCh38
rs747516996 Chr.10: 103383463-103383463 on Build GRCh38
rs1014911403 Chr.10: 103383183-103383183 on Build GRCh38
rs757575612 Chr.10: 103383365-103383365 on Build GRCh38
rs755023143 Chr.10: 103383187-103383187 on Build GRCh38
rs962576272 Chr.10: 103383425-103383425 on Build GRCh38
rs961105486 Chr.10: 103383475-103383475 on Build GRCh38
rs747206978 Chr.10: 103383192-103383192 on Build GRCh38
rs778459312 Chr.10: 103383398-103383398 on Build GRCh38
rs142639122 Chr.10: 103383344-103383344 on Build GRCh38
rs551026020 Chr.10: 103383197-103383197 on Build GRCh38
rs1025739942 Chr.10: 103383473-103383473 on Build GRCh38

View Full Product Details