Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103455436-103455940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALHM1
Entrez Gene ID
255022
Gene Name
calcium homeostasis modulator 1
Gene Aliases
FAM26C
UniGene
Hs.680365
Gene Chromosome Location
Chr.10: 103453387-103458891 on Build GRCh38
Transcripts
NM_001001412.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6619078 c.571T>C p.F191L Chr.10: 103455732-103455732 on Build GRCh38
2059952 c.682C>T p.R228C Chr.10: 103455621-103455621 on Build GRCh38
4409420 c.616C>T p.R206W Chr.10: 103455687-103455687 on Build GRCh38
6850846 c.618G>T p.R206R Chr.10: 103455685-103455685 on Build GRCh38
4668901 c.683G>A p.R228H Chr.10: 103455620-103455620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754682859 Chr.10: 103455696-103455696 on Build GRCh38
rs752676273 Chr.10: 103455550-103455550 on Build GRCh38
rs932190502 Chr.10: 103455821-103455821 on Build GRCh38
rs538993041 Chr.10: 103455493-103455493 on Build GRCh38
rs746852964 Chr.10: 103455665-103455665 on Build GRCh38
rs1012800089 Chr.10: 103455685-103455685 on Build GRCh38
rs761570343 Chr.10: 103455509-103455509 on Build GRCh38
rs529972583 Chr.10: 103455713-103455713 on Build GRCh38
rs762887943 Chr.10: 103455455-103455455 on Build GRCh38
rs367692653 Chr.10: 103455472-103455472 on Build GRCh38
rs756079069 Chr.10: 103455609-103455609 on Build GRCh38
rs10883870 Chr.10: 103455894-103455894 on Build GRCh38
rs752658994 Chr.10: 103455542-103455542 on Build GRCh38
rs767821542 Chr.10: 103455795-103455795 on Build GRCh38
rs763535470 Chr.10: 103455610-103455610 on Build GRCh38
rs575829022 Chr.10: 103455547-103455547 on Build GRCh38
rs914535949 Chr.10: 103455829-103455829 on Build GRCh38
rs768566602 Chr.10: 103455588-103455588 on Build GRCh38
rs746847505 Chr.10: 103455699-103455699 on Build GRCh38
rs201449648 Chr.10: 103455484-103455484 on Build GRCh38
rs138505960 Chr.10: 103455874-103455874 on Build GRCh38
rs760942799 Chr.10: 103455601-103455601 on Build GRCh38
rs1038091144 Chr.10: 103455764-103455764 on Build GRCh38
rs772526563 Chr.10: 103455860-103455860 on Build GRCh38
rs889121496 Chr.10: 103455563-103455563 on Build GRCh38
rs773083586 Chr.10: 103455595-103455595 on Build GRCh38
rs551851691 Chr.10: 103455868-103455868 on Build GRCh38
rs12241622 Chr.10: 103455823-103455823 on Build GRCh38
rs778783921 Chr.10: 103455589-103455589 on Build GRCh38
rs991817072 Chr.10: 103455464-103455464 on Build GRCh38
rs916261032 Chr.10: 103455474-103455474 on Build GRCh38
rs139879192 Chr.10: 103455559-103455559 on Build GRCh38
rs147896633 Chr.10: 103455511-103455511 on Build GRCh38
rs910632929 Chr.10: 103455734-103455734 on Build GRCh38
rs756216448 Chr.10: 103455469-103455469 on Build GRCh38
rs766519044 Chr.10: 103455531-103455531 on Build GRCh38
rs988044625 Chr.10: 103455690-103455690 on Build GRCh38
rs149217930 Chr.10: 103455672-103455672 on Build GRCh38
rs767873202 Chr.10: 103455694-103455694 on Build GRCh38
rs184539423 Chr.10: 103455463-103455463 on Build GRCh38
rs758249278 Chr.10: 103455703-103455703 on Build GRCh38
rs1057361329 Chr.10: 103455800-103455800 on Build GRCh38
rs17853566 Chr.10: 103455513-103455513 on Build GRCh38
rs745535086 Chr.10: 103455490-103455490 on Build GRCh38
rs368732643 Chr.10: 103455624-103455624 on Build GRCh38
rs144739725 Chr.10: 103455510-103455510 on Build GRCh38
rs754005560 Chr.10: 103455535-103455535 on Build GRCh38
rs779383826 Chr.10: 103455477-103455477 on Build GRCh38
rs756975023 Chr.10: 103455624-103455625 on Build GRCh38
rs147103319 Chr.10: 103455607-103455607 on Build GRCh38
rs755567358 Chr.10: 103455475-103455475 on Build GRCh38
rs764322784 Chr.10: 103455616-103455616 on Build GRCh38
rs370494608 Chr.10: 103455772-103455772 on Build GRCh38
rs752295530 Chr.10: 103455702-103455702 on Build GRCh38
rs777527933 Chr.10: 103455697-103455697 on Build GRCh38
rs774772359 Chr.10: 103455790-103455790 on Build GRCh38
rs991140687 Chr.10: 103455534-103455534 on Build GRCh38
rs777828850 Chr.10: 103455717-103455717 on Build GRCh38
rs375082219 Chr.10: 103455719-103455719 on Build GRCh38
rs41287502 Chr.10: 103455459-103455459 on Build GRCh38
rs551766107 Chr.10: 103455462-103455462 on Build GRCh38
rs759847536 Chr.10: 103455670-103455670 on Build GRCh38
rs764613162 Chr.10: 103455700-103455700 on Build GRCh38
rs140855060 Chr.10: 103455491-103455491 on Build GRCh38
rs371599718 Chr.10: 103455686-103455686 on Build GRCh38
rs375270464 Chr.10: 103455687-103455687 on Build GRCh38
rs753341265 Chr.10: 103455625-103455625 on Build GRCh38
rs190475563 Chr.10: 103455667-103455667 on Build GRCh38
rs767338589 Chr.10: 103455497-103455497 on Build GRCh38
rs7072564 Chr.10: 103455819-103455819 on Build GRCh38
rs758358041 Chr.10: 103455565-103455565 on Build GRCh38
rs753431247 Chr.10: 103455695-103455695 on Build GRCh38
rs771321724 Chr.10: 103455602-103455602 on Build GRCh38
rs749992074 Chr.10: 103455566-103455566 on Build GRCh38
rs34235057 Chr.10: 103455510-103455511 on Build GRCh38
rs746044661 Chr.10: 103455730-103455730 on Build GRCh38
rs923574155 Chr.10: 103455593-103455593 on Build GRCh38
rs1023379085 Chr.10: 103455826-103455826 on Build GRCh38
rs143355796 Chr.10: 103455693-103455693 on Build GRCh38
rs573376053 Chr.10: 103455606-103455606 on Build GRCh38
rs780756964 Chr.10: 103455685-103455685 on Build GRCh38
rs746800053 Chr.10: 103455724-103455724 on Build GRCh38
rs749667310 Chr.10: 103455776-103455777 on Build GRCh38
rs760992654 Chr.10: 103455517-103455517 on Build GRCh38
rs770092815 Chr.10: 103455745-103455745 on Build GRCh38
rs748845134 Chr.10: 103455578-103455578 on Build GRCh38
rs527425747 Chr.10: 103455865-103455865 on Build GRCh38
rs765898547 Chr.10: 103455549-103455549 on Build GRCh38
rs374702244 Chr.10: 103455666-103455666 on Build GRCh38
rs141859613 Chr.10: 103455570-103455570 on Build GRCh38
rs145430538 Chr.10: 103455627-103455627 on Build GRCh38
rs777402328 Chr.10: 103455635-103455635 on Build GRCh38
rs1030212556 Chr.10: 103455913-103455913 on Build GRCh38
rs150770390 Chr.10: 103455483-103455483 on Build GRCh38
rs769037813 Chr.10: 103455783-103455783 on Build GRCh38
rs750504998 Chr.10: 103455465-103455465 on Build GRCh38
rs780770385 Chr.10: 103455489-103455489 on Build GRCh38
rs779506837 Chr.10: 103455571-103455571 on Build GRCh38
rs775409079 Chr.10: 103455746-103455746 on Build GRCh38
rs1013029701 Chr.10: 103455825-103455825 on Build GRCh38
rs956372649 Chr.10: 103455893-103455893 on Build GRCh38
rs141282638 Chr.10: 103455470-103455470 on Build GRCh38
rs202160393 Chr.10: 103455729-103455729 on Build GRCh38
rs768617780 Chr.10: 103455499-103455499 on Build GRCh38
rs774172864 Chr.10: 103455501-103455501 on Build GRCh38
rs903012357 Chr.10: 103455641-103455641 on Build GRCh38
rs557348289 Chr.10: 103455496-103455496 on Build GRCh38
rs535176093 Chr.10: 103455621-103455621 on Build GRCh38
rs200674244 Chr.10: 103455794-103455794 on Build GRCh38
rs748121005 Chr.10: 103455592-103455592 on Build GRCh38
rs746192371 Chr.10: 103455817-103455817 on Build GRCh38
rs371478976 Chr.10: 103455630-103455630 on Build GRCh38
rs536458604 Chr.10: 103455564-103455564 on Build GRCh38
rs775666606 Chr.10: 103455674-103455674 on Build GRCh38

View Full Product Details