Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 118685324-118685825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACUL1
Entrez Gene ID
143384
Gene Name
CDK2 associated cullin domain 1
Gene Aliases
C10orf46, CAC1
UniGene
Hs.420024
Gene Chromosome Location
Chr.10: 118680982-118755246 on Build GRCh38
Transcripts
NM_153810.4

SNP Detail

SNP ID Chromosome Location
rs529040139 Chr.10: 118685787-118685787 on Build GRCh38
rs566718256 Chr.10: 118685774-118685774 on Build GRCh38
rs895073620 Chr.10: 118685427-118685427 on Build GRCh38
rs907683079 Chr.10: 118685394-118685394 on Build GRCh38
rs964928316 Chr.10: 118685673-118685673 on Build GRCh38
rs952244252 Chr.10: 118685464-118685464 on Build GRCh38
rs1035095903 Chr.10: 118685700-118685700 on Build GRCh38
rs999501251 Chr.10: 118685426-118685426 on Build GRCh38
rs181576021 Chr.10: 118685374-118685374 on Build GRCh38
rs559228458 Chr.10: 118685578-118685578 on Build GRCh38
rs765215313 Chr.10: 118685617-118685617 on Build GRCh38
rs529999102 Chr.10: 118685607-118685607 on Build GRCh38
rs1037596921 Chr.10: 118685403-118685403 on Build GRCh38
rs147568321 Chr.10: 118685694-118685694 on Build GRCh38
rs759482273 Chr.10: 118685551-118685551 on Build GRCh38
rs867320865 Chr.10: 118685786-118685786 on Build GRCh38
rs933674435 Chr.10: 118685588-118685588 on Build GRCh38
rs796356942 Chr.10: 118685712-118685712 on Build GRCh38
rs889707761 Chr.10: 118685616-118685616 on Build GRCh38
rs923253276 Chr.10: 118685713-118685713 on Build GRCh38
rs147001392 Chr.10: 118685568-118685568 on Build GRCh38
rs189601726 Chr.10: 118685373-118685373 on Build GRCh38
rs1022337725 Chr.10: 118685389-118685389 on Build GRCh38
rs1036677727 Chr.10: 118685564-118685564 on Build GRCh38
rs565275934 Chr.10: 118685735-118685735 on Build GRCh38
rs999999581 Chr.10: 118685699-118685699 on Build GRCh38
rs556877728 Chr.10: 118685486-118685486 on Build GRCh38
rs748266813 Chr.10: 118685364-118685364 on Build GRCh38
rs879462851 Chr.10: 118685774-118685774 on Build GRCh38
rs775402042 Chr.10: 118685667-118685667 on Build GRCh38
rs1037543535 Chr.10: 118685773-118685774 on Build GRCh38
rs960718367 Chr.10: 118685441-118685441 on Build GRCh38
rs1011882220 Chr.10: 118685434-118685434 on Build GRCh38
rs762489601 Chr.10: 118685716-118685716 on Build GRCh38
rs752027096 Chr.10: 118685765-118685765 on Build GRCh38
rs75064628 Chr.10: 118685596-118685596 on Build GRCh38
rs61863911 Chr.10: 118685446-118685446 on Build GRCh38
rs954757070 Chr.10: 118685735-118685742 on Build GRCh38
rs916862867 Chr.10: 118685462-118685462 on Build GRCh38
rs4443992 Chr.10: 118685368-118685368 on Build GRCh38
rs572461642 Chr.10: 118685741-118685741 on Build GRCh38
rs1035001646 Chr.10: 118685432-118685432 on Build GRCh38
rs369347908 Chr.10: 118685555-118685555 on Build GRCh38
rs1049669129 Chr.10: 118685604-118685604 on Build GRCh38
rs985367970 Chr.10: 118685534-118685534 on Build GRCh38
rs1021792324 Chr.10: 118685660-118685660 on Build GRCh38
rs141989475 Chr.10: 118685763-118685763 on Build GRCh38
rs747242435 Chr.10: 118685384-118685385 on Build GRCh38
rs554362824 Chr.10: 118685399-118685399 on Build GRCh38
rs952406070 Chr.10: 118685754-118685754 on Build GRCh38
rs1011707878 Chr.10: 118685602-118685602 on Build GRCh38
rs1043962199 Chr.10: 118685643-118685643 on Build GRCh38
rs759920622 Chr.10: 118685798-118685798 on Build GRCh38
rs1047837362 Chr.10: 118685381-118685381 on Build GRCh38
rs540040909 Chr.10: 118685760-118685760 on Build GRCh38
rs536910293 Chr.10: 118685469-118685469 on Build GRCh38

View Full Product Details