Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124759806-124760317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL10
Entrez Gene ID
399818
Gene Name
methyltransferase like 10
Gene Aliases
C10orf138
UniGene
Hs.720819
Gene Chromosome Location
Chr.10: 124757831-124791941 on Build GRCh38
Transcripts
NM_001304467.1 NM_001304468.1 NM_212554.3

SNP Detail

SNP ID Chromosome Location
rs929860415 Chr.10: 124759898-124759898 on Build GRCh38
rs1026869422 Chr.10: 124760177-124760177 on Build GRCh38
rs922670154 Chr.10: 124759920-124759920 on Build GRCh38
rs573373920 Chr.10: 124759965-124759965 on Build GRCh38
rs951324822 Chr.10: 124760166-124760166 on Build GRCh38
rs569478932 Chr.10: 124759906-124759906 on Build GRCh38
rs780684837 Chr.10: 124760069-124760069 on Build GRCh38
rs1014056733 Chr.10: 124760084-124760084 on Build GRCh38
rs200602002 Chr.10: 124759979-124759979 on Build GRCh38
rs1025409015 Chr.10: 124760094-124760094 on Build GRCh38
rs1031588742 Chr.10: 124760062-124760062 on Build GRCh38
rs919753602 Chr.10: 124759893-124759893 on Build GRCh38
rs984106618 Chr.10: 124759859-124759859 on Build GRCh38
rs191651591 Chr.10: 124759999-124759999 on Build GRCh38
rs974389538 Chr.10: 124760167-124760171 on Build GRCh38
rs999663108 Chr.10: 124760056-124760056 on Build GRCh38
rs1001330897 Chr.10: 124760003-124760003 on Build GRCh38
rs1052575141 Chr.10: 124759938-124759938 on Build GRCh38
rs372939008 Chr.10: 124760234-124760234 on Build GRCh38
rs114614765 Chr.10: 124759937-124759937 on Build GRCh38
rs951379257 Chr.10: 124760249-124760249 on Build GRCh38
rs1044021525 Chr.10: 124760012-124760012 on Build GRCh38
rs914173588 Chr.10: 124759954-124759954 on Build GRCh38
rs528665871 Chr.10: 124759982-124759982 on Build GRCh38
rs536635733 Chr.10: 124759914-124759914 on Build GRCh38
rs551278629 Chr.10: 124759899-124759899 on Build GRCh38
rs982613242 Chr.10: 124760287-124760287 on Build GRCh38
rs916264060 Chr.10: 124759831-124759831 on Build GRCh38
rs542994657 Chr.10: 124759830-124759830 on Build GRCh38
rs756825800 Chr.10: 124760046-124760046 on Build GRCh38
rs969083970 Chr.10: 124760122-124760122 on Build GRCh38
rs539829982 Chr.10: 124759992-124759992 on Build GRCh38
rs1055955649 Chr.10: 124760011-124760011 on Build GRCh38
rs975897747 Chr.10: 124759904-124759904 on Build GRCh38
rs969282225 Chr.10: 124759847-124759847 on Build GRCh38
rs1023290265 Chr.10: 124760089-124760089 on Build GRCh38
rs946933100 Chr.10: 124759972-124759972 on Build GRCh38
rs909857689 Chr.10: 124759885-124759885 on Build GRCh38
rs1005413989 Chr.10: 124760288-124760288 on Build GRCh38

View Full Product Details