Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 126464618-126465126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf90
Entrez Gene ID
118611
Gene Name
chromosome 10 open reading frame 90
Gene Aliases
FATS, bA422P15.2
UniGene
Hs.587663
Gene Chromosome Location
Chr.10: 126424997-126671038 on Build GRCh38
Transcripts
NM_001004298.2 XM_005269491.1 XM_005269492.2 XM_011539216.1 XM_011539217.2 XM_011539220.2 XM_011539222.2 XM_011539223.2 XM_011539224.2 XM_011539226.1 XM_011539227.2 XM_011539230.2 XM_011539231.1 XM_011539232.1 XM_011539233.1 XM_017015634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4055039 c.1460T>G p.L487R Chr.10: 126464770-126464770 on Build GRCh38
4871717 c.1800T>A p.V600V Chr.10: 126464721-126464721 on Build GRCh38
7090178 c.1821C>A p.P607P Chr.10: 126464700-126464700 on Build GRCh38
5919815 c.1825+5G>A p.? Chr.10: 126464691-126464691 on Build GRCh38
5899007 c.1640C>T p.P547L Chr.10: 126464881-126464881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552684926 Chr.10: 126465085-126465085 on Build GRCh38
rs144737013 Chr.10: 126464782-126464782 on Build GRCh38
rs758287530 Chr.10: 126464906-126464906 on Build GRCh38
rs754747554 Chr.10: 126464788-126464788 on Build GRCh38
rs149443145 Chr.10: 126464916-126464916 on Build GRCh38
rs948707503 Chr.10: 126465005-126465005 on Build GRCh38
rs373175630 Chr.10: 126464921-126464921 on Build GRCh38
rs775472544 Chr.10: 126464818-126464818 on Build GRCh38
rs549988140 Chr.10: 126464935-126464935 on Build GRCh38
rs755673362 Chr.10: 126464780-126464780 on Build GRCh38
rs1054487440 Chr.10: 126464773-126464773 on Build GRCh38
rs148047002 Chr.10: 126464752-126464752 on Build GRCh38
rs757896472 Chr.10: 126465017-126465018 on Build GRCh38
rs749861461 Chr.10: 126464777-126464777 on Build GRCh38
rs184501249 Chr.10: 126464686-126464686 on Build GRCh38
rs977962064 Chr.10: 126464939-126464939 on Build GRCh38
rs555900055 Chr.10: 126464710-126464710 on Build GRCh38
rs140833099 Chr.10: 126464771-126464771 on Build GRCh38
rs752201184 Chr.10: 126464942-126464944 on Build GRCh38
rs559110675 Chr.10: 126464668-126464668 on Build GRCh38
rs369535667 Chr.10: 126465083-126465083 on Build GRCh38
rs143599189 Chr.10: 126464723-126464723 on Build GRCh38
rs7079392 Chr.10: 126465054-126465054 on Build GRCh38
rs114882604 Chr.10: 126464816-126464816 on Build GRCh38
rs201855791 Chr.10: 126464902-126464902 on Build GRCh38
rs763231612 Chr.10: 126464847-126464847 on Build GRCh38
rs773520978 Chr.10: 126465018-126465018 on Build GRCh38
rs989151352 Chr.10: 126464757-126464757 on Build GRCh38
rs748977528 Chr.10: 126465079-126465079 on Build GRCh38
rs138339443 Chr.10: 126464815-126464815 on Build GRCh38
rs201947519 Chr.10: 126464911-126464911 on Build GRCh38
rs367883595 Chr.10: 126464688-126464688 on Build GRCh38
rs768828243 Chr.10: 126465000-126465000 on Build GRCh38
rs746933496 Chr.10: 126464912-126464912 on Build GRCh38
rs563866378 Chr.10: 126464899-126464899 on Build GRCh38
rs539426891 Chr.10: 126464767-126464767 on Build GRCh38
rs1004386819 Chr.10: 126464828-126464828 on Build GRCh38
rs1042157391 Chr.10: 126464737-126464737 on Build GRCh38
rs74814928 Chr.10: 126464859-126464859 on Build GRCh38
rs56395014 Chr.10: 126464655-126464655 on Build GRCh38
rs538048163 Chr.10: 126465089-126465089 on Build GRCh38
rs747967277 Chr.10: 126464800-126464800 on Build GRCh38
rs765334221 Chr.10: 126464735-126464735 on Build GRCh38
rs898237050 Chr.10: 126465071-126465071 on Build GRCh38
rs199706107 Chr.10: 126464891-126464891 on Build GRCh38
rs200693227 Chr.10: 126464790-126464790 on Build GRCh38
rs369045364 Chr.10: 126464930-126464930 on Build GRCh38
rs936121805 Chr.10: 126464946-126464946 on Build GRCh38
rs764531718 Chr.10: 126464652-126464652 on Build GRCh38
rs1032736930 Chr.10: 126465061-126465061 on Build GRCh38
rs766812220 Chr.10: 126465032-126465032 on Build GRCh38
rs765912475 Chr.10: 126464886-126464886 on Build GRCh38
rs772395034 Chr.10: 126465012-126465012 on Build GRCh38
rs748096673 Chr.10: 126464699-126464699 on Build GRCh38
rs930696769 Chr.10: 126464835-126464835 on Build GRCh38
rs868074908 Chr.10: 126465053-126465053 on Build GRCh38
rs764410825 Chr.10: 126464754-126464754 on Build GRCh38
rs138712008 Chr.10: 126464863-126464863 on Build GRCh38
rs761130977 Chr.10: 126464882-126464882 on Build GRCh38
rs192378808 Chr.10: 126464728-126464728 on Build GRCh38
rs758118945 Chr.10: 126464738-126464738 on Build GRCh38
rs774331993 Chr.10: 126464695-126464695 on Build GRCh38
rs777794117 Chr.10: 126464813-126464813 on Build GRCh38
rs775799341 Chr.10: 126464747-126464747 on Build GRCh38
rs371768407 Chr.10: 126464675-126464675 on Build GRCh38
rs772065691 Chr.10: 126464803-126464803 on Build GRCh38
rs529141776 Chr.10: 126465042-126465042 on Build GRCh38
rs772196639 Chr.10: 126464720-126464720 on Build GRCh38
rs768922256 Chr.10: 126464851-126464851 on Build GRCh38
rs116419137 Chr.10: 126465084-126465084 on Build GRCh38
rs924674363 Chr.10: 126464945-126464945 on Build GRCh38
rs780674970 Chr.10: 126464658-126464658 on Build GRCh38
rs754804042 Chr.10: 126464662-126464662 on Build GRCh38
rs778709567 Chr.10: 126464900-126464900 on Build GRCh38
rs761234593 Chr.10: 126465023-126465023 on Build GRCh38
rs765173646 Chr.10: 126464863-126464873 on Build GRCh38
rs759424821 Chr.10: 126464788-126464788 on Build GRCh38
rs373116712 Chr.10: 126464999-126464999 on Build GRCh38
rs571005774 Chr.10: 126465088-126465088 on Build GRCh38
rs776138355 Chr.10: 126465034-126465034 on Build GRCh38
rs748988393 Chr.10: 126464687-126464687 on Build GRCh38
rs966925442 Chr.10: 126464936-126464936 on Build GRCh38
rs995302351 Chr.10: 126465083-126465083 on Build GRCh38
rs751986892 Chr.10: 126464758-126464758 on Build GRCh38
rs11244990 Chr.10: 126464650-126464650 on Build GRCh38
rs183852527 Chr.10: 126465039-126465039 on Build GRCh38
rs762072257 Chr.10: 126465007-126465007 on Build GRCh38
rs753501756 Chr.10: 126464783-126464783 on Build GRCh38
rs774633302 Chr.10: 126464855-126464855 on Build GRCh38
rs912431558 Chr.10: 126464719-126464719 on Build GRCh38
rs778798694 Chr.10: 126464672-126464672 on Build GRCh38
rs1054456362 Chr.10: 126464979-126464979 on Build GRCh38

View Full Product Details