Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132104706-132105200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAKMIP3
Entrez Gene ID
282973
Gene Name
Janus kinase and microtubule interacting protein 3
Gene Aliases
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
UniGene
Hs.106254
Gene Chromosome Location
Chr.10: 132036068-132184852 on Build GRCh38
Transcripts
NM_001105521.3 NM_001323086.1 NM_001323087.1 NM_001323088.1 NM_001323089.1 NM_001323090.1 XM_005252674.4 XM_005252676.4 XM_011539676.2 XM_011539682.2 XM_017016095.1 XM_017016096.1 XM_017016097.1 XM_017016098.1 XM_017016099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3751628 c.108G>A p.Q36Q Chr.10: 132104916-132104916 on Build GRCh38
1346841 c.76G>A p.E26K Chr.10: 132104884-132104884 on Build GRCh38
4144492 c.60G>A p.A20A Chr.10: 132104868-132104868 on Build GRCh38
3866842 c.43G>A p.A15T Chr.10: 132104851-132104851 on Build GRCh38
6248117 c.23G>T p.S8I Chr.10: 132104831-132104831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751961106 Chr.10: 132104957-132104957 on Build GRCh38
rs780086305 Chr.10: 132104986-132104986 on Build GRCh38
rs781375095 Chr.10: 132104977-132104977 on Build GRCh38
rs370166502 Chr.10: 132105106-132105106 on Build GRCh38
rs781754484 Chr.10: 132104975-132104975 on Build GRCh38
rs754761854 Chr.10: 132104955-132104955 on Build GRCh38
rs763646214 Chr.10: 132104763-132104763 on Build GRCh38
rs570310861 Chr.10: 132105144-132105144 on Build GRCh38
rs771201494 Chr.10: 132104904-132104904 on Build GRCh38
rs560432144 Chr.10: 132104758-132104758 on Build GRCh38
rs778848311 Chr.10: 132104961-132104961 on Build GRCh38
rs1001549328 Chr.10: 132105048-132105048 on Build GRCh38
rs763181847 Chr.10: 132104929-132104929 on Build GRCh38
rs1038078407 Chr.10: 132104732-132104732 on Build GRCh38
rs1045687280 Chr.10: 132105009-132105009 on Build GRCh38
rs1008576268 Chr.10: 132104874-132104874 on Build GRCh38
rs112564062 Chr.10: 132105070-132105070 on Build GRCh38
rs980078463 Chr.10: 132104788-132104788 on Build GRCh38
rs534023524 Chr.10: 132105111-132105111 on Build GRCh38
rs779707293 Chr.10: 132104823-132104823 on Build GRCh38
rs762299520 Chr.10: 132104939-132104939 on Build GRCh38
rs10870253 Chr.10: 132104962-132104962 on Build GRCh38
rs945912458 Chr.10: 132105149-132105149 on Build GRCh38
rs749004696 Chr.10: 132104877-132104877 on Build GRCh38
rs968773301 Chr.10: 132104746-132104747 on Build GRCh38
rs373674808 Chr.10: 132104859-132104859 on Build GRCh38
rs372275166 Chr.10: 132104817-132104817 on Build GRCh38
rs532786712 Chr.10: 132104776-132104776 on Build GRCh38
rs527334713 Chr.10: 132104982-132104982 on Build GRCh38
rs540336811 Chr.10: 132104729-132104729 on Build GRCh38
rs375491238 Chr.10: 132104894-132104894 on Build GRCh38
rs896805686 Chr.10: 132104738-132104738 on Build GRCh38
rs776839956 Chr.10: 132104905-132104905 on Build GRCh38
rs753689736 Chr.10: 132104954-132104954 on Build GRCh38
rs184847396 Chr.10: 132105098-132105098 on Build GRCh38
rs907038814 Chr.10: 132104803-132104803 on Build GRCh38
rs1018471947 Chr.10: 132105129-132105129 on Build GRCh38
rs1021689436 Chr.10: 132104745-132104745 on Build GRCh38
rs758390400 Chr.10: 132104972-132104972 on Build GRCh38
rs547517909 Chr.10: 132105036-132105036 on Build GRCh38
rs574492779 Chr.10: 132105102-132105102 on Build GRCh38
rs748892200 Chr.10: 132105079-132105079 on Build GRCh38
rs546352989 Chr.10: 132105164-132105164 on Build GRCh38
rs765693147 Chr.10: 132104863-132104863 on Build GRCh38
rs377579463 Chr.10: 132104841-132104841 on Build GRCh38
rs920722597 Chr.10: 132105090-132105090 on Build GRCh38
rs747045263 Chr.10: 132104845-132104845 on Build GRCh38
rs753848756 Chr.10: 132104834-132104834 on Build GRCh38
rs756701841 Chr.10: 132104867-132104867 on Build GRCh38
rs891691272 Chr.10: 132104908-132104908 on Build GRCh38
rs879247668 Chr.10: 132104750-132104750 on Build GRCh38
rs746152484 Chr.10: 132104913-132104913 on Build GRCh38
rs11146177 Chr.10: 132104916-132104916 on Build GRCh38
rs751095579 Chr.10: 132104866-132104866 on Build GRCh38
rs77669554 Chr.10: 132105155-132105155 on Build GRCh38
rs757248875 Chr.10: 132104968-132104969 on Build GRCh38
rs550486685 Chr.10: 132105081-132105081 on Build GRCh38
rs775956780 Chr.10: 132104919-132104919 on Build GRCh38
rs545489271 Chr.10: 132104865-132104865 on Build GRCh38
rs1038271916 Chr.10: 132104844-132104844 on Build GRCh38
rs905153405 Chr.10: 132104735-132104735 on Build GRCh38
rs10870252 Chr.10: 132104868-132104868 on Build GRCh38
rs7095997 Chr.10: 132104728-132104728 on Build GRCh38
rs755866881 Chr.10: 132104821-132104821 on Build GRCh38
rs12098286 Chr.10: 132105063-132105063 on Build GRCh38
rs765906115 Chr.10: 132104952-132104952 on Build GRCh38
rs747590316 Chr.10: 132104981-132104981 on Build GRCh38
rs780777526 Chr.10: 132104813-132104813 on Build GRCh38
rs973576757 Chr.10: 132105084-132105084 on Build GRCh38
rs764601335 Chr.10: 132104932-132104932 on Build GRCh38
rs751040452 Chr.10: 132104767-132104767 on Build GRCh38
rs192795687 Chr.10: 132104966-132104966 on Build GRCh38
rs778944502 Chr.10: 132104883-132104883 on Build GRCh38
rs371883044 Chr.10: 132104949-132104949 on Build GRCh38
rs960094802 Chr.10: 132105064-132105064 on Build GRCh38
rs1021408672 Chr.10: 132104920-132104920 on Build GRCh38
rs768454996 Chr.10: 132104833-132104833 on Build GRCh38
rs774769898 Chr.10: 132104933-132104933 on Build GRCh38
rs771299481 Chr.10: 132104847-132104847 on Build GRCh38
rs570546259 Chr.10: 132105051-132105051 on Build GRCh38
rs200640026 Chr.10: 132104864-132104864 on Build GRCh38
rs148043368 Chr.10: 132105101-132105101 on Build GRCh38
rs1041108131 Chr.10: 132105124-132105124 on Build GRCh38
rs756938396 Chr.10: 132104796-132104796 on Build GRCh38
rs759673925 Chr.10: 132104862-132104862 on Build GRCh38
rs940892992 Chr.10: 132104842-132104842 on Build GRCh38
rs951104007 Chr.10: 132105165-132105165 on Build GRCh38
rs1022311210 Chr.10: 132105090-132105090 on Build GRCh38
rs749302804 Chr.10: 132104825-132104825 on Build GRCh38
rs937254737 Chr.10: 132105066-132105066 on Build GRCh38
rs955688347 Chr.10: 132105104-132105104 on Build GRCh38
rs760414853 Chr.10: 132104950-132104950 on Build GRCh38
rs12768232 Chr.10: 132105153-132105153 on Build GRCh38
rs767069355 Chr.10: 132104942-132104942 on Build GRCh38
rs77653558 Chr.10: 132104824-132104824 on Build GRCh38
rs755706630 Chr.10: 132104876-132104876 on Build GRCh38
rs751322147 Chr.10: 132104858-132104858 on Build GRCh38

View Full Product Details