Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132140322-132140843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAKMIP3
Entrez Gene ID
282973
Gene Name
Janus kinase and microtubule interacting protein 3
Gene Aliases
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
UniGene
Hs.106254
Gene Chromosome Location
Chr.10: 132036068-132184852 on Build GRCh38
Transcripts
NM_001105521.3 NM_001323086.1 NM_001323087.1 NM_001323088.1 NM_001323089.1 NM_001323090.1 XM_005252674.4 XM_005252676.4 XM_011539676.2 XM_011539682.2 XM_017016095.1 XM_017016096.1 XM_017016097.1 XM_017016098.1 XM_017016099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5419647 c.1464_1473+68del78 p.? Chr.10: 132140570-132140647 on Build GRCh38
4186024 c.1471G>C p.E491Q Chr.10: 132140577-132140577 on Build GRCh38
6393621 c.1429A>G p.R477G Chr.10: 132140535-132140535 on Build GRCh38
916323 c.1414T>A p.S472T Chr.10: 132140520-132140520 on Build GRCh38
4526577 c.1403G>A p.G468D Chr.10: 132140509-132140509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530454050 Chr.10: 132140662-132140662 on Build GRCh38
rs376570759 Chr.10: 132140504-132140504 on Build GRCh38
rs531085140 Chr.10: 132140624-132140624 on Build GRCh38
rs148865774 Chr.10: 132140672-132140672 on Build GRCh38
rs774343315 Chr.10: 132140531-132140531 on Build GRCh38
rs370890037 Chr.10: 132140459-132140459 on Build GRCh38
rs1038925827 Chr.10: 132140794-132140794 on Build GRCh38
rs373697625 Chr.10: 132140447-132140447 on Build GRCh38
rs1004248982 Chr.10: 132140718-132140718 on Build GRCh38
rs572490260 Chr.10: 132140390-132140390 on Build GRCh38
rs767320873 Chr.10: 132140606-132140607 on Build GRCh38
rs746459550 Chr.10: 132140436-132140436 on Build GRCh38
rs755802240 Chr.10: 132140567-132140567 on Build GRCh38
rs911529018 Chr.10: 132140770-132140770 on Build GRCh38
rs41311239 Chr.10: 132140676-132140676 on Build GRCh38
rs538323358 Chr.10: 132140432-132140432 on Build GRCh38
rs745493813 Chr.10: 132140445-132140445 on Build GRCh38
rs529702900 Chr.10: 132140368-132140368 on Build GRCh38
rs370909046 Chr.10: 132140431-132140431 on Build GRCh38
rs750460228 Chr.10: 132140403-132140403 on Build GRCh38
rs914203622 Chr.10: 132140805-132140805 on Build GRCh38
rs777459232 Chr.10: 132140435-132140435 on Build GRCh38
rs865964316 Chr.10: 132140372-132140372 on Build GRCh38
rs767621243 Chr.10: 132140401-132140401 on Build GRCh38
rs960570455 Chr.10: 132140731-132140731 on Build GRCh38
rs372190425 Chr.10: 132140539-132140539 on Build GRCh38
rs371762738 Chr.10: 132140391-132140391 on Build GRCh38
rs910873559 Chr.10: 132140817-132140817 on Build GRCh38
rs763072156 Chr.10: 132140431-132140432 on Build GRCh38
rs533801379 Chr.10: 132140754-132140754 on Build GRCh38
rs768588936 Chr.10: 132140524-132140524 on Build GRCh38
rs370170942 Chr.10: 132140585-132140585 on Build GRCh38
rs1052714382 Chr.10: 132140666-132140666 on Build GRCh38
rs367725920 Chr.10: 132140452-132140452 on Build GRCh38
rs1031480276 Chr.10: 132140345-132140345 on Build GRCh38
rs375807579 Chr.10: 132140584-132140584 on Build GRCh38
rs775912421 Chr.10: 132140616-132140616 on Build GRCh38
rs77263725 Chr.10: 132140796-132140796 on Build GRCh38
rs763245397 Chr.10: 132140617-132140617 on Build GRCh38
rs769911129 Chr.10: 132140615-132140615 on Build GRCh38
rs9733882 Chr.10: 132140735-132140735 on Build GRCh38
rs745866105 Chr.10: 132140614-132140614 on Build GRCh38
rs111390655 Chr.10: 132140646-132140646 on Build GRCh38
rs764099664 Chr.10: 132140468-132140470 on Build GRCh38
rs1009183234 Chr.10: 132140505-132140505 on Build GRCh38
rs778660973 Chr.10: 132140600-132140600 on Build GRCh38
rs2818387 Chr.10: 132140507-132140507 on Build GRCh38
rs754162331 Chr.10: 132140411-132140411 on Build GRCh38
rs756973826 Chr.10: 132140442-132140442 on Build GRCh38
rs756046741 Chr.10: 132140610-132140646 on Build GRCh38
rs938871908 Chr.10: 132140397-132140397 on Build GRCh38
rs754828519 Chr.10: 132140599-132140599 on Build GRCh38
rs954522841 Chr.10: 132140355-132140355 on Build GRCh38
rs568653043 Chr.10: 132140708-132140708 on Build GRCh38
rs996950514 Chr.10: 132140707-132140707 on Build GRCh38
rs9733324 Chr.10: 132140607-132140607 on Build GRCh38
rs753347714 Chr.10: 132140614-132140642 on Build GRCh38
rs866412124 Chr.10: 132140717-132140717 on Build GRCh38
rs752106852 Chr.10: 132140486-132140486 on Build GRCh38
rs952946439 Chr.10: 132140387-132140387 on Build GRCh38
rs374337157 Chr.10: 132140565-132140565 on Build GRCh38
rs766336472 Chr.10: 132140613-132140614 on Build GRCh38
rs376411265 Chr.10: 132140540-132140540 on Build GRCh38
rs746980802 Chr.10: 132140465-132140465 on Build GRCh38
rs764540181 Chr.10: 132140560-132140560 on Build GRCh38
rs760911402 Chr.10: 132140404-132140404 on Build GRCh38
rs374584141 Chr.10: 132140480-132140480 on Build GRCh38
rs551246674 Chr.10: 132140344-132140344 on Build GRCh38
rs373132662 Chr.10: 132140598-132140598 on Build GRCh38
rs145468181 Chr.10: 132140768-132140768 on Build GRCh38
rs745571804 Chr.10: 132140508-132140508 on Build GRCh38
rs368231959 Chr.10: 132140481-132140481 on Build GRCh38
rs766533789 Chr.10: 132140407-132140407 on Build GRCh38
rs559054103 Chr.10: 132140357-132140357 on Build GRCh38
rs766445517 Chr.10: 132140473-132140473 on Build GRCh38
rs755679984 Chr.10: 132140509-132140509 on Build GRCh38
rs780964724 Chr.10: 132140443-132140443 on Build GRCh38
rs567997100 Chr.10: 132140741-132140741 on Build GRCh38
rs758944313 Chr.10: 132140434-132140434 on Build GRCh38
rs537648228 Chr.10: 132140732-132140732 on Build GRCh38
rs1015592006 Chr.10: 132140730-132140730 on Build GRCh38
rs565380893 Chr.10: 132140613-132140613 on Build GRCh38
rs755441895 Chr.10: 132140414-132140414 on Build GRCh38
rs911813255 Chr.10: 132140375-132140376 on Build GRCh38
rs2637647 Chr.10: 132140763-132140763 on Build GRCh38
rs571893711 Chr.10: 132140549-132140549 on Build GRCh38
rs761982390 Chr.10: 132140585-132140585 on Build GRCh38
rs943277823 Chr.10: 132140376-132140388 on Build GRCh38
rs1039285730 Chr.10: 132140679-132140679 on Build GRCh38
rs779919328 Chr.10: 132140516-132140516 on Build GRCh38
rs749027078 Chr.10: 132140517-132140517 on Build GRCh38
rs774812498 Chr.10: 132140628-132140628 on Build GRCh38
rs998696199 Chr.10: 132140343-132140343 on Build GRCh38
rs545392654 Chr.10: 132140772-132140772 on Build GRCh38
rs368810795 Chr.10: 132140564-132140564 on Build GRCh38
rs191071195 Chr.10: 132140663-132140663 on Build GRCh38
rs564604532 Chr.10: 132140800-132140800 on Build GRCh38
rs566547846 Chr.10: 132140377-132140377 on Build GRCh38
rs191798603 Chr.10: 132140373-132140373 on Build GRCh38
rs544806114 Chr.10: 132140819-132140819 on Build GRCh38
rs930490396 Chr.10: 132140798-132140798 on Build GRCh38
rs377397710 Chr.10: 132140627-132140627 on Build GRCh38
rs748125309 Chr.10: 132140604-132140604 on Build GRCh38
rs558770517 Chr.10: 132140786-132140786 on Build GRCh38
rs752161665 Chr.10: 132140561-132140561 on Build GRCh38
rs985122754 Chr.10: 132140632-132140632 on Build GRCh38
rs1042712829 Chr.10: 132140470-132140470 on Build GRCh38
rs952431944 Chr.10: 132140775-132140775 on Build GRCh38
rs768374048 Chr.10: 132140484-132140484 on Build GRCh38
rs567967389 Chr.10: 132140360-132140360 on Build GRCh38
rs755055152 Chr.10: 132140647-132140647 on Build GRCh38
rs749851146 Chr.10: 132140566-132140566 on Build GRCh38
rs891906740 Chr.10: 132140474-132140474 on Build GRCh38
rs1048917454 Chr.10: 132140820-132140820 on Build GRCh38
rs964155092 Chr.10: 132140367-132140367 on Build GRCh38
rs1014525551 Chr.10: 132140759-132140759 on Build GRCh38
rs759834774 Chr.10: 132140551-132140551 on Build GRCh38
rs763477350 Chr.10: 132140558-132140558 on Build GRCh38
rs944601274 Chr.10: 132140678-132140678 on Build GRCh38
rs944164558 Chr.10: 132140388-132140388 on Build GRCh38
rs978886493 Chr.10: 132140753-132140753 on Build GRCh38
rs376494575 Chr.10: 132140614-132140691 on Build GRCh38
rs542588608 Chr.10: 132140552-132140552 on Build GRCh38
rs750198622 Chr.10: 132140609-132140609 on Build GRCh38
rs186558312 Chr.10: 132140640-132140640 on Build GRCh38
rs181496069 Chr.10: 132140483-132140483 on Build GRCh38
rs558643590 Chr.10: 132140453-132140453 on Build GRCh38
rs774561333 Chr.10: 132140516-132140520 on Build GRCh38
rs796857986 Chr.10: 132140769-132140769 on Build GRCh38
rs948489914 Chr.10: 132140636-132140636 on Build GRCh38
rs766757160 Chr.10: 132140699-132140699 on Build GRCh38
rs182258408 Chr.10: 132140677-132140677 on Build GRCh38

View Full Product Details