Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 218753-219265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT3
Entrez Gene ID
23410
Gene Name
sirtuin 3
Gene Aliases
SIR2L3
UniGene
Hs.716456
Gene Chromosome Location
Chr.11: 215030-236950 on Build GRCh38
Transcripts
NM_001017524.2 NM_012239.5 XM_005252835.1 XM_011519956.1 XM_011519957.1 XM_017017428.1 XM_017017429.1 XM_017017430.1 XM_017017431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6053530 c.1067G>A p.R356H Chr.11: 218944-218944 on Build GRCh38
925966 c.996G>T p.E332D Chr.11: 219015-219015 on Build GRCh38
925965 c.1034G>A p.R345Q Chr.11: 218977-218977 on Build GRCh38
6243089 c.1031A>C p.N344T Chr.11: 218980-218980 on Build GRCh38
4574277 c.985A>G p.S329G Chr.11: 219026-219026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754529225 Chr.11: 218891-218891 on Build GRCh38
rs371344453 Chr.11: 218786-218786 on Build GRCh38
rs7927733 Chr.11: 219043-219043 on Build GRCh38
rs181924090 Chr.11: 218874-218874 on Build GRCh38
rs202066094 Chr.11: 219008-219008 on Build GRCh38
rs926447027 Chr.11: 218883-218883 on Build GRCh38
rs916449564 Chr.11: 219140-219140 on Build GRCh38
rs778913038 Chr.11: 218817-218817 on Build GRCh38
rs1013389228 Chr.11: 219236-219236 on Build GRCh38
rs558889309 Chr.11: 219219-219219 on Build GRCh38
rs764877600 Chr.11: 218947-218947 on Build GRCh38
rs200051752 Chr.11: 219000-219000 on Build GRCh38
rs147582800 Chr.11: 219011-219011 on Build GRCh38
rs750639090 Chr.11: 218876-218876 on Build GRCh38
rs190926004 Chr.11: 219118-219118 on Build GRCh38
rs200574796 Chr.11: 218912-218912 on Build GRCh38
rs922657232 Chr.11: 218779-218779 on Build GRCh38
rs185287384 Chr.11: 218975-218975 on Build GRCh38
rs568579703 Chr.11: 218928-218928 on Build GRCh38
rs747320623 Chr.11: 219002-219002 on Build GRCh38
rs200076829 Chr.11: 218855-218855 on Build GRCh38
rs542238323 Chr.11: 219217-219217 on Build GRCh38
rs200928165 Chr.11: 218872-218872 on Build GRCh38
rs906226153 Chr.11: 218985-218985 on Build GRCh38
rs758359142 Chr.11: 218972-218972 on Build GRCh38
rs202091258 Chr.11: 218882-218882 on Build GRCh38
rs763323322 Chr.11: 218780-218780 on Build GRCh38
rs1040598135 Chr.11: 219182-219182 on Build GRCh38
rs1794111 Chr.11: 219123-219123 on Build GRCh38
rs1000970460 Chr.11: 219031-219031 on Build GRCh38
rs767397123 Chr.11: 219028-219028 on Build GRCh38
rs894980298 Chr.11: 219225-219225 on Build GRCh38
rs750208270 Chr.11: 219035-219035 on Build GRCh38
rs774898657 Chr.11: 218851-218851 on Build GRCh38
rs753218983 Chr.11: 218885-218885 on Build GRCh38
rs113722904 Chr.11: 218831-218831 on Build GRCh38
rs758529104 Chr.11: 219053-219053 on Build GRCh38
rs371704582 Chr.11: 218944-218944 on Build GRCh38
rs376721867 Chr.11: 219012-219012 on Build GRCh38
rs778916219 Chr.11: 218992-218992 on Build GRCh38
rs750145288 Chr.11: 218797-218797 on Build GRCh38
rs111930133 Chr.11: 218793-218793 on Build GRCh38
rs74477499 Chr.11: 218907-218907 on Build GRCh38
rs1047551814 Chr.11: 218795-218795 on Build GRCh38
rs528185851 Chr.11: 219240-219240 on Build GRCh38
rs769845879 Chr.11: 218911-218911 on Build GRCh38
rs138703964 Chr.11: 218827-218827 on Build GRCh38
rs962828768 Chr.11: 219117-219117 on Build GRCh38
rs202141668 Chr.11: 218982-218982 on Build GRCh38
rs146003473 Chr.11: 218903-218903 on Build GRCh38
rs910700781 Chr.11: 219126-219126 on Build GRCh38
rs202155071 Chr.11: 219059-219059 on Build GRCh38
rs764135602 Chr.11: 218974-218974 on Build GRCh38
rs746247349 Chr.11: 219062-219062 on Build GRCh38
rs755367349 Chr.11: 219007-219007 on Build GRCh38
rs201069150 Chr.11: 218861-218861 on Build GRCh38
rs752983087 Chr.11: 219093-219093 on Build GRCh38
rs200157246 Chr.11: 218916-218916 on Build GRCh38
rs564095864 Chr.11: 218804-218804 on Build GRCh38
rs200898000 Chr.11: 218922-218922 on Build GRCh38
rs199874973 Chr.11: 218993-218993 on Build GRCh38
rs768849128 Chr.11: 218921-218921 on Build GRCh38
rs370703132 Chr.11: 219072-219072 on Build GRCh38
rs565415803 Chr.11: 218901-218901 on Build GRCh38
rs902060553 Chr.11: 219187-219187 on Build GRCh38
rs765448828 Chr.11: 218828-218828 on Build GRCh38
rs772590751 Chr.11: 218933-218933 on Build GRCh38
rs752654725 Chr.11: 218964-218964 on Build GRCh38
rs919618911 Chr.11: 219098-219098 on Build GRCh38
rs748945617 Chr.11: 219220-219220 on Build GRCh38
rs1018915141 Chr.11: 218895-218895 on Build GRCh38
rs199962887 Chr.11: 218943-218943 on Build GRCh38
rs1042758909 Chr.11: 219205-219205 on Build GRCh38
rs753409725 Chr.11: 218809-218809 on Build GRCh38
rs907682812 Chr.11: 218915-218915 on Build GRCh38
rs563421393 Chr.11: 219157-219157 on Build GRCh38
rs201180418 Chr.11: 219070-219070 on Build GRCh38
rs760475653 Chr.11: 219084-219084 on Build GRCh38
rs769516419 Chr.11: 219065-219065 on Build GRCh38
rs749567306 Chr.11: 218984-218984 on Build GRCh38
rs145905941 Chr.11: 219202-219202 on Build GRCh38
rs748168333 Chr.11: 219051-219051 on Build GRCh38
rs201620500 Chr.11: 219018-219018 on Build GRCh38
rs139644721 Chr.11: 218832-218832 on Build GRCh38
rs761763903 Chr.11: 219079-219079 on Build GRCh38
rs755154189 Chr.11: 219044-219044 on Build GRCh38
rs779110722 Chr.11: 219049-219049 on Build GRCh38
rs749317858 Chr.11: 218918-218918 on Build GRCh38
rs1031154844 Chr.11: 218852-218852 on Build GRCh38
rs149887273 Chr.11: 218942-218942 on Build GRCh38
rs756972549 Chr.11: 219026-219026 on Build GRCh38
rs3020901 Chr.11: 218906-218906 on Build GRCh38
rs866285136 Chr.11: 219183-219183 on Build GRCh38
rs762570417 Chr.11: 219017-219017 on Build GRCh38
rs200296470 Chr.11: 218820-218820 on Build GRCh38
rs61740927 Chr.11: 218904-218904 on Build GRCh38
rs139026266 Chr.11: 218930-218930 on Build GRCh38
rs750164863 Chr.11: 219080-219080 on Build GRCh38
rs943578632 Chr.11: 219158-219158 on Build GRCh38
rs941945642 Chr.11: 218805-218805 on Build GRCh38
rs73386658 Chr.11: 218811-218811 on Build GRCh38
rs758962158 Chr.11: 218945-218945 on Build GRCh38
rs79136100 Chr.11: 219078-219078 on Build GRCh38
rs903869127 Chr.11: 219209-219209 on Build GRCh38
rs201115579 Chr.11: 218978-218978 on Build GRCh38
rs1017398995 Chr.11: 219113-219113 on Build GRCh38
rs201630330 Chr.11: 219060-219060 on Build GRCh38
rs511744 Chr.11: 219089-219089 on Build GRCh38
rs907735632 Chr.11: 218801-218801 on Build GRCh38
rs200944124 Chr.11: 218999-218999 on Build GRCh38
rs755591002 Chr.11: 218803-218803 on Build GRCh38
rs200573893 Chr.11: 218934-218934 on Build GRCh38
rs577716108 Chr.11: 219110-219110 on Build GRCh38
rs759278864 Chr.11: 219010-219010 on Build GRCh38
rs780135795 Chr.11: 218980-218980 on Build GRCh38
rs893510049 Chr.11: 219224-219224 on Build GRCh38
rs982047177 Chr.11: 219203-219203 on Build GRCh38
rs374879005 Chr.11: 218990-218990 on Build GRCh38
rs763102480 Chr.11: 218846-218846 on Build GRCh38
rs927882421 Chr.11: 219210-219210 on Build GRCh38
rs778137871 Chr.11: 219057-219057 on Build GRCh38
rs200203977 Chr.11: 218845-218845 on Build GRCh38
rs751856567 Chr.11: 218787-218787 on Build GRCh38
rs771853792 Chr.11: 218777-218777 on Build GRCh38
rs111450242 Chr.11: 218848-218848 on Build GRCh38
rs199635999 Chr.11: 218859-218859 on Build GRCh38
rs760195432 Chr.11: 218881-218881 on Build GRCh38

View Full Product Details