Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 407739-408232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIGIRR
Entrez Gene ID
59307
Gene Name
single Ig and TIR domain containing
Gene Aliases
IL-1R8, TIR8
UniGene
Hs.501624
Gene Chromosome Location
Chr.11: 405716-417454 on Build GRCh38
Transcripts
NM_001135053.1 NM_001135054.1 NM_021805.2 XM_005253044.2 XM_005253045.1 XM_005253046.1 XM_005253048.4 XM_017018099.1 XM_017018100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4629725 c.289A>G p.T97A Chr.11: 408124-408124 on Build GRCh38
5999407 c.224C>T p.S75L Chr.11: 408189-408189 on Build GRCh38
4629726 c.289A>G p.T97A Chr.11: 408124-408124 on Build GRCh38
4428056 c.472G>C p.E158Q Chr.11: 407826-407826 on Build GRCh38
6350873 c.344C>G p.P115R Chr.11: 407954-407954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750809277 Chr.11: 407904-407904 on Build GRCh38
rs752272519 Chr.11: 408024-408024 on Build GRCh38
rs745609961 Chr.11: 408058-408058 on Build GRCh38
rs773658161 Chr.11: 407981-407981 on Build GRCh38
rs754657662 Chr.11: 407928-407928 on Build GRCh38
rs142561304 Chr.11: 408155-408155 on Build GRCh38
rs751038465 Chr.11: 408119-408119 on Build GRCh38
rs767862391 Chr.11: 407901-407901 on Build GRCh38
rs1005838929 Chr.11: 408131-408131 on Build GRCh38
rs201819873 Chr.11: 408063-408063 on Build GRCh38
rs111819059 Chr.11: 407954-407954 on Build GRCh38
rs761316352 Chr.11: 408211-408211 on Build GRCh38
rs943291457 Chr.11: 408068-408068 on Build GRCh38
rs755969858 Chr.11: 408135-408135 on Build GRCh38
rs934476491 Chr.11: 407777-407777 on Build GRCh38
rs924486634 Chr.11: 407765-407765 on Build GRCh38
rs565682134 Chr.11: 407955-407955 on Build GRCh38
rs779810175 Chr.11: 408142-408142 on Build GRCh38
rs538675453 Chr.11: 407764-407764 on Build GRCh38
rs142613676 Chr.11: 408043-408043 on Build GRCh38
rs975292365 Chr.11: 407951-407951 on Build GRCh38
rs1003483573 Chr.11: 408054-408054 on Build GRCh38
rs752128835 Chr.11: 407802-407802 on Build GRCh38
rs201273535 Chr.11: 407969-407969 on Build GRCh38
rs772099766 Chr.11: 408064-408064 on Build GRCh38
rs1047539371 Chr.11: 408140-408140 on Build GRCh38
rs1038940545 Chr.11: 408088-408088 on Build GRCh38
rs373782732 Chr.11: 407892-407892 on Build GRCh38
rs778609327 Chr.11: 407937-407937 on Build GRCh38
rs867378917 Chr.11: 408037-408037 on Build GRCh38
rs766757809 Chr.11: 407926-407926 on Build GRCh38
rs1039115731 Chr.11: 407878-407878 on Build GRCh38
rs757783581 Chr.11: 408031-408031 on Build GRCh38
rs767907903 Chr.11: 407809-407809 on Build GRCh38
rs763159926 Chr.11: 407886-407886 on Build GRCh38
rs368810026 Chr.11: 407961-407961 on Build GRCh38
rs750417412 Chr.11: 408212-408212 on Build GRCh38
rs578216747 Chr.11: 407834-407834 on Build GRCh38
rs773317441 Chr.11: 408078-408078 on Build GRCh38
rs143116663 Chr.11: 408093-408093 on Build GRCh38
rs761228826 Chr.11: 407991-407991 on Build GRCh38
rs759294875 Chr.11: 407998-407998 on Build GRCh38
rs757737493 Chr.11: 407805-407805 on Build GRCh38
rs780979670 Chr.11: 407898-407899 on Build GRCh38
rs762656806 Chr.11: 408112-408112 on Build GRCh38
rs150964982 Chr.11: 408171-408171 on Build GRCh38
rs544044876 Chr.11: 408158-408158 on Build GRCh38
rs751726352 Chr.11: 407865-407866 on Build GRCh38
rs199578659 Chr.11: 408184-408184 on Build GRCh38
rs771429866 Chr.11: 408086-408086 on Build GRCh38
rs187808886 Chr.11: 407870-407870 on Build GRCh38
rs199664141 Chr.11: 407943-407943 on Build GRCh38
rs201028179 Chr.11: 408176-408176 on Build GRCh38
rs887193390 Chr.11: 408098-408098 on Build GRCh38
rs201269567 Chr.11: 407830-407830 on Build GRCh38
rs747625612 Chr.11: 407944-407944 on Build GRCh38
rs755812704 Chr.11: 407828-407828 on Build GRCh38
rs117739035 Chr.11: 408174-408174 on Build GRCh38
rs146668030 Chr.11: 408132-408132 on Build GRCh38
rs756363730 Chr.11: 407922-407922 on Build GRCh38
rs948315663 Chr.11: 408006-408006 on Build GRCh38
rs759922829 Chr.11: 408095-408095 on Build GRCh38
rs764934818 Chr.11: 408004-408004 on Build GRCh38
rs775509544 Chr.11: 407883-407883 on Build GRCh38
rs770699596 Chr.11: 407966-407966 on Build GRCh38
rs767004992 Chr.11: 408057-408057 on Build GRCh38
rs770068555 Chr.11: 407877-407877 on Build GRCh38
rs924015853 Chr.11: 408020-408020 on Build GRCh38
rs748148802 Chr.11: 407861-407861 on Build GRCh38
rs778115855 Chr.11: 407863-407863 on Build GRCh38
rs752468430 Chr.11: 408108-408108 on Build GRCh38
rs779040870 Chr.11: 408062-408062 on Build GRCh38
rs748367004 Chr.11: 408150-408150 on Build GRCh38
rs777335477 Chr.11: 408038-408038 on Build GRCh38
rs761128833 Chr.11: 408081-408081 on Build GRCh38
rs759846718 Chr.11: 407778-407778 on Build GRCh38
rs566469099 Chr.11: 407762-407762 on Build GRCh38
rs1026059327 Chr.11: 407893-407893 on Build GRCh38
rs546582704 Chr.11: 407761-407761 on Build GRCh38
rs768072506 Chr.11: 407980-407980 on Build GRCh38
rs545036605 Chr.11: 408116-408116 on Build GRCh38
rs370322489 Chr.11: 407851-407851 on Build GRCh38
rs762963483 Chr.11: 407785-407785 on Build GRCh38
rs368185991 Chr.11: 407994-407994 on Build GRCh38
rs762338399 Chr.11: 407900-407900 on Build GRCh38
rs774435571 Chr.11: 407979-407979 on Build GRCh38
rs373128798 Chr.11: 407782-407782 on Build GRCh38
rs750713561 Chr.11: 407827-407827 on Build GRCh38
rs754600155 Chr.11: 407839-407839 on Build GRCh38
rs764590975 Chr.11: 407794-407794 on Build GRCh38
rs376128512 Chr.11: 407973-407973 on Build GRCh38
rs267602872 Chr.11: 408080-408080 on Build GRCh38
rs757403538 Chr.11: 407880-407881 on Build GRCh38
rs140256248 Chr.11: 408154-408154 on Build GRCh38
rs772106987 Chr.11: 407864-407864 on Build GRCh38
rs745615956 Chr.11: 408130-408130 on Build GRCh38
rs777513772 Chr.11: 407960-407960 on Build GRCh38
rs537219351 Chr.11: 407887-407887 on Build GRCh38
rs772304578 Chr.11: 408151-408151 on Build GRCh38
rs768875219 Chr.11: 408060-408060 on Build GRCh38
rs73400585 Chr.11: 408191-408191 on Build GRCh38
rs901472987 Chr.11: 407884-407884 on Build GRCh38
rs757160590 Chr.11: 408120-408120 on Build GRCh38
rs778550158 Chr.11: 407844-407844 on Build GRCh38
rs376910339 Chr.11: 407962-407962 on Build GRCh38
rs1047620944 Chr.11: 408080-408081 on Build GRCh38
rs764990107 Chr.11: 408100-408100 on Build GRCh38
rs780878392 Chr.11: 408051-408051 on Build GRCh38
rs746732229 Chr.11: 407871-407871 on Build GRCh38
rs765342442 Chr.11: 407779-407779 on Build GRCh38
rs200272819 Chr.11: 407996-407996 on Build GRCh38
rs777216174 Chr.11: 408156-408156 on Build GRCh38
rs757184742 Chr.11: 408044-408044 on Build GRCh38

View Full Product Details