Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2406559-2407049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPM5
Entrez Gene ID
29850
Gene Name
transient receptor potential cation channel subfamily M member 5
Gene Aliases
LTRPC5, MTR1
UniGene
Hs.272287
Gene Chromosome Location
Chr.11: 2403962-2423424 on Build GRCh38
Transcripts
NM_014555.3 XM_017017628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956458 c.3243C>T p.T1081T Chr.11: 2406669-2406669 on Build GRCh38
1703345 c.3163G>A p.E1055K Chr.11: 2406749-2406749 on Build GRCh38
3446384 c.3153C>T p.V1051V Chr.11: 2406759-2406759 on Build GRCh38
3446383 c.3186C>T p.F1062F Chr.11: 2406726-2406726 on Build GRCh38
4542642 c.3225G>A p.G1075G Chr.11: 2406687-2406687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891830742 Chr.11: 2407013-2407013 on Build GRCh38
rs1053088051 Chr.11: 2407012-2407012 on Build GRCh38
rs201161956 Chr.11: 2406838-2406838 on Build GRCh38
rs373468092 Chr.11: 2406611-2406611 on Build GRCh38
rs761863535 Chr.11: 2406635-2406636 on Build GRCh38
rs779991523 Chr.11: 2406648-2406648 on Build GRCh38
rs760536858 Chr.11: 2406743-2406743 on Build GRCh38
rs892027906 Chr.11: 2406866-2406866 on Build GRCh38
rs1047464340 Chr.11: 2406869-2406869 on Build GRCh38
rs770058245 Chr.11: 2406647-2406647 on Build GRCh38
rs772166323 Chr.11: 2406656-2406657 on Build GRCh38
rs1013902304 Chr.11: 2406910-2406910 on Build GRCh38
rs929095290 Chr.11: 2406865-2406865 on Build GRCh38
rs544818251 Chr.11: 2406614-2406614 on Build GRCh38
rs895038715 Chr.11: 2406903-2406903 on Build GRCh38
rs74930763 Chr.11: 2406877-2406877 on Build GRCh38
rs768757406 Chr.11: 2406624-2406624 on Build GRCh38
rs773139950 Chr.11: 2406703-2406703 on Build GRCh38
rs550827776 Chr.11: 2406943-2406943 on Build GRCh38
rs202069779 Chr.11: 2406677-2406677 on Build GRCh38
rs750278929 Chr.11: 2406944-2406944 on Build GRCh38
rs774596529 Chr.11: 2406634-2406635 on Build GRCh38
rs376500181 Chr.11: 2406624-2406624 on Build GRCh38
rs372790250 Chr.11: 2406839-2406839 on Build GRCh38
rs749440832 Chr.11: 2406652-2406652 on Build GRCh38
rs992143081 Chr.11: 2406695-2406695 on Build GRCh38
rs750761760 Chr.11: 2406777-2406777 on Build GRCh38
rs1032393911 Chr.11: 2406950-2406950 on Build GRCh38
rs766125386 Chr.11: 2406668-2406668 on Build GRCh38
rs750851865 Chr.11: 2406679-2406679 on Build GRCh38
rs1007193895 Chr.11: 2406608-2406608 on Build GRCh38
rs1040541178 Chr.11: 2406613-2406613 on Build GRCh38
rs972360401 Chr.11: 2406924-2406924 on Build GRCh38
rs768453069 Chr.11: 2406654-2406654 on Build GRCh38
rs374941828 Chr.11: 2406806-2406806 on Build GRCh38
rs754039126 Chr.11: 2406830-2406830 on Build GRCh38
rs978174252 Chr.11: 2406948-2406948 on Build GRCh38
rs764678322 Chr.11: 2406717-2406717 on Build GRCh38
rs767251623 Chr.11: 2406612-2406612 on Build GRCh38
rs753276400 Chr.11: 2406669-2406669 on Build GRCh38
rs904618218 Chr.11: 2406997-2406997 on Build GRCh38
rs951871108 Chr.11: 2406663-2406663 on Build GRCh38
rs978157965 Chr.11: 2406727-2406727 on Build GRCh38
rs918283554 Chr.11: 2406942-2406942 on Build GRCh38
rs1031897969 Chr.11: 2406864-2406864 on Build GRCh38
rs774005790 Chr.11: 2406610-2406610 on Build GRCh38
rs966842909 Chr.11: 2406726-2406726 on Build GRCh38
rs748044762 Chr.11: 2406693-2406693 on Build GRCh38
rs374963258 Chr.11: 2406707-2406707 on Build GRCh38
rs188782990 Chr.11: 2406901-2406901 on Build GRCh38
rs569089371 Chr.11: 2406945-2406945 on Build GRCh38
rs371080043 Chr.11: 2406656-2406656 on Build GRCh38
rs184197578 Chr.11: 2406657-2406657 on Build GRCh38
rs192259493 Chr.11: 2406606-2406606 on Build GRCh38
rs527788848 Chr.11: 2406871-2406871 on Build GRCh38
rs1031929086 Chr.11: 2406646-2406646 on Build GRCh38
rs148532424 Chr.11: 2406676-2406676 on Build GRCh38
rs757774385 Chr.11: 2406678-2406678 on Build GRCh38
rs770470349 Chr.11: 2406706-2406706 on Build GRCh38
rs778404231 Chr.11: 2406630-2406630 on Build GRCh38
rs760340076 Chr.11: 2406667-2406667 on Build GRCh38
rs958206216 Chr.11: 2406960-2406960 on Build GRCh38
rs142877041 Chr.11: 2406782-2406782 on Build GRCh38
rs962325775 Chr.11: 2406890-2406893 on Build GRCh38
rs760429971 Chr.11: 2406623-2406623 on Build GRCh38
rs144705982 Chr.11: 2406666-2406666 on Build GRCh38
rs781567959 Chr.11: 2406638-2406638 on Build GRCh38
rs1044473406 Chr.11: 2406988-2406988 on Build GRCh38
rs755173191 Chr.11: 2406832-2406832 on Build GRCh38
rs985604559 Chr.11: 2406962-2406962 on Build GRCh38
rs925481201 Chr.11: 2406735-2406735 on Build GRCh38
rs757659646 Chr.11: 2406635-2406635 on Build GRCh38
rs560541515 Chr.11: 2406870-2406870 on Build GRCh38
rs944333992 Chr.11: 2406966-2406966 on Build GRCh38
rs139971788 Chr.11: 2406640-2406640 on Build GRCh38
rs200235283 Chr.11: 2406690-2406690 on Build GRCh38
rs768069139 Chr.11: 2406761-2406761 on Build GRCh38
rs762301888 Chr.11: 2406759-2406759 on Build GRCh38
rs773096462 Chr.11: 2406671-2406671 on Build GRCh38
rs749721904 Chr.11: 2406622-2406639 on Build GRCh38
rs369241710 Chr.11: 2406627-2406627 on Build GRCh38
rs568251539 Chr.11: 2406880-2406880 on Build GRCh38
rs544431515 Chr.11: 2406589-2406589 on Build GRCh38
rs753395638 Chr.11: 2406626-2406626 on Build GRCh38
rs12275532 Chr.11: 2406596-2406596 on Build GRCh38
rs753713285 Chr.11: 2406758-2406758 on Build GRCh38
rs1042555102 Chr.11: 2406586-2406586 on Build GRCh38
rs751901068 Chr.11: 2406633-2406633 on Build GRCh38
rs756275726 Chr.11: 2406681-2406681 on Build GRCh38
rs757320496 Chr.11: 2406802-2406802 on Build GRCh38
rs1022968553 Chr.11: 2406625-2406625 on Build GRCh38
rs985718927 Chr.11: 2406774-2406774 on Build GRCh38
rs936098279 Chr.11: 2406998-2406998 on Build GRCh38
rs911363207 Chr.11: 2406963-2406963 on Build GRCh38
rs1025004154 Chr.11: 2406921-2406921 on Build GRCh38
rs200237450 Chr.11: 2406792-2406792 on Build GRCh38
rs748683908 Chr.11: 2406602-2406602 on Build GRCh38
rs1057189303 Chr.11: 2406603-2406603 on Build GRCh38
rs760578660 Chr.11: 2406705-2406705 on Build GRCh38
rs200427437 Chr.11: 2406837-2406837 on Build GRCh38
rs749293183 Chr.11: 2406684-2406684 on Build GRCh38
rs992121976 Chr.11: 2406947-2406947 on Build GRCh38
rs367982158 Chr.11: 2406743-2406743 on Build GRCh38
rs772143574 Chr.11: 2406698-2406698 on Build GRCh38
rs895017735 Chr.11: 2406789-2406789 on Build GRCh38
rs1044505203 Chr.11: 2406807-2406807 on Build GRCh38

View Full Product Details