Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5841630-5842139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52E6
Entrez Gene ID
390078
Gene Name
olfactory receptor family 52 subfamily E member 6
Gene Aliases
OR11-58
UniGene
Hs.534693
Gene Chromosome Location
Chr.11: 5840956-5841897 on Build GRCh38
Transcripts
NM_001005167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2034926 c.209C>T p.S70F Chr.11: 5841689-5841689 on Build GRCh38
4145927 c.191G>T p.C64F Chr.11: 5841707-5841707 on Build GRCh38
7132588 c.127A>G p.I43V Chr.11: 5841783-5841783 on Build GRCh38
3450119 c.258C>T p.I86I Chr.11: 5841640-5841640 on Build GRCh38
229190 c.112C>T p.L38F Chr.11: 5841786-5841786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773834562 Chr.11: 5841669-5841669 on Build GRCh38
rs751655245 Chr.11: 5841788-5841788 on Build GRCh38
rs777352925 Chr.11: 5841957-5841957 on Build GRCh38
rs775609182 Chr.11: 5841977-5841977 on Build GRCh38
rs747159566 Chr.11: 5841653-5841653 on Build GRCh38
rs768099944 Chr.11: 5841836-5841836 on Build GRCh38
rs10742810 Chr.11: 5841883-5841883 on Build GRCh38
rs759755991 Chr.11: 5841790-5841790 on Build GRCh38
rs780574148 Chr.11: 5841837-5841837 on Build GRCh38
rs970177291 Chr.11: 5841850-5841850 on Build GRCh38
rs753563290 Chr.11: 5841866-5841866 on Build GRCh38
rs973804382 Chr.11: 5841906-5841906 on Build GRCh38
rs767838749 Chr.11: 5841795-5841795 on Build GRCh38
rs538041006 Chr.11: 5842056-5842056 on Build GRCh38
rs200523283 Chr.11: 5841870-5841870 on Build GRCh38
rs901032511 Chr.11: 5841751-5841751 on Build GRCh38
rs750320064 Chr.11: 5841710-5841710 on Build GRCh38
rs201264128 Chr.11: 5841912-5841912 on Build GRCh38
rs537187497 Chr.11: 5841853-5841853 on Build GRCh38
rs749081931 Chr.11: 5841959-5841959 on Build GRCh38
rs4417311 Chr.11: 5842010-5842010 on Build GRCh38
rs756323219 Chr.11: 5841800-5841800 on Build GRCh38
rs778152659 Chr.11: 5841803-5841803 on Build GRCh38
rs151316251 Chr.11: 5841664-5841664 on Build GRCh38
rs267603032 Chr.11: 5841796-5841796 on Build GRCh38
rs931087841 Chr.11: 5841878-5841878 on Build GRCh38
rs766408063 Chr.11: 5841784-5841784 on Build GRCh38
rs764263619 Chr.11: 5841915-5841915 on Build GRCh38
rs754198173 Chr.11: 5841923-5841923 on Build GRCh38
rs746512737 Chr.11: 5841835-5841835 on Build GRCh38
rs896044297 Chr.11: 5842001-5842001 on Build GRCh38
rs145361821 Chr.11: 5842087-5842087 on Build GRCh38
rs755279804 Chr.11: 5841655-5841655 on Build GRCh38
rs760733815 Chr.11: 5841979-5841979 on Build GRCh38
rs368112349 Chr.11: 5841732-5841732 on Build GRCh38
rs754205272 Chr.11: 5841810-5841810 on Build GRCh38
rs368548347 Chr.11: 5841678-5841678 on Build GRCh38
rs10769272 Chr.11: 5841754-5841754 on Build GRCh38
rs773034560 Chr.11: 5841846-5841846 on Build GRCh38
rs755757333 Chr.11: 5841956-5841956 on Build GRCh38
rs772909446 Chr.11: 5841775-5841775 on Build GRCh38
rs764367364 Chr.11: 5841980-5841980 on Build GRCh38
rs929631166 Chr.11: 5841941-5841941 on Build GRCh38
rs141739512 Chr.11: 5842052-5842052 on Build GRCh38
rs373710987 Chr.11: 5841856-5841856 on Build GRCh38
rs778010560 Chr.11: 5841742-5841742 on Build GRCh38
rs576124866 Chr.11: 5842035-5842035 on Build GRCh38
rs755224921 Chr.11: 5841731-5841731 on Build GRCh38
rs774291314 Chr.11: 5841964-5841964 on Build GRCh38
rs371265942 Chr.11: 5841698-5841698 on Build GRCh38
rs530128204 Chr.11: 5841866-5841867 on Build GRCh38
rs775013450 Chr.11: 5841681-5841681 on Build GRCh38
rs202167540 Chr.11: 5841667-5841667 on Build GRCh38
rs1028375208 Chr.11: 5841756-5841756 on Build GRCh38
rs776779395 Chr.11: 5841981-5841981 on Build GRCh38
rs572257449 Chr.11: 5841974-5841974 on Build GRCh38
rs908226741 Chr.11: 5841891-5841891 on Build GRCh38
rs1000195468 Chr.11: 5841776-5841776 on Build GRCh38
rs78994460 Chr.11: 5841928-5841928 on Build GRCh38
rs760273331 Chr.11: 5841689-5841689 on Build GRCh38
rs769350530 Chr.11: 5841845-5841845 on Build GRCh38
rs759317948 Chr.11: 5841867-5841867 on Build GRCh38
rs769568435 Chr.11: 5841768-5841768 on Build GRCh38
rs752904994 Chr.11: 5841797-5841797 on Build GRCh38
rs745758641 Chr.11: 5841971-5841971 on Build GRCh38
rs139412237 Chr.11: 5841668-5841668 on Build GRCh38
rs202173006 Chr.11: 5841828-5841828 on Build GRCh38
rs61737905 Chr.11: 5841765-5841765 on Build GRCh38
rs745355353 Chr.11: 5841677-5841677 on Build GRCh38
rs757731125 Chr.11: 5841814-5841814 on Build GRCh38
rs376771083 Chr.11: 5841781-5841781 on Build GRCh38
rs747657945 Chr.11: 5841951-5841951 on Build GRCh38
rs367914682 Chr.11: 5841870-5841870 on Build GRCh38
rs190824959 Chr.11: 5842063-5842063 on Build GRCh38
rs758938601 Chr.11: 5841933-5841933 on Build GRCh38
rs763785273 Chr.11: 5841696-5841696 on Build GRCh38
rs746561944 Chr.11: 5841753-5841753 on Build GRCh38
rs780701372 Chr.11: 5841942-5841942 on Build GRCh38
rs779517383 Chr.11: 5841748-5841748 on Build GRCh38
rs759558561 Chr.11: 5841871-5841871 on Build GRCh38
rs766523236 Chr.11: 5841719-5841719 on Build GRCh38
rs369597662 Chr.11: 5842050-5842050 on Build GRCh38
rs4362173 Chr.11: 5841783-5841783 on Build GRCh38
rs896111509 Chr.11: 5841699-5841699 on Build GRCh38
rs750866067 Chr.11: 5841931-5841931 on Build GRCh38
rs4363637 Chr.11: 5842076-5842076 on Build GRCh38
rs1029989733 Chr.11: 5841801-5841801 on Build GRCh38
rs1050061167 Chr.11: 5842054-5842054 on Build GRCh38
rs771451239 Chr.11: 5841747-5841747 on Build GRCh38
rs1056168859 Chr.11: 5841961-5841961 on Build GRCh38
rs542821340 Chr.11: 5841986-5841986 on Build GRCh38
rs367705230 Chr.11: 5841759-5841759 on Build GRCh38
rs762802306 Chr.11: 5841711-5841711 on Build GRCh38
rs759016595 Chr.11: 5841803-5841803 on Build GRCh38
rs4495918 Chr.11: 5841707-5841707 on Build GRCh38
rs760931072 Chr.11: 5841897-5841897 on Build GRCh38
rs747793517 Chr.11: 5841840-5841840 on Build GRCh38
rs758642208 Chr.11: 5842066-5842066 on Build GRCh38
rs61739208 Chr.11: 5841735-5841735 on Build GRCh38
rs375869025 Chr.11: 5841875-5841875 on Build GRCh38
rs565704528 Chr.11: 5841744-5841744 on Build GRCh38

View Full Product Details