Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6239000-6239506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA4
Entrez Gene ID
1262
Gene Name
cyclic nucleotide gated channel alpha 4
Gene Aliases
CNCA2, CNG-4, CNG4, CNG5, CNGB2, OCNC2, OCNCBETA, OCNCb
UniGene
Hs.434618
Gene Chromosome Location
Chr.11: 6234774-6245924 on Build GRCh38
Transcripts
NM_001037329.3 XM_017017217.1 XM_017017218.1 XM_017017219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6133996 c.49C>A p.P17T Chr.11: 6239255-6239255 on Build GRCh38
3451112 c.153C>T p.I51I Chr.11: 6239474-6239474 on Build GRCh38
4529881 c.164G>A p.R55K Chr.11: 6239485-6239485 on Build GRCh38
2039847 c.63G>A p.R21R Chr.11: 6239384-6239384 on Build GRCh38
6014272 c.84C>T p.D28D Chr.11: 6239405-6239405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961722316 Chr.11: 6239176-6239176 on Build GRCh38
rs915748554 Chr.11: 6239248-6239248 on Build GRCh38
rs200595421 Chr.11: 6239196-6239196 on Build GRCh38
rs561237176 Chr.11: 6239160-6239160 on Build GRCh38
rs767349046 Chr.11: 6239255-6239255 on Build GRCh38
rs774696668 Chr.11: 6239291-6239291 on Build GRCh38
rs771727779 Chr.11: 6239465-6239465 on Build GRCh38
rs750846325 Chr.11: 6239427-6239427 on Build GRCh38
rs760471518 Chr.11: 6239478-6239478 on Build GRCh38
rs866642764 Chr.11: 6239396-6239396 on Build GRCh38
rs1019912111 Chr.11: 6239292-6239292 on Build GRCh38
rs775262711 Chr.11: 6239380-6239380 on Build GRCh38
rs374760938 Chr.11: 6239413-6239413 on Build GRCh38
rs755032064 Chr.11: 6239168-6239168 on Build GRCh38
rs769533819 Chr.11: 6239229-6239229 on Build GRCh38
rs774440361 Chr.11: 6239239-6239239 on Build GRCh38
rs200166545 Chr.11: 6239389-6239389 on Build GRCh38
rs749126452 Chr.11: 6239209-6239209 on Build GRCh38
rs778410133 Chr.11: 6239458-6239458 on Build GRCh38
rs150511061 Chr.11: 6239041-6239041 on Build GRCh38
rs772848420 Chr.11: 6239369-6239372 on Build GRCh38
rs768728086 Chr.11: 6239483-6239483 on Build GRCh38
rs772966475 Chr.11: 6239231-6239231 on Build GRCh38
rs770362473 Chr.11: 6239182-6239182 on Build GRCh38
rs749705965 Chr.11: 6239188-6239188 on Build GRCh38
rs757592061 Chr.11: 6239189-6239189 on Build GRCh38
rs754024343 Chr.11: 6239277-6239277 on Build GRCh38
rs188149923 Chr.11: 6239269-6239269 on Build GRCh38
rs553970560 Chr.11: 6239040-6239040 on Build GRCh38
rs1008453704 Chr.11: 6239130-6239130 on Build GRCh38
rs547565362 Chr.11: 6239430-6239430 on Build GRCh38
rs1028455919 Chr.11: 6239115-6239115 on Build GRCh38
rs777962404 Chr.11: 6239439-6239439 on Build GRCh38
rs374451892 Chr.11: 6239249-6239249 on Build GRCh38
rs770763576 Chr.11: 6239443-6239443 on Build GRCh38
rs757641347 Chr.11: 6239279-6239279 on Build GRCh38
rs371032766 Chr.11: 6239207-6239207 on Build GRCh38
rs758032411 Chr.11: 6239222-6239222 on Build GRCh38
rs747955587 Chr.11: 6239293-6239293 on Build GRCh38
rs760324772 Chr.11: 6239369-6239370 on Build GRCh38
rs756428629 Chr.11: 6239184-6239184 on Build GRCh38
rs764343673 Chr.11: 6239273-6239273 on Build GRCh38
rs914625076 Chr.11: 6239344-6239344 on Build GRCh38
rs775525107 Chr.11: 6239477-6239477 on Build GRCh38
rs201804648 Chr.11: 6239345-6239345 on Build GRCh38
rs971148295 Chr.11: 6239323-6239323 on Build GRCh38
rs765442437 Chr.11: 6239415-6239415 on Build GRCh38
rs751497800 Chr.11: 6239167-6239167 on Build GRCh38
rs771264377 Chr.11: 6239055-6239055 on Build GRCh38
rs376537710 Chr.11: 6239205-6239205 on Build GRCh38
rs769146038 Chr.11: 6239318-6239318 on Build GRCh38
rs142160386 Chr.11: 6239442-6239442 on Build GRCh38
rs951807000 Chr.11: 6239221-6239221 on Build GRCh38
rs372687394 Chr.11: 6239346-6239346 on Build GRCh38
rs996962889 Chr.11: 6239078-6239081 on Build GRCh38
rs774352532 Chr.11: 6239360-6239360 on Build GRCh38
rs759000450 Chr.11: 6239296-6239296 on Build GRCh38
rs754073624 Chr.11: 6239398-6239398 on Build GRCh38
rs747733867 Chr.11: 6239314-6239314 on Build GRCh38
rs1008839909 Chr.11: 6239404-6239404 on Build GRCh38
rs569931495 Chr.11: 6239337-6239337 on Build GRCh38
rs773017287 Chr.11: 6239334-6239334 on Build GRCh38
rs760843463 Chr.11: 6239386-6239386 on Build GRCh38
rs892798219 Chr.11: 6239280-6239280 on Build GRCh38
rs542612027 Chr.11: 6239156-6239156 on Build GRCh38
rs891306380 Chr.11: 6239385-6239385 on Build GRCh38
rs746326695 Chr.11: 6239200-6239200 on Build GRCh38
rs934678854 Chr.11: 6239286-6239286 on Build GRCh38
rs971276895 Chr.11: 6239260-6239260 on Build GRCh38
rs778759273 Chr.11: 6239276-6239277 on Build GRCh38
rs111379180 Chr.11: 6239186-6239186 on Build GRCh38
rs767542514 Chr.11: 6239362-6239362 on Build GRCh38
rs771821659 Chr.11: 6239345-6239345 on Build GRCh38
rs759240785 Chr.11: 6239361-6239361 on Build GRCh38
rs781344369 Chr.11: 6239081-6239081 on Build GRCh38
rs564778445 Chr.11: 6239173-6239173 on Build GRCh38
rs766223585 Chr.11: 6239234-6239234 on Build GRCh38
rs752882495 Chr.11: 6239264-6239264 on Build GRCh38
rs536010222 Chr.11: 6239172-6239172 on Build GRCh38
rs780402156 Chr.11: 6239302-6239302 on Build GRCh38
rs751961985 Chr.11: 6239433-6239433 on Build GRCh38
rs762871148 Chr.11: 6239232-6239232 on Build GRCh38
rs779192538 Chr.11: 6239289-6239289 on Build GRCh38
rs909194939 Chr.11: 6239073-6239073 on Build GRCh38
rs959154010 Chr.11: 6239462-6239462 on Build GRCh38
rs1036440197 Chr.11: 6239127-6239127 on Build GRCh38

View Full Product Details