Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 30234699-30235213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHB
Entrez Gene ID
2488
Gene Name
follicle stimulating hormone beta subunit
Gene Aliases
HH24
UniGene
Hs.36975
Gene Chromosome Location
Chr.11: 30231016-30235277 on Build GRCh38
Transcripts
NM_000510.2 NM_001018080.1

SNP Detail

SNP ID Chromosome Location
rs75464895 Chr.11: 30234809-30234809 on Build GRCh38
rs926091092 Chr.11: 30235088-30235088 on Build GRCh38
rs556576879 Chr.11: 30234808-30234808 on Build GRCh38
rs369136961 Chr.11: 30234805-30234805 on Build GRCh38
rs145887479 Chr.11: 30234906-30234906 on Build GRCh38
rs769262752 Chr.11: 30234993-30234993 on Build GRCh38
rs904977105 Chr.11: 30234864-30234864 on Build GRCh38
rs573052973 Chr.11: 30234950-30234950 on Build GRCh38
rs1055015283 Chr.11: 30234863-30234863 on Build GRCh38
rs78946483 Chr.11: 30234748-30234748 on Build GRCh38
rs991878512 Chr.11: 30234781-30234781 on Build GRCh38
rs978890642 Chr.11: 30234737-30234737 on Build GRCh38
rs891970364 Chr.11: 30235176-30235176 on Build GRCh38
rs978943458 Chr.11: 30235065-30235065 on Build GRCh38
rs917818592 Chr.11: 30235038-30235038 on Build GRCh38
rs922639341 Chr.11: 30235128-30235128 on Build GRCh38
rs540432482 Chr.11: 30235137-30235137 on Build GRCh38
rs903584985 Chr.11: 30234838-30234838 on Build GRCh38
rs992487772 Chr.11: 30234996-30234996 on Build GRCh38
rs901166737 Chr.11: 30234782-30234782 on Build GRCh38
rs1029093650 Chr.11: 30234929-30234929 on Build GRCh38
rs1005356676 Chr.11: 30234823-30234823 on Build GRCh38
rs574192474 Chr.11: 30234812-30234812 on Build GRCh38
rs996154182 Chr.11: 30234924-30234924 on Build GRCh38
rs1001899425 Chr.11: 30234876-30234876 on Build GRCh38
rs925059416 Chr.11: 30235107-30235107 on Build GRCh38
rs574242674 Chr.11: 30234768-30234768 on Build GRCh38
rs942796500 Chr.11: 30235099-30235099 on Build GRCh38
rs371525963 Chr.11: 30234942-30234942 on Build GRCh38
rs146658313 Chr.11: 30234794-30234794 on Build GRCh38
rs762424832 Chr.11: 30235108-30235108 on Build GRCh38
rs936203514 Chr.11: 30234848-30234848 on Build GRCh38
rs557783071 Chr.11: 30234943-30234943 on Build GRCh38
rs1010898938 Chr.11: 30234900-30234900 on Build GRCh38
rs775075017 Chr.11: 30235054-30235054 on Build GRCh38
rs1035130463 Chr.11: 30234914-30234914 on Build GRCh38
rs539059758 Chr.11: 30235096-30235098 on Build GRCh38
rs866433090 Chr.11: 30234739-30234739 on Build GRCh38
rs1016701966 Chr.11: 30234836-30234836 on Build GRCh38

View Full Product Details