Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36229428-36229973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD3
Entrez Gene ID
143458
Gene Name
low density lipoprotein receptor class A domain containing 3
Gene Aliases
LRAD3
UniGene
Hs.745134
Gene Chromosome Location
Chr.11: 35943981-36232136 on Build GRCh38
Transcripts
NM_001304263.1 NM_001304264.1 NM_174902.3

SNP Detail

SNP ID Chromosome Location
rs749014752 Chr.11: 36229684-36229684 on Build GRCh38
rs1042689478 Chr.11: 36229683-36229683 on Build GRCh38
rs974454528 Chr.11: 36229554-36229554 on Build GRCh38
rs12793749 Chr.11: 36229806-36229806 on Build GRCh38
rs536738831 Chr.11: 36229730-36229730 on Build GRCh38
rs4755444 Chr.11: 36229496-36229496 on Build GRCh38
rs549996274 Chr.11: 36229468-36229468 on Build GRCh38
rs1044137130 Chr.11: 36229576-36229576 on Build GRCh38
rs940325541 Chr.11: 36229526-36229526 on Build GRCh38
rs531180569 Chr.11: 36229502-36229502 on Build GRCh38
rs111713709 Chr.11: 36229856-36229857 on Build GRCh38
rs780135608 Chr.11: 36229899-36229899 on Build GRCh38
rs945550642 Chr.11: 36229916-36229916 on Build GRCh38
rs920141210 Chr.11: 36229857-36229857 on Build GRCh38
rs192944899 Chr.11: 36229518-36229518 on Build GRCh38
rs60062080 Chr.11: 36229737-36229737 on Build GRCh38
rs1010163341 Chr.11: 36229648-36229648 on Build GRCh38
rs745887198 Chr.11: 36229560-36229560 on Build GRCh38
rs944395376 Chr.11: 36229546-36229546 on Build GRCh38
rs1027690110 Chr.11: 36229782-36229782 on Build GRCh38
rs370738355 Chr.11: 36229541-36229541 on Build GRCh38
rs1033696761 Chr.11: 36229763-36229763 on Build GRCh38
rs563524793 Chr.11: 36229917-36229917 on Build GRCh38
rs184913826 Chr.11: 36229596-36229596 on Build GRCh38
rs151097087 Chr.11: 36229802-36229802 on Build GRCh38
rs745813005 Chr.11: 36229575-36229575 on Build GRCh38
rs572365493 Chr.11: 36229811-36229811 on Build GRCh38
rs532228369 Chr.11: 36229919-36229919 on Build GRCh38
rs560549382 Chr.11: 36229835-36229835 on Build GRCh38
rs373768692 Chr.11: 36229862-36229862 on Build GRCh38
rs540630828 Chr.11: 36229826-36229826 on Build GRCh38
rs935772900 Chr.11: 36229601-36229601 on Build GRCh38
rs945519565 Chr.11: 36229656-36229656 on Build GRCh38
rs556754579 Chr.11: 36229732-36229732 on Build GRCh38
rs1042596217 Chr.11: 36229937-36229937 on Build GRCh38
rs543138211 Chr.11: 36229865-36229865 on Build GRCh38
rs76103857 Chr.11: 36229645-36229645 on Build GRCh38
rs904374941 Chr.11: 36229724-36229724 on Build GRCh38
rs895101960 Chr.11: 36229774-36229774 on Build GRCh38
rs975032288 Chr.11: 36229861-36229861 on Build GRCh38
rs945680672 Chr.11: 36229931-36229931 on Build GRCh38
rs912550353 Chr.11: 36229525-36229525 on Build GRCh38
rs968922879 Chr.11: 36229491-36229491 on Build GRCh38
rs912972227 Chr.11: 36229875-36229875 on Build GRCh38
rs973864742 Chr.11: 36229459-36229459 on Build GRCh38
rs1021493607 Chr.11: 36229479-36229479 on Build GRCh38
rs994972892 Chr.11: 36229779-36229779 on Build GRCh38
rs1002558876 Chr.11: 36229610-36229610 on Build GRCh38
rs920106617 Chr.11: 36229547-36229547 on Build GRCh38
rs954043080 Chr.11: 36229517-36229517 on Build GRCh38
rs1021922814 Chr.11: 36229650-36229650 on Build GRCh38
rs966002791 Chr.11: 36229857-36229857 on Build GRCh38
rs113278126 Chr.11: 36229574-36229574 on Build GRCh38
rs959500867 Chr.11: 36229454-36229454 on Build GRCh38
rs983368878 Chr.11: 36229536-36229536 on Build GRCh38
rs574234653 Chr.11: 36229863-36229863 on Build GRCh38
rs921326186 Chr.11: 36229510-36229510 on Build GRCh38
rs549877231 Chr.11: 36229627-36229627 on Build GRCh38

View Full Product Details