Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44066062-44066565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
UniGene
Hs.126706
Gene Chromosome Location
Chr.11: 44066118-44084237 on Build GRCh38
Transcripts
NM_001127219.1 NM_032592.3 XM_011520410.2

SNP Detail

SNP ID Chromosome Location
rs538898098 Chr.11: 44066358-44066358 on Build GRCh38
rs571468501 Chr.11: 44066318-44066318 on Build GRCh38
rs562453240 Chr.11: 44066248-44066248 on Build GRCh38
rs906797601 Chr.11: 44066408-44066408 on Build GRCh38
rs936318242 Chr.11: 44066344-44066344 on Build GRCh38
rs529557978 Chr.11: 44066096-44066096 on Build GRCh38
rs907330966 Chr.11: 44066231-44066231 on Build GRCh38
rs147180840 Chr.11: 44066178-44066179 on Build GRCh38
rs894891305 Chr.11: 44066356-44066356 on Build GRCh38
rs942375088 Chr.11: 44066087-44066087 on Build GRCh38
rs1026207036 Chr.11: 44066102-44066102 on Build GRCh38
rs184320796 Chr.11: 44066331-44066331 on Build GRCh38
rs933210813 Chr.11: 44066325-44066325 on Build GRCh38
rs967354164 Chr.11: 44066379-44066379 on Build GRCh38
rs963763540 Chr.11: 44066470-44066470 on Build GRCh38
rs549108012 Chr.11: 44066211-44066211 on Build GRCh38
rs1025620656 Chr.11: 44066200-44066200 on Build GRCh38
rs2074038 Chr.11: 44066439-44066439 on Build GRCh38
rs188719261 Chr.11: 44066529-44066529 on Build GRCh38
rs560954815 Chr.11: 44066093-44066093 on Build GRCh38
rs534515410 Chr.11: 44066249-44066249 on Build GRCh38
rs910951540 Chr.11: 44066463-44066463 on Build GRCh38
rs895343842 Chr.11: 44066166-44066166 on Build GRCh38
rs974565108 Chr.11: 44066258-44066258 on Build GRCh38
rs1025215310 Chr.11: 44066173-44066173 on Build GRCh38
rs986677175 Chr.11: 44066455-44066455 on Build GRCh38
rs775722284 Chr.11: 44066097-44066097 on Build GRCh38
rs1012368529 Chr.11: 44066167-44066167 on Build GRCh38
rs763147089 Chr.11: 44066343-44066343 on Build GRCh38
rs898774198 Chr.11: 44066094-44066094 on Build GRCh38
rs1047313594 Chr.11: 44066227-44066227 on Build GRCh38
rs1005961904 Chr.11: 44066492-44066492 on Build GRCh38
rs923728554 Chr.11: 44066188-44066188 on Build GRCh38
rs958949780 Chr.11: 44066122-44066122 on Build GRCh38
rs1042462536 Chr.11: 44066339-44066339 on Build GRCh38
rs888476846 Chr.11: 44066488-44066488 on Build GRCh38
rs1021557511 Chr.11: 44066371-44066371 on Build GRCh38
rs762382246 Chr.11: 44066192-44066192 on Build GRCh38
rs78783230 Chr.11: 44066196-44066196 on Build GRCh38
rs556959626 Chr.11: 44066387-44066387 on Build GRCh38
rs542686527 Chr.11: 44066232-44066232 on Build GRCh38
rs919608167 Chr.11: 44066213-44066213 on Build GRCh38
rs1053508798 Chr.11: 44066354-44066354 on Build GRCh38
rs757119312 Chr.11: 44066100-44066100 on Build GRCh38
rs1034447439 Chr.11: 44066109-44066109 on Build GRCh38
rs907387138 Chr.11: 44066104-44066104 on Build GRCh38
rs878981860 Chr.11: 44066214-44066214 on Build GRCh38
rs572931721 Chr.11: 44066537-44066537 on Build GRCh38
rs1011629223 Chr.11: 44066370-44066370 on Build GRCh38
rs150642270 Chr.11: 44066209-44066209 on Build GRCh38
rs547964157 Chr.11: 44066123-44066123 on Build GRCh38
rs958163378 Chr.11: 44066089-44066089 on Build GRCh38
rs1000594029 Chr.11: 44066163-44066163 on Build GRCh38
rs1018402735 Chr.11: 44066518-44066518 on Build GRCh38
rs554708705 Chr.11: 44066536-44066536 on Build GRCh38
rs1003330946 Chr.11: 44066108-44066108 on Build GRCh38
rs553092040 Chr.11: 44066255-44066255 on Build GRCh38
rs999362713 Chr.11: 44066390-44066390 on Build GRCh38
rs178513 Chr.11: 44066165-44066165 on Build GRCh38
rs911011288 Chr.11: 44066204-44066204 on Build GRCh38
rs769312300 Chr.11: 44066128-44066128 on Build GRCh38
rs894591804 Chr.11: 44066364-44066364 on Build GRCh38
rs766553142 Chr.11: 44066412-44066412 on Build GRCh38
rs955379800 Chr.11: 44066454-44066454 on Build GRCh38
rs551504703 Chr.11: 44066190-44066190 on Build GRCh38
rs559801955 Chr.11: 44066135-44066135 on Build GRCh38
rs967909370 Chr.11: 44066151-44066151 on Build GRCh38

View Full Product Details