Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45224906-45225403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM11
Entrez Gene ID
56981
Gene Name
PR domain 11
Gene Aliases
PFM8
UniGene
Hs.178715
Gene Chromosome Location
Chr.11: 45094013-45235124 on Build GRCh38
Transcripts
NM_001256695.1 NM_001256696.1 NR_046338.1 XM_011520222.2 XM_011520223.2 XM_011520224.2 XM_011520225.2 XM_011520226.2 XM_011520227.2 XM_011520228.2 XM_011520231.2

SNP Detail

SNP ID Chromosome Location
rs1024367293 Chr.11: 45224976-45224976 on Build GRCh38
rs989684598 Chr.11: 45225056-45225056 on Build GRCh38
rs1049781560 Chr.11: 45225139-45225139 on Build GRCh38
rs907948837 Chr.11: 45225082-45225082 on Build GRCh38
rs1010562876 Chr.11: 45225322-45225322 on Build GRCh38
rs969764459 Chr.11: 45225001-45225001 on Build GRCh38
rs1024255531 Chr.11: 45225305-45225305 on Build GRCh38
rs548830700 Chr.11: 45225083-45225083 on Build GRCh38
rs565282569 Chr.11: 45224967-45224967 on Build GRCh38
rs1014174956 Chr.11: 45224961-45224961 on Build GRCh38
rs932217359 Chr.11: 45225197-45225197 on Build GRCh38
rs988560688 Chr.11: 45225367-45225367 on Build GRCh38
rs201762109 Chr.11: 45224933-45224933 on Build GRCh38
rs537409044 Chr.11: 45225348-45225348 on Build GRCh38
rs982363459 Chr.11: 45225003-45225003 on Build GRCh38
rs1052364908 Chr.11: 45225198-45225198 on Build GRCh38
rs935094310 Chr.11: 45225248-45225248 on Build GRCh38
rs771127561 Chr.11: 45224937-45224937 on Build GRCh38
rs1001241556 Chr.11: 45225337-45225337 on Build GRCh38
rs888568991 Chr.11: 45225148-45225148 on Build GRCh38
rs760899507 Chr.11: 45224958-45224958 on Build GRCh38
rs1037233213 Chr.11: 45225184-45225184 on Build GRCh38
rs534565026 Chr.11: 45225237-45225237 on Build GRCh38
rs932681596 Chr.11: 45225137-45225137 on Build GRCh38
rs565861251 Chr.11: 45225217-45225217 on Build GRCh38
rs1005333723 Chr.11: 45225182-45225182 on Build GRCh38
rs549373630 Chr.11: 45224987-45224987 on Build GRCh38
rs764676582 Chr.11: 45224959-45224959 on Build GRCh38
rs775958697 Chr.11: 45224954-45224954 on Build GRCh38
rs983508441 Chr.11: 45225047-45225047 on Build GRCh38
rs1032756886 Chr.11: 45225345-45225345 on Build GRCh38
rs1041315981 Chr.11: 45225095-45225095 on Build GRCh38
rs189968740 Chr.11: 45225249-45225249 on Build GRCh38
rs765610202 Chr.11: 45224938-45224938 on Build GRCh38
rs749380331 Chr.11: 45224934-45224934 on Build GRCh38
rs1023266837 Chr.11: 45225335-45225335 on Build GRCh38
rs925718607 Chr.11: 45225049-45225049 on Build GRCh38
rs3740784 Chr.11: 45225019-45225019 on Build GRCh38
rs535075636 Chr.11: 45225227-45225227 on Build GRCh38
rs755278529 Chr.11: 45225043-45225043 on Build GRCh38
rs927781267 Chr.11: 45225010-45225010 on Build GRCh38
rs112453689 Chr.11: 45224999-45224999 on Build GRCh38
rs714215 Chr.11: 45225318-45225318 on Build GRCh38
rs746082736 Chr.11: 45224950-45224950 on Build GRCh38
rs532718561 Chr.11: 45224985-45224985 on Build GRCh38
rs529003522 Chr.11: 45225054-45225054 on Build GRCh38
rs879039490 Chr.11: 45225232-45225232 on Build GRCh38
rs914083563 Chr.11: 45225057-45225057 on Build GRCh38
rs922209960 Chr.11: 45225146-45225146 on Build GRCh38
rs958191284 Chr.11: 45225050-45225050 on Build GRCh38
rs781242756 Chr.11: 45225116-45225116 on Build GRCh38

View Full Product Details