Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47359612-47360138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPI1
Entrez Gene ID
6688
Gene Name
Spi-1 proto-oncogene
Gene Aliases
OF, PU.1, SFPI1, SPI-1, SPI-A
UniGene
Hs.502511
Gene Chromosome Location
Chr.11: 47354858-47395640 on Build GRCh38
Transcripts
NM_001080547.1 NM_003120.2 XM_011520307.1 XM_017018173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6551987 c.210C>A p.N70K Chr.11: 47359976-47359976 on Build GRCh38
4784572 c.199G>A p.A67T Chr.11: 47359987-47359987 on Build GRCh38
6494047 c.243_245delCAT p.H81_M82>Q Chr.11: 47359920-47359922 on Build GRCh38
4785306 c.303G>A p.M101I Chr.11: 47359883-47359883 on Build GRCh38
6788916 c.291C>T p.L97L Chr.11: 47359895-47359895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867959117 Chr.11: 47360057-47360057 on Build GRCh38
rs959342600 Chr.11: 47359973-47359973 on Build GRCh38
rs901705968 Chr.11: 47360114-47360114 on Build GRCh38
rs772416102 Chr.11: 47359951-47359951 on Build GRCh38
rs943258367 Chr.11: 47359783-47359783 on Build GRCh38
rs149432722 Chr.11: 47359901-47359901 on Build GRCh38
rs185801580 Chr.11: 47359822-47359822 on Build GRCh38
rs775926060 Chr.11: 47359958-47359958 on Build GRCh38
rs542063740 Chr.11: 47359728-47359728 on Build GRCh38
rs368215281 Chr.11: 47359997-47359997 on Build GRCh38
rs372814106 Chr.11: 47360012-47360012 on Build GRCh38
rs772865590 Chr.11: 47360070-47360070 on Build GRCh38
rs564341452 Chr.11: 47360027-47360027 on Build GRCh38
rs769051484 Chr.11: 47359971-47359971 on Build GRCh38
rs576231631 Chr.11: 47359727-47359727 on Build GRCh38
rs546163869 Chr.11: 47359683-47359683 on Build GRCh38
rs779680681 Chr.11: 47359808-47359808 on Build GRCh38
rs181611720 Chr.11: 47359658-47359658 on Build GRCh38
rs745879794 Chr.11: 47360017-47360017 on Build GRCh38
rs931284142 Chr.11: 47359754-47359754 on Build GRCh38
rs138934569 Chr.11: 47359890-47359890 on Build GRCh38
rs556191578 Chr.11: 47359706-47359706 on Build GRCh38
rs1040224335 Chr.11: 47360109-47360109 on Build GRCh38
rs772645546 Chr.11: 47360022-47360022 on Build GRCh38
rs200444347 Chr.11: 47360045-47360045 on Build GRCh38
rs201951592 Chr.11: 47360059-47360059 on Build GRCh38
rs781009883 Chr.11: 47360003-47360003 on Build GRCh38
rs758384599 Chr.11: 47359885-47359885 on Build GRCh38
rs1050233603 Chr.11: 47359856-47359856 on Build GRCh38
rs773349782 Chr.11: 47359823-47359823 on Build GRCh38
rs73453722 Chr.11: 47359635-47359635 on Build GRCh38
rs113825984 Chr.11: 47359987-47359987 on Build GRCh38
rs982239614 Chr.11: 47359649-47359649 on Build GRCh38
rs113583625 Chr.11: 47360111-47360111 on Build GRCh38
rs751249636 Chr.11: 47359895-47359895 on Build GRCh38
rs756375781 Chr.11: 47359900-47359900 on Build GRCh38
rs144812993 Chr.11: 47359946-47359946 on Build GRCh38
rs1040574101 Chr.11: 47359799-47359799 on Build GRCh38
rs760405055 Chr.11: 47359841-47359841 on Build GRCh38
rs1020808352 Chr.11: 47359888-47359888 on Build GRCh38
rs918177259 Chr.11: 47359659-47359659 on Build GRCh38
rs769506463 Chr.11: 47360063-47360063 on Build GRCh38
rs757424163 Chr.11: 47360011-47360011 on Build GRCh38
rs764832542 Chr.11: 47359868-47359868 on Build GRCh38
rs576752137 Chr.11: 47359651-47359651 on Build GRCh38
rs544183724 Chr.11: 47359694-47359696 on Build GRCh38
rs377219165 Chr.11: 47360044-47360044 on Build GRCh38
rs992733487 Chr.11: 47359657-47359657 on Build GRCh38
rs989101604 Chr.11: 47359734-47359734 on Build GRCh38
rs746398377 Chr.11: 47359947-47359947 on Build GRCh38
rs753401693 Chr.11: 47359850-47359850 on Build GRCh38
rs749507809 Chr.11: 47359913-47359913 on Build GRCh38
rs749312250 Chr.11: 47359831-47359831 on Build GRCh38
rs940220556 Chr.11: 47359730-47359730 on Build GRCh38
rs748448119 Chr.11: 47359811-47359811 on Build GRCh38
rs377150823 Chr.11: 47359840-47359840 on Build GRCh38
rs760889389 Chr.11: 47359970-47359970 on Build GRCh38
rs889863613 Chr.11: 47359769-47359769 on Build GRCh38
rs750423772 Chr.11: 47359873-47359873 on Build GRCh38
rs537934199 Chr.11: 47359927-47359927 on Build GRCh38
rs777111732 Chr.11: 47360049-47360049 on Build GRCh38
rs1049866930 Chr.11: 47359763-47359763 on Build GRCh38
rs774905069 Chr.11: 47359834-47359834 on Build GRCh38
rs770989166 Chr.11: 47359832-47359832 on Build GRCh38
rs759618208 Chr.11: 47359839-47359839 on Build GRCh38
rs529405462 Chr.11: 47360110-47360110 on Build GRCh38
rs911314892 Chr.11: 47360089-47360089 on Build GRCh38
rs776489859 Chr.11: 47359985-47359985 on Build GRCh38
rs919902184 Chr.11: 47359742-47359742 on Build GRCh38
rs1037048988 Chr.11: 47359737-47359737 on Build GRCh38
rs992345752 Chr.11: 47359976-47359976 on Build GRCh38
rs762991052 Chr.11: 47359994-47359994 on Build GRCh38
rs745783323 Chr.11: 47359804-47359804 on Build GRCh38
rs61897390 Chr.11: 47360091-47360091 on Build GRCh38
rs762432821 Chr.11: 47360088-47360088 on Build GRCh38
rs767461301 Chr.11: 47359934-47359934 on Build GRCh38
rs749900902 Chr.11: 47359988-47359988 on Build GRCh38
rs766172098 Chr.11: 47359996-47359996 on Build GRCh38
rs915524816 Chr.11: 47359772-47359772 on Build GRCh38
rs761463108 Chr.11: 47359866-47359866 on Build GRCh38
rs1003294395 Chr.11: 47360021-47360021 on Build GRCh38
rs929711247 Chr.11: 47359702-47359702 on Build GRCh38
rs771040203 Chr.11: 47359915-47359915 on Build GRCh38

View Full Product Details