Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56176862-56177368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5J2
Entrez Gene ID
282775
Gene Name
olfactory receptor family 5 subfamily J member 2
Gene Aliases
OR11-266
UniGene
Hs.537145
Gene Chromosome Location
Chr.11: 56176618-56177556 on Build GRCh38
Transcripts
NM_001005492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245692 c.680G>C p.R227T Chr.11: 56177297-56177297 on Build GRCh38
5916096 c.544C>A p.P182T Chr.11: 56177161-56177161 on Build GRCh38
6442352 c.251A>G p.N84S Chr.11: 56176868-56176868 on Build GRCh38
358543 c.405C>A p.A135A Chr.11: 56177022-56177022 on Build GRCh38
7107904 c.365G>T p.R122L Chr.11: 56176982-56176982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536548848 Chr.11: 56177339-56177339 on Build GRCh38
rs764285747 Chr.11: 56176897-56176897 on Build GRCh38
rs760965659 Chr.11: 56176939-56176939 on Build GRCh38
rs762612596 Chr.11: 56177088-56177088 on Build GRCh38
rs747781999 Chr.11: 56176970-56176970 on Build GRCh38
rs753560178 Chr.11: 56177159-56177159 on Build GRCh38
rs759512906 Chr.11: 56177116-56177116 on Build GRCh38
rs373119214 Chr.11: 56176929-56176929 on Build GRCh38
rs779578731 Chr.11: 56176904-56176904 on Build GRCh38
rs142761104 Chr.11: 56177315-56177315 on Build GRCh38
rs760717579 Chr.11: 56177332-56177332 on Build GRCh38
rs143609460 Chr.11: 56177211-56177211 on Build GRCh38
rs373367045 Chr.11: 56177208-56177208 on Build GRCh38
rs199689729 Chr.11: 56177057-56177057 on Build GRCh38
rs553805754 Chr.11: 56176961-56176961 on Build GRCh38
rs745595664 Chr.11: 56177316-56177316 on Build GRCh38
rs755394606 Chr.11: 56177287-56177287 on Build GRCh38
rs961561145 Chr.11: 56177270-56177270 on Build GRCh38
rs145372004 Chr.11: 56176912-56176912 on Build GRCh38
rs757026720 Chr.11: 56176946-56176946 on Build GRCh38
rs941097033 Chr.11: 56177244-56177244 on Build GRCh38
rs779523273 Chr.11: 56176955-56176955 on Build GRCh38
rs754433187 Chr.11: 56177026-56177026 on Build GRCh38
rs780669321 Chr.11: 56177065-56177065 on Build GRCh38
rs148761337 Chr.11: 56176935-56176935 on Build GRCh38
rs762816704 Chr.11: 56177024-56177024 on Build GRCh38
rs770684787 Chr.11: 56177220-56177220 on Build GRCh38
rs764988902 Chr.11: 56176950-56176950 on Build GRCh38
rs965059795 Chr.11: 56176901-56176901 on Build GRCh38
rs758333440 Chr.11: 56176952-56176952 on Build GRCh38
rs762080941 Chr.11: 56177079-56177079 on Build GRCh38
rs542294807 Chr.11: 56176980-56176980 on Build GRCh38
rs766836725 Chr.11: 56177277-56177277 on Build GRCh38
rs1028542088 Chr.11: 56176926-56176926 on Build GRCh38
rs572285751 Chr.11: 56176964-56176964 on Build GRCh38
rs199679290 Chr.11: 56177291-56177291 on Build GRCh38
rs767014132 Chr.11: 56177122-56177122 on Build GRCh38
rs746455332 Chr.11: 56176957-56176957 on Build GRCh38
rs763684226 Chr.11: 56177241-56177241 on Build GRCh38
rs752211231 Chr.11: 56177124-56177124 on Build GRCh38
rs151315491 Chr.11: 56176982-56176982 on Build GRCh38
rs746659808 Chr.11: 56176908-56176908 on Build GRCh38
rs747566544 Chr.11: 56177070-56177070 on Build GRCh38
rs781507139 Chr.11: 56177289-56177289 on Build GRCh38
rs866231331 Chr.11: 56177334-56177334 on Build GRCh38
rs776338136 Chr.11: 56177337-56177337 on Build GRCh38
rs749585315 Chr.11: 56176916-56176916 on Build GRCh38
rs1024595713 Chr.11: 56177039-56177039 on Build GRCh38
rs915845433 Chr.11: 56177340-56177340 on Build GRCh38
rs141314313 Chr.11: 56177031-56177031 on Build GRCh38
rs775502945 Chr.11: 56177237-56177237 on Build GRCh38
rs757900646 Chr.11: 56176899-56176899 on Build GRCh38
rs754686350 Chr.11: 56177175-56177175 on Build GRCh38
rs144285429 Chr.11: 56176896-56176896 on Build GRCh38
rs916057433 Chr.11: 56177256-56177256 on Build GRCh38
rs548534792 Chr.11: 56177323-56177323 on Build GRCh38
rs755600455 Chr.11: 56177203-56177203 on Build GRCh38
rs376086008 Chr.11: 56176930-56176930 on Build GRCh38
rs761221646 Chr.11: 56176973-56176973 on Build GRCh38
rs552271667 Chr.11: 56177213-56177213 on Build GRCh38
rs771307106 Chr.11: 56176971-56176971 on Build GRCh38
rs757035260 Chr.11: 56177272-56177272 on Build GRCh38
rs764356359 Chr.11: 56176942-56176942 on Build GRCh38
rs909570306 Chr.11: 56177242-56177242 on Build GRCh38
rs746385650 Chr.11: 56176975-56176975 on Build GRCh38
rs750241290 Chr.11: 56177005-56177005 on Build GRCh38
rs767303798 Chr.11: 56177232-56177232 on Build GRCh38
rs977070126 Chr.11: 56177217-56177217 on Build GRCh38
rs779189427 Chr.11: 56176974-56176974 on Build GRCh38
rs12279899 Chr.11: 56177025-56177025 on Build GRCh38
rs760890519 Chr.11: 56177341-56177342 on Build GRCh38
rs140282757 Chr.11: 56177303-56177303 on Build GRCh38
rs779326980 Chr.11: 56177046-56177046 on Build GRCh38
rs773483070 Chr.11: 56177086-56177086 on Build GRCh38
rs760064405 Chr.11: 56177240-56177240 on Build GRCh38
rs1055499203 Chr.11: 56177245-56177245 on Build GRCh38
rs771934315 Chr.11: 56177321-56177321 on Build GRCh38
rs753399221 Chr.11: 56177265-56177265 on Build GRCh38
rs750127531 Chr.11: 56177343-56177343 on Build GRCh38
rs761979289 Chr.11: 56176996-56176996 on Build GRCh38
rs530410826 Chr.11: 56177075-56177075 on Build GRCh38
rs903404369 Chr.11: 56177294-56177294 on Build GRCh38
rs868722629 Chr.11: 56177293-56177293 on Build GRCh38
rs776969067 Chr.11: 56176995-56176995 on Build GRCh38
rs767607788 Chr.11: 56176934-56176934 on Build GRCh38
rs535936033 Chr.11: 56177221-56177221 on Build GRCh38
rs4489763 Chr.11: 56176998-56176998 on Build GRCh38
rs201477399 Chr.11: 56177338-56177338 on Build GRCh38
rs984356551 Chr.11: 56177247-56177247 on Build GRCh38
rs143728764 Chr.11: 56177227-56177227 on Build GRCh38
rs756122621 Chr.11: 56177301-56177301 on Build GRCh38
rs142469225 Chr.11: 56176981-56176981 on Build GRCh38
rs771204046 Chr.11: 56177312-56177312 on Build GRCh38
rs751914437 Chr.11: 56177280-56177280 on Build GRCh38
rs147148092 Chr.11: 56177302-56177302 on Build GRCh38
rs763505261 Chr.11: 56177333-56177333 on Build GRCh38
rs570586477 Chr.11: 56177226-56177226 on Build GRCh38
rs568231265 Chr.11: 56177327-56177327 on Build GRCh38
rs747513107 Chr.11: 56177190-56177190 on Build GRCh38
rs148071910 Chr.11: 56177214-56177214 on Build GRCh38
rs548340665 Chr.11: 56177081-56177081 on Build GRCh38
rs751479633 Chr.11: 56177105-56177105 on Build GRCh38
rs149623931 Chr.11: 56176991-56176991 on Build GRCh38
rs758788165 Chr.11: 56177162-56177162 on Build GRCh38
rs138599396 Chr.11: 56177054-56177054 on Build GRCh38
rs1046323612 Chr.11: 56177264-56177264 on Build GRCh38
rs922252564 Chr.11: 56176987-56176987 on Build GRCh38
rs149284707 Chr.11: 56177134-56177134 on Build GRCh38
rs1014908598 Chr.11: 56177038-56177038 on Build GRCh38
rs146115412 Chr.11: 56177032-56177032 on Build GRCh38
rs950287067 Chr.11: 56177263-56177263 on Build GRCh38
rs573804726 Chr.11: 56177044-56177044 on Build GRCh38
rs868322923 Chr.11: 56176909-56176909 on Build GRCh38
rs986236291 Chr.11: 56177110-56177110 on Build GRCh38
rs773308507 Chr.11: 56177145-56177147 on Build GRCh38
rs766179867 Chr.11: 56177103-56177103 on Build GRCh38
rs776641028 Chr.11: 56177077-56177077 on Build GRCh38

View Full Product Details