Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56360848-56361348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8J1
Entrez Gene ID
219477
Gene Name
olfactory receptor family 8 subfamily J member 1
Gene Aliases
OR11-183
UniGene
Hs.626841
Gene Chromosome Location
Chr.11: 56360215-56361288 on Build GRCh38
Transcripts
NM_001005205.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1354631 c.697G>T p.G233* Chr.11: 56360943-56360943 on Build GRCh38
4033882 c.604T>C p.S202P Chr.11: 56360850-56360850 on Build GRCh38
6735377 c.709G>T p.A237S Chr.11: 56360955-56360955 on Build GRCh38
928350 c.766C>A p.L256I Chr.11: 56361012-56361012 on Build GRCh38
3449386 c.702G>A p.R234R Chr.11: 56360948-56360948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7928704 Chr.11: 56361048-56361048 on Build GRCh38
rs373490643 Chr.11: 56361219-56361219 on Build GRCh38
rs373924384 Chr.11: 56360956-56360956 on Build GRCh38
rs776758363 Chr.11: 56361087-56361087 on Build GRCh38
rs747769877 Chr.11: 56360893-56360893 on Build GRCh38
rs747801339 Chr.11: 56360993-56360993 on Build GRCh38
rs144364540 Chr.11: 56361167-56361167 on Build GRCh38
rs1016045427 Chr.11: 56361271-56361271 on Build GRCh38
rs748958270 Chr.11: 56361075-56361075 on Build GRCh38
rs923088808 Chr.11: 56361302-56361302 on Build GRCh38
rs781486467 Chr.11: 56361194-56361194 on Build GRCh38
rs758306578 Chr.11: 56360881-56360881 on Build GRCh38
rs776078174 Chr.11: 56361019-56361019 on Build GRCh38
rs774613851 Chr.11: 56360897-56360897 on Build GRCh38
rs370640996 Chr.11: 56361030-56361030 on Build GRCh38
rs763680308 Chr.11: 56361148-56361148 on Build GRCh38
rs758480984 Chr.11: 56361317-56361318 on Build GRCh38
rs771822660 Chr.11: 56361290-56361290 on Build GRCh38
rs753498218 Chr.11: 56361150-56361150 on Build GRCh38
rs112511179 Chr.11: 56361242-56361242 on Build GRCh38
rs142433814 Chr.11: 56361031-56361031 on Build GRCh38
rs759396655 Chr.11: 56361238-56361238 on Build GRCh38
rs747486541 Chr.11: 56361086-56361086 on Build GRCh38
rs577424821 Chr.11: 56360891-56360891 on Build GRCh38
rs941821150 Chr.11: 56361110-56361110 on Build GRCh38
rs372843205 Chr.11: 56361171-56361171 on Build GRCh38
rs7938038 Chr.11: 56361296-56361296 on Build GRCh38
rs748244503 Chr.11: 56361092-56361092 on Build GRCh38
rs186240912 Chr.11: 56361041-56361041 on Build GRCh38
rs775237310 Chr.11: 56361247-56361247 on Build GRCh38
rs774124424 Chr.11: 56361224-56361224 on Build GRCh38
rs34649252 Chr.11: 56361197-56361198 on Build GRCh38
rs755517175 Chr.11: 56361147-56361147 on Build GRCh38
rs958172569 Chr.11: 56360953-56360953 on Build GRCh38
rs989958112 Chr.11: 56361017-56361017 on Build GRCh38
rs761282770 Chr.11: 56361021-56361021 on Build GRCh38
rs767416211 Chr.11: 56361129-56361129 on Build GRCh38
rs746582070 Chr.11: 56360887-56360887 on Build GRCh38
rs769923091 Chr.11: 56361096-56361096 on Build GRCh38
rs757014604 Chr.11: 56360871-56360871 on Build GRCh38
rs1030034496 Chr.11: 56360890-56360890 on Build GRCh38
rs769374845 Chr.11: 56361002-56361002 on Build GRCh38
rs148720229 Chr.11: 56361182-56361182 on Build GRCh38
rs199725360 Chr.11: 56360896-56360896 on Build GRCh38
rs548575754 Chr.11: 56361260-56361260 on Build GRCh38
rs77431564 Chr.11: 56361320-56361320 on Build GRCh38
rs757385960 Chr.11: 56361001-56361001 on Build GRCh38
rs766125912 Chr.11: 56361123-56361123 on Build GRCh38
rs143337271 Chr.11: 56361034-56361034 on Build GRCh38
rs754427336 Chr.11: 56360966-56360966 on Build GRCh38
rs919216713 Chr.11: 56361008-56361008 on Build GRCh38
rs755781753 Chr.11: 56361065-56361065 on Build GRCh38
rs761951424 Chr.11: 56361050-56361050 on Build GRCh38
rs780542668 Chr.11: 56361165-56361165 on Build GRCh38
rs547343412 Chr.11: 56361195-56361195 on Build GRCh38
rs201154227 Chr.11: 56360935-56360935 on Build GRCh38
rs772567105 Chr.11: 56361012-56361012 on Build GRCh38
rs868011806 Chr.11: 56361046-56361046 on Build GRCh38
rs757008828 Chr.11: 56361159-56361159 on Build GRCh38
rs936881197 Chr.11: 56361161-56361161 on Build GRCh38
rs149645647 Chr.11: 56361145-56361145 on Build GRCh38
rs868123171 Chr.11: 56361109-56361109 on Build GRCh38
rs537599009 Chr.11: 56361300-56361300 on Build GRCh38
rs375781706 Chr.11: 56360957-56360957 on Build GRCh38
rs745778425 Chr.11: 56360877-56360877 on Build GRCh38
rs780756932 Chr.11: 56360892-56360892 on Build GRCh38
rs199894933 Chr.11: 56361220-56361220 on Build GRCh38
rs760911965 Chr.11: 56361287-56361287 on Build GRCh38
rs56266017 Chr.11: 56361201-56361202 on Build GRCh38
rs970934144 Chr.11: 56360889-56360889 on Build GRCh38
rs758783993 Chr.11: 56361076-56361076 on Build GRCh38
rs1011032909 Chr.11: 56361262-56361262 on Build GRCh38
rs771388497 Chr.11: 56361166-56361167 on Build GRCh38
rs140351903 Chr.11: 56361099-56361099 on Build GRCh38
rs760767327 Chr.11: 56360931-56360931 on Build GRCh38
rs374279068 Chr.11: 56361033-56361033 on Build GRCh38
rs779515422 Chr.11: 56360884-56360884 on Build GRCh38
rs536283757 Chr.11: 56361249-56361249 on Build GRCh38
rs780725965 Chr.11: 56361055-56361055 on Build GRCh38
rs763186669 Chr.11: 56361114-56361114 on Build GRCh38
rs1012428227 Chr.11: 56361174-56361174 on Build GRCh38
rs769715571 Chr.11: 56361202-56361202 on Build GRCh38
rs888835355 Chr.11: 56361201-56361201 on Build GRCh38
rs778594068 Chr.11: 56360876-56360876 on Build GRCh38
rs777149289 Chr.11: 56361167-56361168 on Build GRCh38
rs200839909 Chr.11: 56361218-56361218 on Build GRCh38
rs376550127 Chr.11: 56361007-56361007 on Build GRCh38
rs759451788 Chr.11: 56361128-56361128 on Build GRCh38
rs750232856 Chr.11: 56360955-56360955 on Build GRCh38
rs1025232247 Chr.11: 56360875-56360875 on Build GRCh38
rs761580073 Chr.11: 56361131-56361131 on Build GRCh38
rs886088463 Chr.11: 56361101-56361101 on Build GRCh38
rs780593894 Chr.11: 56361084-56361084 on Build GRCh38
rs768815456 Chr.11: 56361025-56361025 on Build GRCh38
rs1016523854 Chr.11: 56360915-56360915 on Build GRCh38
rs370213627 Chr.11: 56360943-56360943 on Build GRCh38
rs963536292 Chr.11: 56361289-56361289 on Build GRCh38
rs754943666 Chr.11: 56361053-56361053 on Build GRCh38
rs760695563 Chr.11: 56361248-56361248 on Build GRCh38
rs201063906 Chr.11: 56360908-56360908 on Build GRCh38
rs780705942 Chr.11: 56360992-56360992 on Build GRCh38
rs777219973 Chr.11: 56361066-56361066 on Build GRCh38
rs62001032 Chr.11: 56360930-56360930 on Build GRCh38
rs1031031204 Chr.11: 56361234-56361234 on Build GRCh38
rs146758768 Chr.11: 56361057-56361057 on Build GRCh38
rs748753596 Chr.11: 56361029-56361029 on Build GRCh38
rs764278726 Chr.11: 56360933-56360933 on Build GRCh38
rs760059517 Chr.11: 56360898-56360898 on Build GRCh38
rs761972886 Chr.11: 56360941-56360941 on Build GRCh38
rs759851747 Chr.11: 56361190-56361190 on Build GRCh38
rs867854319 Chr.11: 56361155-56361155 on Build GRCh38
rs7938865 Chr.11: 56361299-56361299 on Build GRCh38
rs564005808 Chr.11: 56361115-56361115 on Build GRCh38

View Full Product Details