Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57597990-57598484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPING1
Entrez Gene ID
710
Gene Name
serpin family G member 1
Gene Aliases
C1IN, C1INH, C1NH, HAE1, HAE2
UniGene
Hs.384598
Gene Chromosome Location
Chr.11: 57597554-57614853 on Build GRCh38
Transcripts
NM_000062.2 NM_001032295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7277438 c.33G>C p.L11L Chr.11: 57598303-57598303 on Build GRCh38
4574490 c.31_33delCTG p.L15delL Chr.11: 57598301-57598303 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758650443 Chr.11: 57598400-57598400 on Build GRCh38
rs778873658 Chr.11: 57598133-57598133 on Build GRCh38
rs532231419 Chr.11: 57598070-57598070 on Build GRCh38
rs900603288 Chr.11: 57598175-57598175 on Build GRCh38
rs554852233 Chr.11: 57598379-57598379 on Build GRCh38
rs919177783 Chr.11: 57598434-57598434 on Build GRCh38
rs967418951 Chr.11: 57598096-57598096 on Build GRCh38
rs199473715 Chr.11: 57598321-57598321 on Build GRCh38
rs766182685 Chr.11: 57598244-57598244 on Build GRCh38
rs765774755 Chr.11: 57598290-57598290 on Build GRCh38
rs780220125 Chr.11: 57598166-57598166 on Build GRCh38
rs768364958 Chr.11: 57598183-57598183 on Build GRCh38
rs1024704974 Chr.11: 57598089-57598089 on Build GRCh38
rs919196001 Chr.11: 57598187-57598187 on Build GRCh38
rs759225698 Chr.11: 57598254-57598254 on Build GRCh38
rs28362942 Chr.11: 57598037-57598037 on Build GRCh38
rs780903287 Chr.11: 57598352-57598352 on Build GRCh38
rs28362945 Chr.11: 57598422-57598422 on Build GRCh38
rs747100943 Chr.11: 57598446-57598446 on Build GRCh38
rs772664522 Chr.11: 57598236-57598236 on Build GRCh38
rs762519399 Chr.11: 57598238-57598238 on Build GRCh38
rs752539201 Chr.11: 57598205-57598206 on Build GRCh38
rs28362943 Chr.11: 57598160-57598160 on Build GRCh38
rs1048798650 Chr.11: 57598191-57598191 on Build GRCh38
rs867235086 Chr.11: 57598118-57598118 on Build GRCh38
rs201455616 Chr.11: 57598295-57598295 on Build GRCh38
rs887549942 Chr.11: 57598056-57598056 on Build GRCh38
rs754174303 Chr.11: 57598269-57598269 on Build GRCh38
rs28362944 Chr.11: 57598250-57598250 on Build GRCh38
rs758453439 Chr.11: 57598302-57598302 on Build GRCh38
rs182197371 Chr.11: 57598218-57598218 on Build GRCh38
rs908954589 Chr.11: 57598199-57598199 on Build GRCh38
rs867406954 Chr.11: 57598317-57598317 on Build GRCh38
rs1055933809 Chr.11: 57598205-57598205 on Build GRCh38
rs979921873 Chr.11: 57598016-57598016 on Build GRCh38
rs1028080362 Chr.11: 57598241-57598241 on Build GRCh38
rs890700546 Chr.11: 57598141-57598141 on Build GRCh38
rs1005720526 Chr.11: 57598057-57598057 on Build GRCh38
rs910386251 Chr.11: 57598144-57598144 on Build GRCh38
rs999292918 Chr.11: 57598110-57598110 on Build GRCh38
rs927057007 Chr.11: 57598026-57598026 on Build GRCh38
rs757716611 Chr.11: 57598131-57598131 on Build GRCh38
rs745938839 Chr.11: 57598149-57598149 on Build GRCh38
rs957482992 Chr.11: 57598125-57598125 on Build GRCh38
rs957360903 Chr.11: 57598345-57598345 on Build GRCh38
rs995335918 Chr.11: 57598463-57598463 on Build GRCh38
rs201088164 Chr.11: 57598291-57598291 on Build GRCh38
rs867100870 Chr.11: 57598316-57598316 on Build GRCh38
rs1014893726 Chr.11: 57598231-57598231 on Build GRCh38
rs769753632 Chr.11: 57598220-57598220 on Build GRCh38
rs900637150 Chr.11: 57598444-57598444 on Build GRCh38
rs912891412 Chr.11: 57598351-57598351 on Build GRCh38
rs548528069 Chr.11: 57598185-57598185 on Build GRCh38
rs937752655 Chr.11: 57598201-57598201 on Build GRCh38
rs565866483 Chr.11: 57598103-57598103 on Build GRCh38
rs569933712 Chr.11: 57598186-57598186 on Build GRCh38
rs966331098 Chr.11: 57598159-57598159 on Build GRCh38
rs1007614693 Chr.11: 57598374-57598374 on Build GRCh38
rs572325752 Chr.11: 57598329-57598329 on Build GRCh38
rs185342631 Chr.11: 57598275-57598275 on Build GRCh38
rs1056008556 Chr.11: 57598064-57598064 on Build GRCh38
rs199873764 Chr.11: 57598349-57598349 on Build GRCh38
rs746858240 Chr.11: 57598176-57598176 on Build GRCh38
rs747969766 Chr.11: 57598354-57598354 on Build GRCh38
rs1054546604 Chr.11: 57598058-57598058 on Build GRCh38
rs183507388 Chr.11: 57598076-57598076 on Build GRCh38
rs764371613 Chr.11: 57598265-57598265 on Build GRCh38
rs1019516520 Chr.11: 57598311-57598312 on Build GRCh38
rs758086092 Chr.11: 57598301-57598303 on Build GRCh38
rs927125149 Chr.11: 57598315-57598320 on Build GRCh38
rs976343932 Chr.11: 57598433-57598433 on Build GRCh38
rs1014512262 Chr.11: 57598082-57598082 on Build GRCh38
rs776417611 Chr.11: 57598188-57598188 on Build GRCh38

View Full Product Details