Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57743890-57744397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD18
Entrez Gene ID
643376
Gene Name
BTB domain containing 18
Gene Aliases
-
UniGene
Hs.606355
Gene Chromosome Location
Chr.11: 57743514-57753176 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4841085 c.2028G>T p.E676D Chr.11: 57744245-57744245 on Build GRCh38
5739245 c.2087T>C p.V696A Chr.11: 57744186-57744186 on Build GRCh38
4638122 c.1905G>A p.E635E Chr.11: 57744368-57744368 on Build GRCh38
1152358 c.2065G>T p.V689L Chr.11: 57744208-57744208 on Build GRCh38
3748531 c.1927A>T p.T643S Chr.11: 57744346-57744346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751493867 Chr.11: 57744368-57744368 on Build GRCh38
rs989275426 Chr.11: 57744273-57744273 on Build GRCh38
rs776796813 Chr.11: 57744233-57744233 on Build GRCh38
rs565850042 Chr.11: 57744292-57744292 on Build GRCh38
rs548447040 Chr.11: 57744356-57744356 on Build GRCh38
rs117734748 Chr.11: 57744029-57744029 on Build GRCh38
rs1053131522 Chr.11: 57743958-57743958 on Build GRCh38
rs750275119 Chr.11: 57744346-57744346 on Build GRCh38
rs535021475 Chr.11: 57744203-57744203 on Build GRCh38
rs994284166 Chr.11: 57744089-57744089 on Build GRCh38
rs889641370 Chr.11: 57744025-57744025 on Build GRCh38
rs547628240 Chr.11: 57744117-57744117 on Build GRCh38
rs896911620 Chr.11: 57744080-57744080 on Build GRCh38
rs769270935 Chr.11: 57744129-57744129 on Build GRCh38
rs960333170 Chr.11: 57744118-57744118 on Build GRCh38
rs774798950 Chr.11: 57744132-57744132 on Build GRCh38
rs987787781 Chr.11: 57744188-57744188 on Build GRCh38
rs536181991 Chr.11: 57744332-57744332 on Build GRCh38
rs933976213 Chr.11: 57744288-57744288 on Build GRCh38
rs564969764 Chr.11: 57744067-57744067 on Build GRCh38
rs770747825 Chr.11: 57743946-57743949 on Build GRCh38
rs756478722 Chr.11: 57744058-57744058 on Build GRCh38
rs932265301 Chr.11: 57743980-57743980 on Build GRCh38
rs780342057 Chr.11: 57744100-57744100 on Build GRCh38
rs772670005 Chr.11: 57744176-57744176 on Build GRCh38
rs1049673631 Chr.11: 57744331-57744331 on Build GRCh38
rs1038562380 Chr.11: 57744043-57744043 on Build GRCh38
rs1023499606 Chr.11: 57744096-57744096 on Build GRCh38
rs370927769 Chr.11: 57743969-57743969 on Build GRCh38
rs994913483 Chr.11: 57744079-57744079 on Build GRCh38
rs531321522 Chr.11: 57744011-57744011 on Build GRCh38
rs1049620958 Chr.11: 57744018-57744018 on Build GRCh38
rs1014542324 Chr.11: 57744121-57744121 on Build GRCh38
rs765180887 Chr.11: 57744245-57744245 on Build GRCh38
rs868371204 Chr.11: 57744245-57744250 on Build GRCh38
rs370477173 Chr.11: 57744170-57744170 on Build GRCh38
rs747801969 Chr.11: 57744126-57744126 on Build GRCh38
rs558312760 Chr.11: 57744347-57744347 on Build GRCh38
rs975120142 Chr.11: 57744277-57744277 on Build GRCh38
rs1034918311 Chr.11: 57744159-57744159 on Build GRCh38
rs367567782 Chr.11: 57744365-57744365 on Build GRCh38
rs764278629 Chr.11: 57744213-57744213 on Build GRCh38
rs941241719 Chr.11: 57744363-57744363 on Build GRCh38
rs760908244 Chr.11: 57744205-57744205 on Build GRCh38
rs1003481661 Chr.11: 57744157-57744157 on Build GRCh38
rs890465822 Chr.11: 57743996-57743996 on Build GRCh38
rs956583066 Chr.11: 57744166-57744166 on Build GRCh38
rs947545452 Chr.11: 57743950-57743950 on Build GRCh38
rs532204685 Chr.11: 57744082-57744082 on Build GRCh38
rs867714864 Chr.11: 57744217-57744217 on Build GRCh38
rs914964934 Chr.11: 57743942-57743942 on Build GRCh38
rs978940012 Chr.11: 57743961-57743961 on Build GRCh38
rs1051941332 Chr.11: 57743986-57743986 on Build GRCh38
rs899284490 Chr.11: 57744051-57744051 on Build GRCh38
rs956149466 Chr.11: 57744249-57744249 on Build GRCh38

View Full Product Details