Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61956792-61957303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST1
Entrez Gene ID
7439
Gene Name
bestrophin 1
Gene Aliases
ARB, BEST, BMD, RP50, TU15B, VMD2
UniGene
Hs.524910
Gene Chromosome Location
Chr.11: 61946722-61964465 on Build GRCh38
Transcripts
NM_001139443.1 NM_001300786.1 NM_001300787.1 NM_004183.3 NR_134580.1 XM_005274210.3 XM_005274215.3 XM_005274216.3 XM_005274219.3 XM_005274221.3 XM_011545229.2 XM_011545230.2 XM_017018228.1 XM_017018229.1 XM_017018230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4574560 c.404C>T p.A135V Chr.11: 61956946-61956946 on Build GRCh38
3676245 c.456+1G>C p.? Chr.11: 61956999-61956999 on Build GRCh38
5786960 c.412G>A p.G138R Chr.11: 61956954-61956954 on Build GRCh38
3676243 c.456+1G>A p.? Chr.11: 61956999-61956999 on Build GRCh38
4574561 c.584C>T p.A195V Chr.11: 61956946-61956946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752607915 Chr.11: 61956877-61956877 on Build GRCh38
rs199529046 Chr.11: 61956964-61956964 on Build GRCh38
rs766085130 Chr.11: 61956841-61956841 on Build GRCh38
rs202048573 Chr.11: 61956823-61956823 on Build GRCh38
rs757428952 Chr.11: 61957007-61957007 on Build GRCh38
rs144663679 Chr.11: 61957155-61957155 on Build GRCh38
rs62637337 Chr.11: 61956999-61956999 on Build GRCh38
rs768885579 Chr.11: 61957104-61957104 on Build GRCh38
rs763222021 Chr.11: 61957012-61957013 on Build GRCh38
rs267606680 Chr.11: 61956976-61956976 on Build GRCh38
rs760306 Chr.11: 61956820-61956820 on Build GRCh38
rs121918286 Chr.11: 61956960-61956960 on Build GRCh38
rs74653691 Chr.11: 61956981-61956981 on Build GRCh38
rs753066086 Chr.11: 61957029-61957030 on Build GRCh38
rs201010315 Chr.11: 61956829-61956829 on Build GRCh38
rs973117160 Chr.11: 61957103-61957103 on Build GRCh38
rs998252744 Chr.11: 61957176-61957176 on Build GRCh38
rs368770776 Chr.11: 61957036-61957036 on Build GRCh38
rs771276800 Chr.11: 61956926-61956926 on Build GRCh38
rs913644904 Chr.11: 61957030-61957030 on Build GRCh38
rs917475338 Chr.11: 61957223-61957223 on Build GRCh38
rs62641693 Chr.11: 61956980-61956980 on Build GRCh38
rs138552020 Chr.11: 61957242-61957242 on Build GRCh38
rs989465282 Chr.11: 61957264-61957264 on Build GRCh38
rs758380277 Chr.11: 61956990-61956990 on Build GRCh38
rs765998048 Chr.11: 61956966-61956966 on Build GRCh38
rs557643704 Chr.11: 61957027-61957027 on Build GRCh38
rs753652930 Chr.11: 61956855-61956855 on Build GRCh38
rs996005390 Chr.11: 61957032-61957032 on Build GRCh38
rs386754168 Chr.11: 61956980-61956981 on Build GRCh38
rs182941675 Chr.11: 61956857-61956857 on Build GRCh38
rs927811985 Chr.11: 61957066-61957066 on Build GRCh38
rs532866226 Chr.11: 61956939-61956939 on Build GRCh38
rs150247275 Chr.11: 61956986-61956986 on Build GRCh38
rs1005976439 Chr.11: 61957118-61957118 on Build GRCh38
rs574089424 Chr.11: 61957044-61957044 on Build GRCh38
rs534281398 Chr.11: 61957038-61957038 on Build GRCh38
rs777978717 Chr.11: 61956908-61956908 on Build GRCh38
rs764196815 Chr.11: 61956998-61956998 on Build GRCh38
rs202092275 Chr.11: 61957033-61957033 on Build GRCh38
rs200277476 Chr.11: 61956946-61956946 on Build GRCh38
rs281865237 Chr.11: 61956988-61956988 on Build GRCh38
rs543002966 Chr.11: 61957058-61957058 on Build GRCh38
rs1006982767 Chr.11: 61957231-61957231 on Build GRCh38
rs889969375 Chr.11: 61957208-61957208 on Build GRCh38
rs1037072548 Chr.11: 61957146-61957146 on Build GRCh38
rs781412251 Chr.11: 61957025-61957025 on Build GRCh38
rs195161 Chr.11: 61957111-61957111 on Build GRCh38
rs957604837 Chr.11: 61957250-61957250 on Build GRCh38
rs576635161 Chr.11: 61957142-61957142 on Build GRCh38
rs758504759 Chr.11: 61956899-61956899 on Build GRCh38
rs746317719 Chr.11: 61956833-61956833 on Build GRCh38
rs769188077 Chr.11: 61956961-61956961 on Build GRCh38
rs753437612 Chr.11: 61956967-61956967 on Build GRCh38
rs775979290 Chr.11: 61956895-61956897 on Build GRCh38
rs12225132 Chr.11: 61957165-61957165 on Build GRCh38
rs776011957 Chr.11: 61956947-61956947 on Build GRCh38
rs748477131 Chr.11: 61957159-61957159 on Build GRCh38
rs191220756 Chr.11: 61957249-61957249 on Build GRCh38
rs186544610 Chr.11: 61956870-61956870 on Build GRCh38
rs195162 Chr.11: 61957042-61957042 on Build GRCh38
rs937810251 Chr.11: 61957075-61957075 on Build GRCh38
rs75769763 Chr.11: 61957179-61957179 on Build GRCh38
rs546228761 Chr.11: 61956944-61956944 on Build GRCh38
rs57846215 Chr.11: 61957026-61957026 on Build GRCh38

View Full Product Details