Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73940786-73941102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM161B
Entrez Gene ID
145483
Gene Name
family with sequence similarity 161 member B
Gene Aliases
C14orf44, c14_5547
UniGene
Hs.550547
Gene Chromosome Location
Chr.14: 73932031-73950414 on Build GRCh38
Transcripts
NM_152445.2 XM_011536475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5980642 c.1468C>T p.P490S Chr.14: 73941047-73941047 on Build GRCh38
4887968 c.1469C>A p.P490Q Chr.14: 73941046-73941046 on Build GRCh38
1493301 c.1373C>T p.A458V Chr.14: 73940953-73940953 on Build GRCh38
4578008 c.1550A>G p.H517R Chr.14: 73940965-73940965 on Build GRCh38
5980643 c.1279C>T p.P427S Chr.14: 73941047-73941047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368631236 Chr.14: 73941003-73941003 on Build GRCh38
rs985654268 Chr.14: 73940917-73940917 on Build GRCh38
rs752547544 Chr.14: 73940937-73940937 on Build GRCh38
rs753326803 Chr.14: 73940919-73940919 on Build GRCh38
rs149451151 Chr.14: 73940935-73940935 on Build GRCh38
rs749460073 Chr.14: 73941002-73941002 on Build GRCh38
rs756272650 Chr.14: 73941075-73941075 on Build GRCh38
rs190674916 Chr.14: 73940820-73940820 on Build GRCh38
rs763457868 Chr.14: 73940952-73940953 on Build GRCh38
rs753391393 Chr.14: 73940956-73940956 on Build GRCh38
rs915887642 Chr.14: 73940907-73940907 on Build GRCh38
rs545669063 Chr.14: 73940838-73940839 on Build GRCh38
rs979280272 Chr.14: 73940977-73940977 on Build GRCh38
rs762474039 Chr.14: 73941036-73941036 on Build GRCh38
rs759037058 Chr.14: 73941027-73941027 on Build GRCh38
rs771133695 Chr.14: 73941024-73941024 on Build GRCh38
rs781379472 Chr.14: 73940971-73940971 on Build GRCh38
rs1004907199 Chr.14: 73940842-73940842 on Build GRCh38
rs756471049 Chr.14: 73940980-73940980 on Build GRCh38
rs751547856 Chr.14: 73940965-73940965 on Build GRCh38
rs762156975 Chr.14: 73940906-73940906 on Build GRCh38
rs978835367 Chr.14: 73940946-73940946 on Build GRCh38
rs763969761 Chr.14: 73941038-73941038 on Build GRCh38
rs968811384 Chr.14: 73940921-73940921 on Build GRCh38
rs748742144 Chr.14: 73941008-73941008 on Build GRCh38
rs79402303 Chr.14: 73940901-73940901 on Build GRCh38
rs541959659 Chr.14: 73941057-73941057 on Build GRCh38
rs1051992392 Chr.14: 73941010-73941010 on Build GRCh38
rs776037158 Chr.14: 73940875-73940875 on Build GRCh38
rs780213346 Chr.14: 73941076-73941076 on Build GRCh38
rs910880688 Chr.14: 73940948-73940948 on Build GRCh38
rs758203402 Chr.14: 73940951-73940951 on Build GRCh38
rs772430119 Chr.14: 73941013-73941013 on Build GRCh38
rs372074550 Chr.14: 73940902-73940902 on Build GRCh38
rs762185650 Chr.14: 73940973-73940974 on Build GRCh38
rs201171936 Chr.14: 73941031-73941031 on Build GRCh38
rs374244820 Chr.14: 73940912-73940912 on Build GRCh38
rs750585356 Chr.14: 73941069-73941069 on Build GRCh38
rs1035955743 Chr.14: 73940872-73940872 on Build GRCh38
rs911830052 Chr.14: 73941056-73941056 on Build GRCh38
rs960531526 Chr.14: 73940879-73940879 on Build GRCh38
rs751288370 Chr.14: 73941039-73941039 on Build GRCh38
rs759230992 Chr.14: 73940931-73940931 on Build GRCh38
rs765795373 Chr.14: 73941022-73941026 on Build GRCh38
rs143909733 Chr.14: 73941007-73941007 on Build GRCh38
rs780406073 Chr.14: 73940990-73940990 on Build GRCh38
rs1038704528 Chr.14: 73941071-73941071 on Build GRCh38
rs777366901 Chr.14: 73940963-73940963 on Build GRCh38
rs776194396 Chr.14: 73941016-73941016 on Build GRCh38
rs757258224 Chr.14: 73941049-73941049 on Build GRCh38
rs951578031 Chr.14: 73940819-73940819 on Build GRCh38
rs745911799 Chr.14: 73940978-73940978 on Build GRCh38
rs757092869 Chr.14: 73940969-73940969 on Build GRCh38

View Full Product Details