Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201620858-201621197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM237
Entrez Gene ID
65062
Gene Name
transmembrane protein 237
Gene Aliases
ALS2CR4, JBTS14
UniGene
Hs.12319
Gene Chromosome Location
Chr.2: 201620184-201643529 on Build GRCh38
Transcripts
NM_001044385.2 NM_152388.3

SNP Detail

SNP ID Chromosome Location
rs79378497 Chr.2: 201621131-201621131 on Build GRCh38
rs561312621 Chr.2: 201620905-201620905 on Build GRCh38
rs546598867 Chr.2: 201621017-201621017 on Build GRCh38
rs530012065 Chr.2: 201621088-201621088 on Build GRCh38
rs900022106 Chr.2: 201620966-201620966 on Build GRCh38
rs898567191 Chr.2: 201621130-201621130 on Build GRCh38
rs1016943095 Chr.2: 201621124-201621124 on Build GRCh38
rs951725039 Chr.2: 201620881-201620881 on Build GRCh38
rs1051259298 Chr.2: 201621024-201621024 on Build GRCh38
rs754950149 Chr.2: 201621172-201621172 on Build GRCh38
rs947776588 Chr.2: 201620920-201620920 on Build GRCh38
rs779803604 Chr.2: 201620962-201620962 on Build GRCh38
rs758241557 Chr.2: 201620896-201620896 on Build GRCh38
rs943170007 Chr.2: 201620935-201620935 on Build GRCh38
rs183590415 Chr.2: 201621161-201621161 on Build GRCh38
rs927758866 Chr.2: 201620970-201620970 on Build GRCh38
rs914274527 Chr.2: 201620925-201620925 on Build GRCh38
rs939181432 Chr.2: 201621093-201621093 on Build GRCh38
rs980470875 Chr.2: 201620957-201620957 on Build GRCh38
rs192214909 Chr.2: 201620883-201620883 on Build GRCh38
rs971282536 Chr.2: 201621159-201621159 on Build GRCh38
rs1040276382 Chr.2: 201620958-201620958 on Build GRCh38
rs887380242 Chr.2: 201621040-201621040 on Build GRCh38
rs528512481 Chr.2: 201620945-201620945 on Build GRCh38
rs866645983 Chr.2: 201621132-201621132 on Build GRCh38
rs562558327 Chr.2: 201620984-201620984 on Build GRCh38
rs565592584 Chr.2: 201621121-201621121 on Build GRCh38

View Full Product Details