Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 213363626-213364210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPAG16
Entrez Gene ID
79582
Gene Name
sperm associated antigen 16
Gene Aliases
PF20, WDR29
UniGene
Hs.743365
Gene Chromosome Location
Chr.2: 213284369-214410501 on Build GRCh38
Transcripts
NM_024532.4 NR_047659.1 NR_047660.1 XM_006712746.2 XM_011511814.1 XM_011511815.1 XM_011511816.2 XM_011511817.1 XM_011511818.1 XM_011511819.2 XM_011511820.1 XM_011511821.2 XM_011511822.1 XM_011511823.2 XM_011511824.1 XM_011511827.1 XM_011511828.1 XM_011511829.1 XM_011511831.1 XM_011511832.1 XM_011511835.2 XM_011511837.2 XM_017004896.1 XM_017004897.1 XM_017004898.1 XM_017004899.1 XM_017004900.1 XM_017004901.1 XM_017004902.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5826161 c.704A>G p.Q235R Chr.2: 213364110-213364110 on Build GRCh38
5065287 c.797A>G p.Q266R Chr.2: 213364110-213364110 on Build GRCh38
1565105 c.797A>G p.Q266R Chr.2: 213364110-213364110 on Build GRCh38
3909521 c.802G>A p.G268R Chr.2: 213364115-213364115 on Build GRCh38
81758 c.819T>C p.G273G Chr.2: 213364132-213364132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056001114 Chr.2: 213363812-213363812 on Build GRCh38
rs376987409 Chr.2: 213364183-213364183 on Build GRCh38
rs988148632 Chr.2: 213363858-213363858 on Build GRCh38
rs766813751 Chr.2: 213364089-213364089 on Build GRCh38
rs534677884 Chr.2: 213363653-213363653 on Build GRCh38
rs752908640 Chr.2: 213364108-213364108 on Build GRCh38
rs971047365 Chr.2: 213364039-213364039 on Build GRCh38
rs578039911 Chr.2: 213363706-213363706 on Build GRCh38
rs949126170 Chr.2: 213364131-213364131 on Build GRCh38
rs915562766 Chr.2: 213363716-213363716 on Build GRCh38
rs886064985 Chr.2: 213364180-213364180 on Build GRCh38
rs1029497138 Chr.2: 213363843-213363843 on Build GRCh38
rs549322658 Chr.2: 213363808-213363808 on Build GRCh38
rs761506627 Chr.2: 213364059-213364059 on Build GRCh38
rs762795433 Chr.2: 213363791-213363791 on Build GRCh38
rs998078650 Chr.2: 213363840-213363840 on Build GRCh38
rs539471307 Chr.2: 213363972-213363972 on Build GRCh38
rs188182208 Chr.2: 213363832-213363832 on Build GRCh38
rs563806725 Chr.2: 213363989-213363989 on Build GRCh38
rs919542165 Chr.2: 213364096-213364096 on Build GRCh38
rs913207299 Chr.2: 213364008-213364008 on Build GRCh38
rs560496597 Chr.2: 213363720-213363720 on Build GRCh38
rs937636862 Chr.2: 213364066-213364066 on Build GRCh38
rs545281255 Chr.2: 213363705-213363705 on Build GRCh38
rs79246157 Chr.2: 213363867-213363867 on Build GRCh38
rs866537068 Chr.2: 213364091-213364091 on Build GRCh38
rs12469417 Chr.2: 213364068-213364068 on Build GRCh38
rs913259742 Chr.2: 213363692-213363692 on Build GRCh38
rs761174763 Chr.2: 213364088-213364088 on Build GRCh38
rs957372486 Chr.2: 213363965-213363965 on Build GRCh38
rs544032310 Chr.2: 213363961-213363962 on Build GRCh38
rs967360598 Chr.2: 213364021-213364021 on Build GRCh38
rs184537297 Chr.2: 213363689-213363689 on Build GRCh38
rs773917559 Chr.2: 213364049-213364049 on Build GRCh38
rs937503195 Chr.2: 213363804-213363804 on Build GRCh38
rs758594176 Chr.2: 213364111-213364111 on Build GRCh38
rs976682306 Chr.2: 213363679-213363679 on Build GRCh38
rs963367400 Chr.2: 213363941-213363941 on Build GRCh38
rs954852439 Chr.2: 213363657-213363657 on Build GRCh38
rs749221128 Chr.2: 213364162-213364162 on Build GRCh38
rs901813016 Chr.2: 213363835-213363835 on Build GRCh38
rs954940286 Chr.2: 213364005-213364005 on Build GRCh38
rs759600418 Chr.2: 213364097-213364097 on Build GRCh38
rs1045990589 Chr.2: 213363721-213363721 on Build GRCh38
rs926189299 Chr.2: 213364056-213364056 on Build GRCh38
rs886117487 Chr.2: 213363911-213363911 on Build GRCh38
rs991602402 Chr.2: 213363710-213363710 on Build GRCh38
rs534993770 Chr.2: 213364119-213364119 on Build GRCh38
rs964327093 Chr.2: 213363673-213363673 on Build GRCh38
rs374194517 Chr.2: 213363991-213363991 on Build GRCh38
rs926090373 Chr.2: 213363740-213363740 on Build GRCh38
rs527877415 Chr.2: 213363752-213363752 on Build GRCh38
rs574124178 Chr.2: 213364027-213364027 on Build GRCh38
rs780809875 Chr.2: 213364127-213364132 on Build GRCh38
rs774193622 Chr.2: 213364003-213364003 on Build GRCh38
rs996405866 Chr.2: 213363979-213363979 on Build GRCh38
rs73990322 Chr.2: 213364034-213364034 on Build GRCh38
rs1004766951 Chr.2: 213363936-213363936 on Build GRCh38
rs755965099 Chr.2: 213364121-213364121 on Build GRCh38
rs1049250008 Chr.2: 213363819-213363819 on Build GRCh38
rs561777476 Chr.2: 213363831-213363831 on Build GRCh38
rs754328855 Chr.2: 213364094-213364094 on Build GRCh38
rs191400347 Chr.2: 213363686-213363686 on Build GRCh38
rs1016190346 Chr.2: 213363939-213363939 on Build GRCh38
rs1032804903 Chr.2: 213363932-213363932 on Build GRCh38
rs200726046 Chr.2: 213364154-213364154 on Build GRCh38
rs765326677 Chr.2: 213364101-213364101 on Build GRCh38
rs751193019 Chr.2: 213364156-213364157 on Build GRCh38
rs568857869 Chr.2: 213363964-213363964 on Build GRCh38
rs946279332 Chr.2: 213363703-213363703 on Build GRCh38
rs772346688 Chr.2: 213364060-213364060 on Build GRCh38
rs781161267 Chr.2: 213364114-213364114 on Build GRCh38
rs987535864 Chr.2: 213364006-213364006 on Build GRCh38

View Full Product Details