Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230713197-230713746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAB39
Entrez Gene ID
51719
Gene Name
calcium binding protein 39
Gene Aliases
CGI-66, MO25
UniGene
Hs.603930
Gene Chromosome Location
Chr.2: 230712842-230821075 on Build GRCh38
Transcripts
NM_001130849.1 NM_001130850.1 NM_016289.3 XM_011511350.2

SNP Detail

SNP ID Chromosome Location
rs982142698 Chr.2: 230713361-230713361 on Build GRCh38
rs971573717 Chr.2: 230713356-230713356 on Build GRCh38
rs1050647542 Chr.2: 230713397-230713397 on Build GRCh38
rs1047046959 Chr.2: 230713559-230713560 on Build GRCh38
rs113117425 Chr.2: 230713270-230713270 on Build GRCh38
rs907287386 Chr.2: 230713653-230713653 on Build GRCh38
rs951130062 Chr.2: 230713284-230713284 on Build GRCh38
rs188726464 Chr.2: 230713511-230713511 on Build GRCh38
rs961685097 Chr.2: 230713510-230713510 on Build GRCh38
rs922674809 Chr.2: 230713674-230713674 on Build GRCh38
rs1001905481 Chr.2: 230713487-230713487 on Build GRCh38
rs1034649335 Chr.2: 230713599-230713599 on Build GRCh38
rs887022669 Chr.2: 230713559-230713559 on Build GRCh38
rs936603572 Chr.2: 230713570-230713570 on Build GRCh38
rs926977332 Chr.2: 230713244-230713244 on Build GRCh38
rs765240278 Chr.2: 230713501-230713501 on Build GRCh38
rs923137087 Chr.2: 230713389-230713389 on Build GRCh38
rs1003051989 Chr.2: 230713459-230713459 on Build GRCh38
rs985998539 Chr.2: 230713547-230713547 on Build GRCh38
rs922559079 Chr.2: 230713552-230713552 on Build GRCh38
rs568698178 Chr.2: 230713311-230713311 on Build GRCh38
rs759091858 Chr.2: 230713387-230713387 on Build GRCh38
rs540989068 Chr.2: 230713612-230713612 on Build GRCh38
rs767027424 Chr.2: 230713406-230713406 on Build GRCh38
rs937020686 Chr.2: 230713250-230713250 on Build GRCh38
rs1009177693 Chr.2: 230713615-230713615 on Build GRCh38
rs1005790110 Chr.2: 230713429-230713429 on Build GRCh38
rs148062240 Chr.2: 230713267-230713267 on Build GRCh38
rs573831683 Chr.2: 230713572-230713572 on Build GRCh38
rs1032972307 Chr.2: 230713323-230713323 on Build GRCh38
rs368862160 Chr.2: 230713555-230713555 on Build GRCh38
rs933158555 Chr.2: 230713390-230713390 on Build GRCh38
rs1053686698 Chr.2: 230713606-230713606 on Build GRCh38
rs557112520 Chr.2: 230713498-230713498 on Build GRCh38
rs984425657 Chr.2: 230713368-230713368 on Build GRCh38
rs889040036 Chr.2: 230713398-230713398 on Build GRCh38
rs901074738 Chr.2: 230713257-230713257 on Build GRCh38
rs933256412 Chr.2: 230713294-230713294 on Build GRCh38
rs1034892938 Chr.2: 230713476-230713476 on Build GRCh38
rs923051626 Chr.2: 230713539-230713539 on Build GRCh38
rs550149356 Chr.2: 230713293-230713293 on Build GRCh38
rs940472301 Chr.2: 230713386-230713386 on Build GRCh38
rs531766322 Chr.2: 230713272-230713272 on Build GRCh38
rs962723865 Chr.2: 230713637-230713637 on Build GRCh38
rs1020494815 Chr.2: 230713354-230713354 on Build GRCh38
rs893130887 Chr.2: 230713419-230713419 on Build GRCh38
rs576657916 Chr.2: 230713502-230713502 on Build GRCh38
rs942752043 Chr.2: 230713542-230713542 on Build GRCh38
rs1033573058 Chr.2: 230713657-230713657 on Build GRCh38
rs1050361986 Chr.2: 230713300-230713300 on Build GRCh38
rs1054474967 Chr.2: 230713409-230713409 on Build GRCh38
rs1007225365 Chr.2: 230713251-230713251 on Build GRCh38
rs964164759 Chr.2: 230713658-230713658 on Build GRCh38
rs914618174 Chr.2: 230713623-230713623 on Build GRCh38
rs2459923 Chr.2: 230713423-230713423 on Build GRCh38
rs374725907 Chr.2: 230713532-230713532 on Build GRCh38
rs562982134 Chr.2: 230713511-230713512 on Build GRCh38
rs371363297 Chr.2: 230713723-230713723 on Build GRCh38
rs535780138 Chr.2: 230713343-230713343 on Build GRCh38
rs2459922 Chr.2: 230713370-230713370 on Build GRCh38
rs1020572391 Chr.2: 230713616-230713616 on Build GRCh38
rs933873769 Chr.2: 230713417-230713417 on Build GRCh38
rs904953679 Chr.2: 230713577-230713577 on Build GRCh38
rs946024913 Chr.2: 230713625-230713625 on Build GRCh38
rs981707130 Chr.2: 230713243-230713243 on Build GRCh38
rs539876925 Chr.2: 230713446-230713446 on Build GRCh38
rs954569721 Chr.2: 230713545-230713545 on Build GRCh38
rs559264562 Chr.2: 230713711-230713711 on Build GRCh38
rs893242124 Chr.2: 230713344-230713344 on Build GRCh38
rs896120224 Chr.2: 230713607-230713607 on Build GRCh38
rs942742902 Chr.2: 230713379-230713379 on Build GRCh38
rs192334870 Chr.2: 230713563-230713563 on Build GRCh38
rs772926014 Chr.2: 230713275-230713275 on Build GRCh38
rs150620678 Chr.2: 230713713-230713713 on Build GRCh38
rs1006897864 Chr.2: 230713318-230713318 on Build GRCh38
rs1015905113 Chr.2: 230713503-230713503 on Build GRCh38
rs368113505 Chr.2: 230713664-230713664 on Build GRCh38
rs551074212 Chr.2: 230713218-230713218 on Build GRCh38
rs1036165306 Chr.2: 230713276-230713276 on Build GRCh38
rs959178996 Chr.2: 230713364-230713364 on Build GRCh38
rs1039684954 Chr.2: 230713253-230713253 on Build GRCh38
rs886912553 Chr.2: 230713424-230713424 on Build GRCh38
rs1041805176 Chr.2: 230713444-230713444 on Build GRCh38
rs1001884873 Chr.2: 230713584-230713584 on Build GRCh38
rs972071456 Chr.2: 230713223-230713223 on Build GRCh38
rs1034599268 Chr.2: 230713362-230713362 on Build GRCh38
rs889816059 Chr.2: 230713306-230713306 on Build GRCh38
rs984538913 Chr.2: 230713264-230713264 on Build GRCh38
rs1010298674 Chr.2: 230713351-230713351 on Build GRCh38
rs960297325 Chr.2: 230713499-230713499 on Build GRCh38

View Full Product Details