Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129140074-129140484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISY1
Entrez Gene ID
57461
Gene Name
ISY1 splicing factor homolog
Gene Aliases
FSAP33
UniGene
Hs.512661
Gene Chromosome Location
Chr.3: 129127416-129161230 on Build GRCh38
Transcripts
NM_001199469.1 NM_020701.3 NM_001204890.1

Gene Symbol
ISY1-RAB43
Entrez Gene ID
100534599
Gene Name
ISY1-RAB43 readthrough
Gene Aliases
-
UniGene
Hs.512661
Gene Chromosome Location
Chr.3: 129087569-129161230 on Build GRCh38
Transcripts
NM_001199469.1 NM_020701.3 NM_001204890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4318804 c.418C>A p.P140T Chr.3: 129140368-129140368 on Build GRCh38
1418795 c.380A>G p.K127R Chr.3: 129140406-129140406 on Build GRCh38
6225220 c.352C>T p.R118* Chr.3: 129140434-129140434 on Build GRCh38
4650617 c.418C>T p.P140S Chr.3: 129140368-129140368 on Build GRCh38
4947926 c.380A>G p.K127R Chr.3: 129140406-129140406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757823960 Chr.3: 129140371-129140371 on Build GRCh38
rs551298283 Chr.3: 129140342-129140342 on Build GRCh38
rs756459654 Chr.3: 129140417-129140417 on Build GRCh38
rs764775402 Chr.3: 129140349-129140349 on Build GRCh38
rs550454268 Chr.3: 129140180-129140180 on Build GRCh38
rs754692121 Chr.3: 129140346-129140346 on Build GRCh38
rs961209144 Chr.3: 129140225-129140225 on Build GRCh38
rs749467210 Chr.3: 129140433-129140433 on Build GRCh38
rs761669405 Chr.3: 129140325-129140325 on Build GRCh38
rs200389055 Chr.3: 129140370-129140370 on Build GRCh38
rs537176918 Chr.3: 129140184-129140184 on Build GRCh38
rs10684214 Chr.3: 129140362-129140363 on Build GRCh38
rs781776433 Chr.3: 129140394-129140394 on Build GRCh38
rs773302503 Chr.3: 129140124-129140124 on Build GRCh38
rs772862989 Chr.3: 129140339-129140339 on Build GRCh38
rs906030648 Chr.3: 129140102-129140102 on Build GRCh38
rs377433231 Chr.3: 129140331-129140331 on Build GRCh38
rs892600154 Chr.3: 129140351-129140351 on Build GRCh38
rs753481494 Chr.3: 129140343-129140343 on Build GRCh38
rs10664499 Chr.3: 129140359-129140360 on Build GRCh38
rs984056810 Chr.3: 129140198-129140198 on Build GRCh38
rs151260655 Chr.3: 129140321-129140321 on Build GRCh38
rs560925826 Chr.3: 129140121-129140121 on Build GRCh38
rs1008395885 Chr.3: 129140236-129140236 on Build GRCh38
rs200793292 Chr.3: 129140372-129140372 on Build GRCh38
rs973972368 Chr.3: 129140336-129140336 on Build GRCh38
rs199890591 Chr.3: 129140359-129140359 on Build GRCh38
rs34504018 Chr.3: 129140360-129140361 on Build GRCh38
rs955186351 Chr.3: 129140202-129140202 on Build GRCh38
rs62265338 Chr.3: 129140363-129140363 on Build GRCh38
rs566531045 Chr.3: 129140410-129140410 on Build GRCh38
rs780300226 Chr.3: 129140420-129140420 on Build GRCh38
rs778953332 Chr.3: 129140461-129140461 on Build GRCh38
rs779661959 Chr.3: 129140117-129140117 on Build GRCh38
rs562689230 Chr.3: 129140190-129140190 on Build GRCh38
rs755118308 Chr.3: 129140320-129140325 on Build GRCh38
rs902465136 Chr.3: 129140145-129140145 on Build GRCh38
rs181238539 Chr.3: 129140295-129140295 on Build GRCh38
rs923390489 Chr.3: 129140310-129140310 on Build GRCh38
rs768867156 Chr.3: 129140450-129140450 on Build GRCh38
rs972136578 Chr.3: 129140278-129140278 on Build GRCh38
rs531629218 Chr.3: 129140178-129140178 on Build GRCh38
rs869275228 Chr.3: 129140363-129140364 on Build GRCh38
rs1018083075 Chr.3: 129140244-129140244 on Build GRCh38
rs774425159 Chr.3: 129140323-129140323 on Build GRCh38
rs1020452477 Chr.3: 129140189-129140189 on Build GRCh38
rs202218284 Chr.3: 129140368-129140368 on Build GRCh38
rs1009882259 Chr.3: 129140179-129140179 on Build GRCh38

View Full Product Details