Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197776516-197777084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYTTD1
Entrez Gene ID
84248
Gene Name
forty-two-three domain containing 1
Gene Aliases
UIF
UniGene
Hs.277533
Gene Chromosome Location
Chr.3: 197749553-197784446 on Build GRCh38
Transcripts
NM_001011537.2 NM_032288.6 NR_027840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
730454 c.673A>T p.T225S Chr.3: 197776943-197776943 on Build GRCh38
336193 c.673A>G p.T225A Chr.3: 197776943-197776943 on Build GRCh38
7079886 c.698C>G p.S233C Chr.3: 197776968-197776968 on Build GRCh38
7414561 c.709C>T p.P237S Chr.3: 197776979-197776979 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773870463 Chr.3: 197776828-197776828 on Build GRCh38
rs139214163 Chr.3: 197776957-197776957 on Build GRCh38
rs374472675 Chr.3: 197777017-197777020 on Build GRCh38
rs189070195 Chr.3: 197777033-197777033 on Build GRCh38
rs759753903 Chr.3: 197777037-197777037 on Build GRCh38
rs763392617 Chr.3: 197776900-197776900 on Build GRCh38
rs369840594 Chr.3: 197776932-197776932 on Build GRCh38
rs899662860 Chr.3: 197776586-197776586 on Build GRCh38
rs796087503 Chr.3: 197776822-197776822 on Build GRCh38
rs745430353 Chr.3: 197776968-197776968 on Build GRCh38
rs971673037 Chr.3: 197777041-197777041 on Build GRCh38
rs773711801 Chr.3: 197776891-197776891 on Build GRCh38
rs376587309 Chr.3: 197776881-197776881 on Build GRCh38
rs200792127 Chr.3: 197776979-197776979 on Build GRCh38
rs759300973 Chr.3: 197776873-197776873 on Build GRCh38
rs756422306 Chr.3: 197777050-197777050 on Build GRCh38
rs991288566 Chr.3: 197776780-197776780 on Build GRCh38
rs1028900501 Chr.3: 197776659-197776659 on Build GRCh38
rs971722141 Chr.3: 197776736-197776739 on Build GRCh38
rs893177873 Chr.3: 197776686-197776686 on Build GRCh38
rs762234131 Chr.3: 197776878-197776878 on Build GRCh38
rs952527760 Chr.3: 197776848-197776848 on Build GRCh38
rs4622943 Chr.3: 197776803-197776803 on Build GRCh38
rs540461585 Chr.3: 197776798-197776798 on Build GRCh38
rs183744271 Chr.3: 197776711-197776711 on Build GRCh38
rs766446741 Chr.3: 197776901-197776901 on Build GRCh38
rs767906479 Chr.3: 197776924-197776924 on Build GRCh38
rs143872457 Chr.3: 197777053-197777053 on Build GRCh38
rs779359771 Chr.3: 197776871-197776871 on Build GRCh38
rs958881150 Chr.3: 197776773-197776773 on Build GRCh38
rs767590025 Chr.3: 197777010-197777013 on Build GRCh38
rs763515831 Chr.3: 197776902-197776902 on Build GRCh38
rs767114562 Chr.3: 197776874-197776874 on Build GRCh38
rs981679141 Chr.3: 197776706-197776706 on Build GRCh38
rs565509086 Chr.3: 197776827-197776827 on Build GRCh38
rs891276310 Chr.3: 197776952-197776952 on Build GRCh38
rs201860162 Chr.3: 197776908-197776908 on Build GRCh38
rs771710665 Chr.3: 197776970-197776970 on Build GRCh38
rs561248342 Chr.3: 197776762-197776762 on Build GRCh38
rs943955327 Chr.3: 197776539-197776539 on Build GRCh38
rs575251237 Chr.3: 197776701-197776701 on Build GRCh38
rs1033067388 Chr.3: 197776610-197776610 on Build GRCh38
rs1011580476 Chr.3: 197776689-197776689 on Build GRCh38
rs777438189 Chr.3: 197776942-197776942 on Build GRCh38
rs773638797 Chr.3: 197776901-197776902 on Build GRCh38
rs767707319 Chr.3: 197777040-197777040 on Build GRCh38
rs773484547 Chr.3: 197777009-197777009 on Build GRCh38
rs766847862 Chr.3: 197777020-197777020 on Build GRCh38
rs755259270 Chr.3: 197776673-197776673 on Build GRCh38
rs757172718 Chr.3: 197776950-197776950 on Build GRCh38
rs1040890318 Chr.3: 197776541-197776541 on Build GRCh38
rs915726947 Chr.3: 197776785-197776785 on Build GRCh38
rs755231098 Chr.3: 197776910-197776910 on Build GRCh38
rs746678476 Chr.3: 197776997-197776997 on Build GRCh38
rs930398022 Chr.3: 197776766-197776766 on Build GRCh38
rs770147940 Chr.3: 197777000-197777000 on Build GRCh38
rs766429012 Chr.3: 197776902-197776906 on Build GRCh38
rs116806598 Chr.3: 197776700-197776700 on Build GRCh38
rs1053991776 Chr.3: 197776966-197776966 on Build GRCh38
rs375632231 Chr.3: 197777049-197777049 on Build GRCh38
rs111226542 Chr.3: 197776628-197776628 on Build GRCh38
rs548460773 Chr.3: 197776991-197776991 on Build GRCh38
rs763488452 Chr.3: 197777016-197777016 on Build GRCh38
rs144055089 Chr.3: 197776931-197776931 on Build GRCh38
rs751684426 Chr.3: 197776910-197776911 on Build GRCh38
rs920307144 Chr.3: 197776763-197776763 on Build GRCh38
rs1046049735 Chr.3: 197776799-197776799 on Build GRCh38
rs558753665 Chr.3: 197776609-197776609 on Build GRCh38
rs115383101 Chr.3: 197776948-197776948 on Build GRCh38
rs919693292 Chr.3: 197776838-197776838 on Build GRCh38
rs539149381 Chr.3: 197776662-197776662 on Build GRCh38
rs755120916 Chr.3: 197776911-197776911 on Build GRCh38
rs996990259 Chr.3: 197776646-197776646 on Build GRCh38
rs369740997 Chr.3: 197776772-197776772 on Build GRCh38

View Full Product Details