Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43650223-43650611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPH9
Entrez Gene ID
221421
Gene Name
radial spoke head 9 homolog
Gene Aliases
C6orf206, CILD12, MRPS18AL1
UniGene
Hs.534585
Gene Chromosome Location
Chr.6: 43645030-43671011 on Build GRCh38
Transcripts
NM_001193341.1 NM_152732.4 XM_005248901.3 XM_006715014.1 XM_017010426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4722948 c.288G>A p.S96S Chr.6: 43650435-43650435 on Build GRCh38
1292329 c.320C>T p.P107L Chr.6: 43650467-43650467 on Build GRCh38
6354106 c.346C>T p.L116L Chr.6: 43650493-43650493 on Build GRCh38
3381611 c.372A>G p.K124K Chr.6: 43650519-43650519 on Build GRCh38
4542206 c.313G>A p.G105R Chr.6: 43650460-43650460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054247330 Chr.6: 43650541-43650541 on Build GRCh38
rs570326490 Chr.6: 43650308-43650308 on Build GRCh38
rs944591850 Chr.6: 43650493-43650493 on Build GRCh38
rs6915370 Chr.6: 43650258-43650258 on Build GRCh38
rs774004334 Chr.6: 43650451-43650451 on Build GRCh38
rs761387413 Chr.6: 43650460-43650460 on Build GRCh38
rs140285713 Chr.6: 43650474-43650474 on Build GRCh38
rs886061477 Chr.6: 43650532-43650532 on Build GRCh38
rs573487210 Chr.6: 43650448-43650448 on Build GRCh38
rs769699376 Chr.6: 43650365-43650365 on Build GRCh38
rs1044367157 Chr.6: 43650375-43650375 on Build GRCh38
rs577273074 Chr.6: 43650288-43650288 on Build GRCh38
rs757420621 Chr.6: 43650427-43650427 on Build GRCh38
rs773953326 Chr.6: 43650396-43650396 on Build GRCh38
rs76818047 Chr.6: 43650514-43650514 on Build GRCh38
rs751900140 Chr.6: 43650533-43650533 on Build GRCh38
rs201474277 Chr.6: 43650372-43650372 on Build GRCh38
rs777885477 Chr.6: 43650419-43650419 on Build GRCh38
rs766954824 Chr.6: 43650403-43650403 on Build GRCh38
rs142194323 Chr.6: 43650428-43650428 on Build GRCh38
rs769598177 Chr.6: 43650434-43650434 on Build GRCh38
rs368242248 Chr.6: 43650564-43650564 on Build GRCh38
rs77473340 Chr.6: 43650273-43650273 on Build GRCh38
rs914643708 Chr.6: 43650309-43650309 on Build GRCh38
rs200694391 Chr.6: 43650501-43650501 on Build GRCh38
rs368692729 Chr.6: 43650435-43650435 on Build GRCh38
rs6915523 Chr.6: 43650289-43650289 on Build GRCh38
rs774989438 Chr.6: 43650407-43650407 on Build GRCh38
rs771436734 Chr.6: 43650287-43650287 on Build GRCh38
rs372188063 Chr.6: 43650563-43650563 on Build GRCh38
rs747120923 Chr.6: 43650328-43650328 on Build GRCh38
rs760134926 Chr.6: 43650477-43650477 on Build GRCh38
rs924786748 Chr.6: 43650291-43650291 on Build GRCh38
rs199874020 Chr.6: 43650520-43650520 on Build GRCh38
rs934843364 Chr.6: 43650295-43650295 on Build GRCh38
rs60297264 Chr.6: 43650497-43650497 on Build GRCh38
rs765626320 Chr.6: 43650492-43650492 on Build GRCh38
rs762716238 Chr.6: 43650384-43650384 on Build GRCh38
rs551113626 Chr.6: 43650553-43650553 on Build GRCh38
rs868204071 Chr.6: 43650571-43650571 on Build GRCh38
rs1024584329 Chr.6: 43650389-43650389 on Build GRCh38
rs1032347070 Chr.6: 43650466-43650466 on Build GRCh38
rs367856581 Chr.6: 43650395-43650395 on Build GRCh38
rs758684050 Chr.6: 43650416-43650416 on Build GRCh38
rs35110984 Chr.6: 43650410-43650410 on Build GRCh38
rs151197990 Chr.6: 43650429-43650429 on Build GRCh38
rs199710227 Chr.6: 43650371-43650371 on Build GRCh38
rs373658809 Chr.6: 43650406-43650406 on Build GRCh38
rs142583052 Chr.6: 43650551-43650551 on Build GRCh38
rs1010820034 Chr.6: 43650388-43650388 on Build GRCh38
rs761334350 Chr.6: 43650399-43650399 on Build GRCh38
rs145695737 Chr.6: 43650313-43650313 on Build GRCh38
rs147687479 Chr.6: 43650502-43650502 on Build GRCh38
rs763244385 Chr.6: 43650517-43650517 on Build GRCh38
rs745736090 Chr.6: 43650364-43650364 on Build GRCh38
rs995756440 Chr.6: 43650408-43650408 on Build GRCh38
rs150021750 Chr.6: 43650475-43650475 on Build GRCh38
rs935743330 Chr.6: 43650536-43650536 on Build GRCh38
rs147713380 Chr.6: 43650423-43650423 on Build GRCh38
rs768448558 Chr.6: 43650440-43650440 on Build GRCh38
rs757541624 Chr.6: 43650540-43650540 on Build GRCh38
rs752978111 Chr.6: 43650411-43650411 on Build GRCh38
rs775136764 Chr.6: 43650369-43650369 on Build GRCh38
rs749068506 Chr.6: 43650439-43650439 on Build GRCh38
rs781295043 Chr.6: 43650342-43650342 on Build GRCh38
rs771089026 Chr.6: 43650340-43650340 on Build GRCh38
rs999921787 Chr.6: 43650301-43650301 on Build GRCh38

View Full Product Details