Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114527698-114528122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXPE1
Entrez Gene ID
120400
Gene Name
neurexophilin and PC-esterase domain family member 1
Gene Aliases
FAM55A
UniGene
Hs.721328
Gene Chromosome Location
Chr.11: 114518946-114560244 on Build GRCh38
Transcripts
NM_152315.3 XM_011542594.2 XM_011542595.2 XM_011542596.2 XM_011542597.2 XM_011542598.2 XM_011542599.2 XM_017017207.1 XM_017017209.1 XM_017017210.1

Gene Symbol
NXPE2
Entrez Gene ID
120406
Gene Name
neurexophilin and PC-esterase domain family member 2
Gene Aliases
FAM55B
UniGene
Hs.446760
Gene Chromosome Location
Chr.11: 114464276-114813702 on Build GRCh38
Transcripts
NM_152315.3 XM_011542594.2 XM_011542595.2 XM_011542596.2 XM_011542597.2 XM_011542598.2 XM_011542599.2 XM_017017207.1 XM_017017209.1 XM_017017210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3666101 c.418G>T p.G140* Chr.11: 114527891-114527891 on Build GRCh38
5046095 c.451G>A p.D151N Chr.11: 114527858-114527858 on Build GRCh38
3716773 c.458C>G p.T153S Chr.11: 114527851-114527851 on Build GRCh38
673803 c.440G>A p.R147H Chr.11: 114527869-114527869 on Build GRCh38
4939842 c.410C>A p.S137Y Chr.11: 114527899-114527899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770957797 Chr.11: 114527922-114527922 on Build GRCh38
rs755314910 Chr.11: 114527907-114527907 on Build GRCh38
rs754269717 Chr.11: 114527870-114527870 on Build GRCh38
rs796092142 Chr.11: 114527906-114527907 on Build GRCh38
rs539396912 Chr.11: 114527813-114527813 on Build GRCh38
rs760316653 Chr.11: 114527945-114527945 on Build GRCh38
rs773215540 Chr.11: 114527845-114527845 on Build GRCh38
rs777141697 Chr.11: 114527905-114527906 on Build GRCh38
rs145084457 Chr.11: 114527879-114527879 on Build GRCh38
rs12292536 Chr.11: 114527723-114527723 on Build GRCh38
rs748070045 Chr.11: 114527862-114527862 on Build GRCh38
rs969996868 Chr.11: 114527766-114527766 on Build GRCh38
rs907627182 Chr.11: 114527833-114527833 on Build GRCh38
rs893180453 Chr.11: 114527931-114527931 on Build GRCh38
rs143110380 Chr.11: 114527869-114527869 on Build GRCh38
rs771498579 Chr.11: 114527928-114527928 on Build GRCh38
rs760974592 Chr.11: 114527867-114527867 on Build GRCh38
rs748214199 Chr.11: 114527932-114527944 on Build GRCh38
rs76450715 Chr.11: 114527905-114527905 on Build GRCh38
rs748807076 Chr.11: 114527785-114527785 on Build GRCh38
rs370936440 Chr.11: 114527847-114527847 on Build GRCh38
rs771796822 Chr.11: 114528022-114528022 on Build GRCh38
rs1024227096 Chr.11: 114527730-114527730 on Build GRCh38
rs759766415 Chr.11: 114528045-114528045 on Build GRCh38
rs555065705 Chr.11: 114528000-114528000 on Build GRCh38
rs79589894 Chr.11: 114527916-114527916 on Build GRCh38
rs759005061 Chr.11: 114527855-114527855 on Build GRCh38
rs186842426 Chr.11: 114527768-114527768 on Build GRCh38
rs757683735 Chr.11: 114528014-114528016 on Build GRCh38
rs773885488 Chr.11: 114527851-114527851 on Build GRCh38
rs745337730 Chr.11: 114527927-114527927 on Build GRCh38
rs756470288 Chr.11: 114527792-114527792 on Build GRCh38
rs377413921 Chr.11: 114527826-114527826 on Build GRCh38
rs775144629 Chr.11: 114527935-114527935 on Build GRCh38
rs752319679 Chr.11: 114527860-114527860 on Build GRCh38
rs931967801 Chr.11: 114527872-114527872 on Build GRCh38
rs1003437637 Chr.11: 114528086-114528086 on Build GRCh38
rs780263636 Chr.11: 114527893-114527893 on Build GRCh38
rs763795477 Chr.11: 114527948-114527948 on Build GRCh38
rs768683517 Chr.11: 114527831-114527831 on Build GRCh38
rs777694703 Chr.11: 114527908-114527908 on Build GRCh38
rs754359712 Chr.11: 114527798-114527798 on Build GRCh38
rs778206967 Chr.11: 114527797-114527797 on Build GRCh38
rs772316847 Chr.11: 114527825-114527825 on Build GRCh38
rs116309340 Chr.11: 114527873-114527873 on Build GRCh38
rs1011174054 Chr.11: 114527936-114527936 on Build GRCh38
rs772926845 Chr.11: 114528029-114528029 on Build GRCh38
rs776729390 Chr.11: 114527921-114527922 on Build GRCh38
rs749139516 Chr.11: 114527914-114527914 on Build GRCh38
rs757263056 Chr.11: 114527805-114527805 on Build GRCh38
rs546752465 Chr.11: 114527830-114527830 on Build GRCh38
rs765713906 Chr.11: 114527878-114527878 on Build GRCh38
rs902341163 Chr.11: 114528010-114528010 on Build GRCh38
rs35058533 Chr.11: 114527906-114527906 on Build GRCh38
rs34542949 Chr.11: 114527906-114527907 on Build GRCh38
rs778409669 Chr.11: 114527876-114527878 on Build GRCh38
rs374323318 Chr.11: 114528025-114528025 on Build GRCh38
rs778961439 Chr.11: 114527810-114527810 on Build GRCh38
rs770030852 Chr.11: 114527944-114527945 on Build GRCh38
rs774189740 Chr.11: 114527924-114527924 on Build GRCh38
rs758429750 Chr.11: 114527880-114527880 on Build GRCh38
rs397815602 Chr.11: 114527915-114527916 on Build GRCh38
rs550851607 Chr.11: 114527745-114527745 on Build GRCh38
rs201428698 Chr.11: 114527840-114527840 on Build GRCh38
rs771047826 Chr.11: 114527848-114527848 on Build GRCh38
rs767162163 Chr.11: 114527858-114527858 on Build GRCh38
rs1052908389 Chr.11: 114527997-114527997 on Build GRCh38

View Full Product Details