Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118748416-118748930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX6
Entrez Gene ID
1656
Gene Name
DEAD-box helicase 6
Gene Aliases
HLR2, P54, RCK
UniGene
Hs.408461
Gene Chromosome Location
Chr.11: 118747763-118791263 on Build GRCh38
Transcripts
NM_001257191.2 NM_004397.5 XM_011542644.1 XM_011542645.1 XM_017017251.1

SNP Detail

SNP ID Chromosome Location
rs782623535 Chr.11: 118748791-118748791 on Build GRCh38
rs781878025 Chr.11: 118748531-118748531 on Build GRCh38
rs529761609 Chr.11: 118748630-118748630 on Build GRCh38
rs570958662 Chr.11: 118748488-118748488 on Build GRCh38
rs970120171 Chr.11: 118748521-118748521 on Build GRCh38
rs533765928 Chr.11: 118748897-118748897 on Build GRCh38
rs782393560 Chr.11: 118748849-118748849 on Build GRCh38
rs1043535300 Chr.11: 118748619-118748619 on Build GRCh38
rs537968963 Chr.11: 118748498-118748498 on Build GRCh38
rs962550586 Chr.11: 118748707-118748707 on Build GRCh38
rs923701857 Chr.11: 118748858-118748858 on Build GRCh38
rs566654335 Chr.11: 118748624-118748624 on Build GRCh38
rs782657954 Chr.11: 118748552-118748552 on Build GRCh38
rs754702328 Chr.11: 118748534-118748534 on Build GRCh38
rs1039627786 Chr.11: 118748594-118748594 on Build GRCh38
rs1023434473 Chr.11: 118748701-118748701 on Build GRCh38
rs185228198 Chr.11: 118748832-118748832 on Build GRCh38
rs199960214 Chr.11: 118748757-118748758 on Build GRCh38
rs184005423 Chr.11: 118748729-118748729 on Build GRCh38
rs994877150 Chr.11: 118748710-118748710 on Build GRCh38
rs983870162 Chr.11: 118748775-118748775 on Build GRCh38
rs900212964 Chr.11: 118748599-118748599 on Build GRCh38
rs777939872 Chr.11: 118748837-118748837 on Build GRCh38
rs1027716547 Chr.11: 118748759-118748759 on Build GRCh38
rs535712388 Chr.11: 118748562-118748562 on Build GRCh38
rs2508925 Chr.11: 118748721-118748721 on Build GRCh38
rs201564500 Chr.11: 118748714-118748714 on Build GRCh38
rs377385391 Chr.11: 118748747-118748748 on Build GRCh38
rs946892185 Chr.11: 118748836-118748836 on Build GRCh38
rs929345890 Chr.11: 118748543-118748543 on Build GRCh38
rs193050781 Chr.11: 118748805-118748805 on Build GRCh38
rs570514640 Chr.11: 118748747-118748747 on Build GRCh38
rs191774228 Chr.11: 118748523-118748523 on Build GRCh38
rs1961138 Chr.11: 118748714-118748714 on Build GRCh38
rs566343848 Chr.11: 118748663-118748663 on Build GRCh38
rs140512615 Chr.11: 118748586-118748586 on Build GRCh38
rs552367368 Chr.11: 118748448-118748448 on Build GRCh38
rs902162899 Chr.11: 118748634-118748634 on Build GRCh38
rs782758219 Chr.11: 118748746-118748747 on Build GRCh38
rs556351163 Chr.11: 118748818-118748818 on Build GRCh38
rs975057385 Chr.11: 118748807-118748807 on Build GRCh38
rs189583461 Chr.11: 118748769-118748769 on Build GRCh38
rs1056357419 Chr.11: 118748658-118748658 on Build GRCh38
rs150013784 Chr.11: 118748699-118748699 on Build GRCh38
rs1961139 Chr.11: 118748676-118748676 on Build GRCh38
rs946443144 Chr.11: 118748613-118748613 on Build GRCh38
rs896413700 Chr.11: 118748659-118748659 on Build GRCh38
rs999626905 Chr.11: 118748652-118748652 on Build GRCh38
rs2004781 Chr.11: 118748544-118748544 on Build GRCh38

View Full Product Details