Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119338850-119339352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF5
Entrez Gene ID
114902
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
CTRP5
UniGene
Hs.632102
Gene Chromosome Location
Chr.11: 119338934-119346673 on Build GRCh38
Transcripts
NM_001278431.1 NM_015645.4 NM_031433.3

Gene Symbol
MFRP
Entrez Gene ID
83552
Gene Name
membrane frizzled-related protein
Gene Aliases
MCOP5, NNO2, RD6
UniGene
Hs.632102
Gene Chromosome Location
Chr.11: 119338934-119346673 on Build GRCh38
Transcripts
NM_001278431.1 NM_015645.4 NM_031433.3

SNP Detail

SNP ID Chromosome Location
rs549464072 Chr.11: 119339199-119339199 on Build GRCh38
rs11217240 Chr.11: 119339206-119339206 on Build GRCh38
rs1019399817 Chr.11: 119339259-119339259 on Build GRCh38
rs777149267 Chr.11: 119339313-119339313 on Build GRCh38
rs551431328 Chr.11: 119338986-119338986 on Build GRCh38
rs1042379965 Chr.11: 119339177-119339177 on Build GRCh38
rs983080084 Chr.11: 119339295-119339295 on Build GRCh38
rs369261260 Chr.11: 119339323-119339323 on Build GRCh38
rs540189370 Chr.11: 119339172-119339172 on Build GRCh38
rs932857442 Chr.11: 119339059-119339059 on Build GRCh38
rs559347092 Chr.11: 119338944-119338944 on Build GRCh38
rs189336965 Chr.11: 119338873-119338873 on Build GRCh38
rs1003867368 Chr.11: 119339197-119339197 on Build GRCh38
rs1044817417 Chr.11: 119339139-119339139 on Build GRCh38
rs528395612 Chr.11: 119339055-119339055 on Build GRCh38
rs533245586 Chr.11: 119338959-119338959 on Build GRCh38
rs773466129 Chr.11: 119339328-119339328 on Build GRCh38
rs530757292 Chr.11: 119339113-119339113 on Build GRCh38
rs935028818 Chr.11: 119339032-119339032 on Build GRCh38
rs886047816 Chr.11: 119339080-119339080 on Build GRCh38
rs540630665 Chr.11: 119338942-119338942 on Build GRCh38
rs886047817 Chr.11: 119339173-119339173 on Build GRCh38
rs774976658 Chr.11: 119339280-119339280 on Build GRCh38
rs1033626316 Chr.11: 119339198-119339198 on Build GRCh38
rs200181343 Chr.11: 119339297-119339297 on Build GRCh38
rs746563698 Chr.11: 119339290-119339290 on Build GRCh38
rs757093089 Chr.11: 119339213-119339213 on Build GRCh38
rs906052940 Chr.11: 119339158-119339158 on Build GRCh38
rs944278239 Chr.11: 119338926-119338926 on Build GRCh38
rs765574187 Chr.11: 119339325-119339325 on Build GRCh38
rs1004893867 Chr.11: 119339249-119339249 on Build GRCh38
rs9640 Chr.11: 119339269-119339269 on Build GRCh38
rs1056492542 Chr.11: 119339041-119339041 on Build GRCh38
rs921427838 Chr.11: 119339052-119339052 on Build GRCh38
rs567782248 Chr.11: 119339240-119339240 on Build GRCh38
rs1009485333 Chr.11: 119339143-119339143 on Build GRCh38
rs537084889 Chr.11: 119339324-119339324 on Build GRCh38
rs906579086 Chr.11: 119339194-119339194 on Build GRCh38
rs115670501 Chr.11: 119339086-119339086 on Build GRCh38
rs373753331 Chr.11: 119339329-119339329 on Build GRCh38
rs766865012 Chr.11: 119338968-119338968 on Build GRCh38

View Full Product Details