Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 120226625-120227122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OAF
Entrez Gene ID
220323
Gene Name
out at first homolog
Gene Aliases
NS5ATP13TP2
UniGene
Hs.445081
Gene Chromosome Location
Chr.11: 120211039-120229941 on Build GRCh38
Transcripts
NM_178507.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1676508 c.370A>C p.N124H Chr.11: 120226819-120226819 on Build GRCh38
5371832 c.373C>T p.P125S Chr.11: 120226822-120226822 on Build GRCh38
5422032 c.490G>T p.E164* Chr.11: 120226939-120226939 on Build GRCh38
3985995 c.423C>A p.H141Q Chr.11: 120226872-120226872 on Build GRCh38
6351170 c.489C>T p.A163A Chr.11: 120226938-120226938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564005561 Chr.11: 120226648-120226648 on Build GRCh38
rs765765008 Chr.11: 120226925-120226925 on Build GRCh38
rs873058 Chr.11: 120226842-120226842 on Build GRCh38
rs776223244 Chr.11: 120226903-120226903 on Build GRCh38
rs147029182 Chr.11: 120226664-120226664 on Build GRCh38
rs775330605 Chr.11: 120226847-120226852 on Build GRCh38
rs756793176 Chr.11: 120226787-120226787 on Build GRCh38
rs200189572 Chr.11: 120226978-120226978 on Build GRCh38
rs768543253 Chr.11: 120226979-120226979 on Build GRCh38
rs149057050 Chr.11: 120226929-120226929 on Build GRCh38
rs750002128 Chr.11: 120226849-120226849 on Build GRCh38
rs144494021 Chr.11: 120226824-120226824 on Build GRCh38
rs1001148467 Chr.11: 120226723-120226723 on Build GRCh38
rs759546390 Chr.11: 120226828-120226828 on Build GRCh38
rs748586285 Chr.11: 120226939-120226939 on Build GRCh38
rs963923811 Chr.11: 120227068-120227068 on Build GRCh38
rs1048277657 Chr.11: 120226773-120226773 on Build GRCh38
rs781359992 Chr.11: 120226863-120226863 on Build GRCh38
rs771764357 Chr.11: 120226842-120226844 on Build GRCh38
rs755399151 Chr.11: 120226857-120226857 on Build GRCh38
rs753552892 Chr.11: 120226843-120226843 on Build GRCh38
rs113497939 Chr.11: 120226892-120226892 on Build GRCh38
rs931245772 Chr.11: 120226761-120226761 on Build GRCh38
rs200035341 Chr.11: 120226830-120226830 on Build GRCh38
rs904668889 Chr.11: 120226877-120226877 on Build GRCh38
rs867204745 Chr.11: 120226943-120226943 on Build GRCh38
rs199742045 Chr.11: 120227006-120227006 on Build GRCh38
rs747441332 Chr.11: 120226853-120226853 on Build GRCh38
rs750965471 Chr.11: 120227016-120227016 on Build GRCh38
rs773559616 Chr.11: 120226881-120226881 on Build GRCh38
rs375967354 Chr.11: 120226930-120226930 on Build GRCh38
rs753228658 Chr.11: 120227031-120227031 on Build GRCh38
rs764731731 Chr.11: 120226847-120226847 on Build GRCh38
rs879007742 Chr.11: 120226841-120226841 on Build GRCh38
rs760549931 Chr.11: 120227021-120227021 on Build GRCh38
rs990999480 Chr.11: 120226699-120226699 on Build GRCh38
rs376221495 Chr.11: 120226687-120226687 on Build GRCh38
rs572334937 Chr.11: 120227057-120227057 on Build GRCh38
rs1024562348 Chr.11: 120226936-120226936 on Build GRCh38
rs777534523 Chr.11: 120226780-120226780 on Build GRCh38
rs76523273 Chr.11: 120226701-120226701 on Build GRCh38
rs747799548 Chr.11: 120226928-120226928 on Build GRCh38
rs369472498 Chr.11: 120226793-120226793 on Build GRCh38
rs1042359268 Chr.11: 120226709-120226709 on Build GRCh38
rs140254492 Chr.11: 120226963-120226963 on Build GRCh38
rs551785090 Chr.11: 120226914-120226914 on Build GRCh38
rs749433430 Chr.11: 120226882-120226882 on Build GRCh38
rs151137876 Chr.11: 120226944-120226944 on Build GRCh38
rs757890745 Chr.11: 120226850-120226850 on Build GRCh38
rs377251120 Chr.11: 120227005-120227005 on Build GRCh38
rs775531621 Chr.11: 120226889-120226889 on Build GRCh38
rs778184167 Chr.11: 120226945-120226945 on Build GRCh38
rs778237008 Chr.11: 120227045-120227045 on Build GRCh38
rs145186688 Chr.11: 120226852-120226852 on Build GRCh38
rs768878445 Chr.11: 120226815-120226815 on Build GRCh38
rs753403302 Chr.11: 120226782-120226782 on Build GRCh38
rs370438022 Chr.11: 120227009-120227009 on Build GRCh38
rs771067831 Chr.11: 120226964-120226964 on Build GRCh38
rs374542166 Chr.11: 120226798-120226798 on Build GRCh38
rs972547433 Chr.11: 120226710-120226710 on Build GRCh38
rs902332462 Chr.11: 120226719-120226719 on Build GRCh38
rs1053869537 Chr.11: 120226745-120226745 on Build GRCh38
rs572795920 Chr.11: 120227087-120227087 on Build GRCh38
rs911140020 Chr.11: 120227035-120227035 on Build GRCh38
rs771107311 Chr.11: 120226884-120226884 on Build GRCh38
rs200957652 Chr.11: 120226834-120226834 on Build GRCh38
rs113533099 Chr.11: 120227022-120227022 on Build GRCh38
rs995116028 Chr.11: 120227090-120227090 on Build GRCh38
rs770071046 Chr.11: 120226875-120226875 on Build GRCh38
rs773363476 Chr.11: 120226826-120226826 on Build GRCh38
rs942732175 Chr.11: 120227076-120227076 on Build GRCh38
rs776950790 Chr.11: 120226822-120226822 on Build GRCh38
rs761644882 Chr.11: 120227004-120227004 on Build GRCh38
rs367651245 Chr.11: 120227012-120227012 on Build GRCh38
rs767623985 Chr.11: 120226829-120226829 on Build GRCh38
rs145262765 Chr.11: 120226965-120226965 on Build GRCh38
rs372575959 Chr.11: 120227001-120227001 on Build GRCh38
rs761591936 Chr.11: 120226910-120226910 on Build GRCh38
rs949254668 Chr.11: 120226702-120226702 on Build GRCh38
rs772065124 Chr.11: 120226918-120226918 on Build GRCh38
rs554128021 Chr.11: 120227032-120227032 on Build GRCh38
rs745463220 Chr.11: 120226789-120226789 on Build GRCh38
rs873059 Chr.11: 120226789-120226789 on Build GRCh38
rs757941904 Chr.11: 120226920-120226920 on Build GRCh38
rs746388253 Chr.11: 120226791-120226791 on Build GRCh38
rs3740736 Chr.11: 120226938-120226938 on Build GRCh38
rs199517878 Chr.11: 120226804-120226804 on Build GRCh38
rs773895089 Chr.11: 120226787-120226787 on Build GRCh38
rs918378022 Chr.11: 120226750-120226750 on Build GRCh38
rs559289815 Chr.11: 120226865-120226865 on Build GRCh38
rs772500904 Chr.11: 120226792-120226792 on Build GRCh38
rs147440018 Chr.11: 120226825-120226825 on Build GRCh38
rs756846378 Chr.11: 120226844-120226844 on Build GRCh38
rs539049275 Chr.11: 120227084-120227084 on Build GRCh38

View Full Product Details