Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 120332458-120333005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM136
Entrez Gene ID
219902
Gene Name
transmembrane protein 136
Gene Aliases
-
UniGene
Hs.643516
Gene Chromosome Location
Chr.11: 120324298-120333688 on Build GRCh38
Transcripts
NM_001198670.1 NM_001198671.1 NM_001198672.1 NM_001198673.1 NM_001198674.1 NM_001198675.1 NM_174926.2 XM_017017327.1

SNP Detail

SNP ID Chromosome Location
rs936033299 Chr.11: 120332746-120332746 on Build GRCh38
rs34298380 Chr.11: 120332850-120332850 on Build GRCh38
rs894693895 Chr.11: 120332514-120332514 on Build GRCh38
rs1045420516 Chr.11: 120332523-120332523 on Build GRCh38
rs576371970 Chr.11: 120332571-120332571 on Build GRCh38
rs1046011549 Chr.11: 120332820-120332820 on Build GRCh38
rs909125249 Chr.11: 120332941-120332941 on Build GRCh38
rs966694853 Chr.11: 120332705-120332705 on Build GRCh38
rs990994020 Chr.11: 120332744-120332744 on Build GRCh38
rs541132205 Chr.11: 120332658-120332658 on Build GRCh38
rs1018811769 Chr.11: 120332981-120332981 on Build GRCh38
rs572517637 Chr.11: 120332503-120332503 on Build GRCh38
rs1049234813 Chr.11: 120332927-120332927 on Build GRCh38
rs543411575 Chr.11: 120332592-120332592 on Build GRCh38
rs58377417 Chr.11: 120332549-120332550 on Build GRCh38
rs1048992833 Chr.11: 120332572-120332572 on Build GRCh38
rs531300237 Chr.11: 120332520-120332521 on Build GRCh38
rs991418457 Chr.11: 120332490-120332490 on Build GRCh38
rs980069038 Chr.11: 120332723-120332723 on Build GRCh38
rs12806740 Chr.11: 120332919-120332919 on Build GRCh38
rs1014679454 Chr.11: 120332856-120332856 on Build GRCh38
rs773499720 Chr.11: 120332521-120332523 on Build GRCh38
rs1056051973 Chr.11: 120332488-120332488 on Build GRCh38
rs777374008 Chr.11: 120332800-120332800 on Build GRCh38
rs552598194 Chr.11: 120332517-120332517 on Build GRCh38
rs1032575919 Chr.11: 120332733-120332733 on Build GRCh38
rs764893851 Chr.11: 120332523-120332524 on Build GRCh38
rs559820016 Chr.11: 120332695-120332695 on Build GRCh38
rs556900405 Chr.11: 120332953-120332953 on Build GRCh38
rs368088242 Chr.11: 120332837-120332837 on Build GRCh38
rs758824823 Chr.11: 120332584-120332584 on Build GRCh38
rs1020878569 Chr.11: 120332692-120332692 on Build GRCh38
rs185919466 Chr.11: 120332569-120332569 on Build GRCh38
rs12364701 Chr.11: 120332943-120332943 on Build GRCh38
rs147519184 Chr.11: 120332645-120332645 on Build GRCh38
rs78505084 Chr.11: 120332549-120332549 on Build GRCh38
rs894462914 Chr.11: 120332786-120332786 on Build GRCh38
rs561777927 Chr.11: 120332618-120332618 on Build GRCh38
rs949814137 Chr.11: 120332511-120332511 on Build GRCh38
rs562793502 Chr.11: 120332721-120332721 on Build GRCh38
rs901933504 Chr.11: 120332655-120332655 on Build GRCh38
rs754390833 Chr.11: 120332526-120332526 on Build GRCh38
rs1017889935 Chr.11: 120332653-120332653 on Build GRCh38
rs1004350350 Chr.11: 120332851-120332851 on Build GRCh38
rs527449192 Chr.11: 120332756-120332756 on Build GRCh38
rs372589124 Chr.11: 120332660-120332660 on Build GRCh38
rs79259333 Chr.11: 120332964-120332964 on Build GRCh38
rs191462626 Chr.11: 120332783-120332783 on Build GRCh38
rs112946342 Chr.11: 120332743-120332743 on Build GRCh38
rs566995459 Chr.11: 120332866-120332866 on Build GRCh38
rs921890566 Chr.11: 120332640-120332640 on Build GRCh38
rs1024506995 Chr.11: 120332521-120332532 on Build GRCh38
rs1012907213 Chr.11: 120332774-120332774 on Build GRCh38
rs918127011 Chr.11: 120332778-120332778 on Build GRCh38
rs951507496 Chr.11: 120332548-120332548 on Build GRCh38
rs1014442382 Chr.11: 120332521-120332526 on Build GRCh38

View Full Product Details