Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 121102588-121103094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TECTA
Entrez Gene ID
7007
Gene Name
tectorin alpha
Gene Aliases
DFNA12, DFNA8, DFNB21
UniGene
Hs.248162
Gene Chromosome Location
Chr.11: 121102666-121190806 on Build GRCh38
Transcripts
NM_005422.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923993 c.43T>G p.F15V Chr.11: 121102708-121102708 on Build GRCh38
923992 c.7T>G p.Y3D Chr.11: 121102672-121102672 on Build GRCh38
5050601 c.6T>A p.N2K Chr.11: 121102671-121102671 on Build GRCh38
5570860 c.52G>C p.V18L Chr.11: 121102717-121102717 on Build GRCh38
6290282 c.31G>A p.V11I Chr.11: 121102696-121102696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759114822 Chr.11: 121102772-121102772 on Build GRCh38
rs775605340 Chr.11: 121102999-121102999 on Build GRCh38
rs754200393 Chr.11: 121102631-121102631 on Build GRCh38
rs773805206 Chr.11: 121102767-121102767 on Build GRCh38
rs759915862 Chr.11: 121102765-121102765 on Build GRCh38
rs773207318 Chr.11: 121102745-121102745 on Build GRCh38
rs539772308 Chr.11: 121102998-121102998 on Build GRCh38
rs749080253 Chr.11: 121102747-121102747 on Build GRCh38
rs983400762 Chr.11: 121102821-121102821 on Build GRCh38
rs118013075 Chr.11: 121102798-121102798 on Build GRCh38
rs567656439 Chr.11: 121102878-121102878 on Build GRCh38
rs569883658 Chr.11: 121102827-121102827 on Build GRCh38
rs756491046 Chr.11: 121102626-121102626 on Build GRCh38
rs187420900 Chr.11: 121102984-121102984 on Build GRCh38
rs140393508 Chr.11: 121102698-121102698 on Build GRCh38
rs558472378 Chr.11: 121102680-121102680 on Build GRCh38
rs371295280 Chr.11: 121102776-121102776 on Build GRCh38
rs904561133 Chr.11: 121103020-121103020 on Build GRCh38
rs531024679 Chr.11: 121102859-121102859 on Build GRCh38
rs946189628 Chr.11: 121102990-121102990 on Build GRCh38
rs192066633 Chr.11: 121103025-121103025 on Build GRCh38
rs558100664 Chr.11: 121103016-121103016 on Build GRCh38
rs35507522 Chr.11: 121102721-121102721 on Build GRCh38
rs747926423 Chr.11: 121103069-121103069 on Build GRCh38
rs911573556 Chr.11: 121102925-121102925 on Build GRCh38
rs1034989523 Chr.11: 121103052-121103052 on Build GRCh38
rs772378473 Chr.11: 121102870-121102870 on Build GRCh38
rs763424515 Chr.11: 121102633-121102633 on Build GRCh38
rs549415293 Chr.11: 121102872-121102872 on Build GRCh38
rs921725430 Chr.11: 121102617-121102617 on Build GRCh38
rs559610902 Chr.11: 121102755-121102755 on Build GRCh38
rs764574591 Chr.11: 121102778-121102778 on Build GRCh38
rs28481890 Chr.11: 121102825-121102825 on Build GRCh38
rs1053648092 Chr.11: 121103042-121103042 on Build GRCh38
rs756460108 Chr.11: 121102710-121102710 on Build GRCh38
rs868714310 Chr.11: 121102884-121102884 on Build GRCh38
rs375404227 Chr.11: 121102777-121102777 on Build GRCh38
rs528834518 Chr.11: 121102667-121102667 on Build GRCh38
rs907573546 Chr.11: 121102812-121102812 on Build GRCh38
rs149919658 Chr.11: 121102711-121102711 on Build GRCh38
rs894087235 Chr.11: 121102985-121102985 on Build GRCh38
rs921652741 Chr.11: 121102873-121102873 on Build GRCh38
rs935021236 Chr.11: 121102939-121102939 on Build GRCh38
rs931735344 Chr.11: 121102900-121102900 on Build GRCh38
rs922151769 Chr.11: 121102875-121102875 on Build GRCh38
rs778157765 Chr.11: 121102627-121102627 on Build GRCh38
rs1035020891 Chr.11: 121102809-121102809 on Build GRCh38
rs145916279 Chr.11: 121102653-121102653 on Build GRCh38
rs200130858 Chr.11: 121102633-121102634 on Build GRCh38
rs553132015 Chr.11: 121102940-121102940 on Build GRCh38
rs960205480 Chr.11: 121102815-121102815 on Build GRCh38
rs746066900 Chr.11: 121102866-121102866 on Build GRCh38
rs757560920 Chr.11: 121102643-121102643 on Build GRCh38
rs754218636 Chr.11: 121102781-121102781 on Build GRCh38
rs1012502802 Chr.11: 121103068-121103068 on Build GRCh38
rs56358414 Chr.11: 121102807-121102807 on Build GRCh38
rs751969965 Chr.11: 121102634-121102634 on Build GRCh38
rs535014231 Chr.11: 121102923-121102923 on Build GRCh38
rs554453576 Chr.11: 121102760-121102763 on Build GRCh38

View Full Product Details