Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 122655722-122656217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBASH3B
Entrez Gene ID
84959
Gene Name
ubiquitin associated and SH3 domain containing B
Gene Aliases
STS-1, STS1, TULA-2, TULA2, p70
UniGene
Hs.444075
Gene Chromosome Location
Chr.11: 122655690-122814479 on Build GRCh38
Transcripts
NM_032873.4 XM_011543042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7396468 c.154G>C p.A52P Chr.11: 122656203-122656203 on Build GRCh38
6263216 c.12C>T p.Y4Y Chr.11: 122656061-122656061 on Build GRCh38
4738643 c.68delC p.R25fs*76 Chr.11: 122656117-122656117 on Build GRCh38
6396587 c.58A>T p.S20C Chr.11: 122656107-122656107 on Build GRCh38
6383140 c.130C>A p.L44I Chr.11: 122656179-122656179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042598538 Chr.11: 122655831-122655831 on Build GRCh38
rs778658634 Chr.11: 122656027-122656027 on Build GRCh38
rs745545499 Chr.11: 122655759-122655759 on Build GRCh38
rs934374141 Chr.11: 122655922-122655922 on Build GRCh38
rs140079462 Chr.11: 122656148-122656148 on Build GRCh38
rs776599575 Chr.11: 122656121-122656121 on Build GRCh38
rs770318439 Chr.11: 122656076-122656076 on Build GRCh38
rs868417514 Chr.11: 122656141-122656141 on Build GRCh38
rs1028890304 Chr.11: 122655838-122655838 on Build GRCh38
rs751679167 Chr.11: 122656014-122656014 on Build GRCh38
rs776918107 Chr.11: 122656116-122656117 on Build GRCh38
rs761708229 Chr.11: 122656130-122656130 on Build GRCh38
rs779678247 Chr.11: 122655832-122655832 on Build GRCh38
rs899265830 Chr.11: 122656083-122656083 on Build GRCh38
rs766062163 Chr.11: 122656146-122656146 on Build GRCh38
rs758910676 Chr.11: 122655971-122655971 on Build GRCh38
rs991140724 Chr.11: 122655949-122655949 on Build GRCh38
rs924369679 Chr.11: 122655907-122655907 on Build GRCh38
rs757101586 Chr.11: 122656025-122656025 on Build GRCh38
rs948203801 Chr.11: 122655775-122655775 on Build GRCh38
rs905403650 Chr.11: 122655988-122655988 on Build GRCh38
rs758073778 Chr.11: 122656145-122656145 on Build GRCh38
rs750256698 Chr.11: 122656143-122656143 on Build GRCh38
rs780814631 Chr.11: 122656168-122656168 on Build GRCh38
rs753770185 Chr.11: 122656017-122656017 on Build GRCh38
rs576825078 Chr.11: 122655762-122655762 on Build GRCh38
rs747617461 Chr.11: 122656182-122656182 on Build GRCh38
rs560524802 Chr.11: 122655943-122655943 on Build GRCh38
rs940862514 Chr.11: 122656061-122656061 on Build GRCh38
rs1045720304 Chr.11: 122655942-122655942 on Build GRCh38
rs754596519 Chr.11: 122656156-122656156 on Build GRCh38
rs762632430 Chr.11: 122655748-122655748 on Build GRCh38
rs994852725 Chr.11: 122655836-122655836 on Build GRCh38
rs768217066 Chr.11: 122656062-122656062 on Build GRCh38
rs150696870 Chr.11: 122656113-122656113 on Build GRCh38
rs558665410 Chr.11: 122655750-122655750 on Build GRCh38
rs767570158 Chr.11: 122656117-122656117 on Build GRCh38
rs906601336 Chr.11: 122655792-122655792 on Build GRCh38
rs200380590 Chr.11: 122656015-122656015 on Build GRCh38
rs962632382 Chr.11: 122655795-122655795 on Build GRCh38
rs766470283 Chr.11: 122656010-122656010 on Build GRCh38
rs868423666 Chr.11: 122656198-122656198 on Build GRCh38
rs1019288408 Chr.11: 122656131-122656131 on Build GRCh38
rs563840858 Chr.11: 122655915-122655929 on Build GRCh38
rs765046434 Chr.11: 122656142-122656142 on Build GRCh38
rs758102795 Chr.11: 122656040-122656040 on Build GRCh38
rs149619094 Chr.11: 122656068-122656068 on Build GRCh38
rs1052627478 Chr.11: 122655923-122655923 on Build GRCh38
rs865861575 Chr.11: 122656190-122656190 on Build GRCh38
rs542381822 Chr.11: 122655903-122655903 on Build GRCh38
rs773652683 Chr.11: 122656077-122656077 on Build GRCh38
rs779525094 Chr.11: 122656050-122656050 on Build GRCh38
rs7128198 Chr.11: 122655893-122655893 on Build GRCh38
rs771196593 Chr.11: 122656091-122656091 on Build GRCh38
rs1003959037 Chr.11: 122656000-122656000 on Build GRCh38
rs7127978 Chr.11: 122655793-122655793 on Build GRCh38
rs953343919 Chr.11: 122655843-122655843 on Build GRCh38
rs920980082 Chr.11: 122655861-122655861 on Build GRCh38
rs986449836 Chr.11: 122655894-122655894 on Build GRCh38
rs915450846 Chr.11: 122656172-122656172 on Build GRCh38
rs946985478 Chr.11: 122655938-122655938 on Build GRCh38
rs893965174 Chr.11: 122655924-122655924 on Build GRCh38
rs544229297 Chr.11: 122655787-122655787 on Build GRCh38
rs916803414 Chr.11: 122655979-122655979 on Build GRCh38
rs375787428 Chr.11: 122656200-122656200 on Build GRCh38
rs748655872 Chr.11: 122656073-122656073 on Build GRCh38
rs750083695 Chr.11: 122656033-122656033 on Build GRCh38
rs199692628 Chr.11: 122656016-122656016 on Build GRCh38
rs951430994 Chr.11: 122655788-122655788 on Build GRCh38
rs763175516 Chr.11: 122656086-122656086 on Build GRCh38
rs564196528 Chr.11: 122656058-122656058 on Build GRCh38
rs987704612 Chr.11: 122655887-122655887 on Build GRCh38
rs982590453 Chr.11: 122655791-122655791 on Build GRCh38

View Full Product Details