Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32679007-32679500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1L
Entrez Gene ID
10059
Gene Name
dynamin 1 like
Gene Aliases
DLP1, DRP1, DVLP, DYMPLE, EMPF, HDYNIV
UniGene
Hs.556296
Gene Chromosome Location
Chr.12: 32679200-32745650 on Build GRCh38
Transcripts
NM_001278463.1 NM_001278464.1 NM_001278465.1 NM_001278466.1 NM_005690.4 NM_012062.4 NM_012063.3 XM_005253282.4 XM_011520543.2 XM_011520544.2 XM_017018663.1 XM_017018664.1 XM_017018665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3739614 c.58G>A p.A20T Chr.12: 32679421-32679421 on Build GRCh38
3396069 c.56G>T p.G19V Chr.12: 32679419-32679419 on Build GRCh38
3396070 c.56G>T p.G19V Chr.12: 32679419-32679419 on Build GRCh38
7429252 c.38A>T p.D13V Chr.12: 32679401-32679401 on Build GRCh38
3739613 c.58G>A p.A20T Chr.12: 32679421-32679421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896148786 Chr.12: 32679409-32679409 on Build GRCh38
rs767578893 Chr.12: 32679354-32679354 on Build GRCh38
rs116816682 Chr.12: 32679092-32679092 on Build GRCh38
rs747115615 Chr.12: 32679347-32679347 on Build GRCh38
rs766636851 Chr.12: 32679311-32679311 on Build GRCh38
rs151285821 Chr.12: 32679363-32679363 on Build GRCh38
rs754614254 Chr.12: 32679357-32679357 on Build GRCh38
rs201968952 Chr.12: 32679350-32679350 on Build GRCh38
rs191084732 Chr.12: 32679339-32679339 on Build GRCh38
rs771990382 Chr.12: 32679438-32679438 on Build GRCh38
rs201348675 Chr.12: 32679399-32679399 on Build GRCh38
rs781103166 Chr.12: 32679388-32679388 on Build GRCh38
rs760304487 Chr.12: 32679355-32679355 on Build GRCh38
rs758693731 Chr.12: 32679361-32679361 on Build GRCh38
rs200938430 Chr.12: 32679411-32679411 on Build GRCh38
rs772586756 Chr.12: 32679344-32679344 on Build GRCh38
rs1026398918 Chr.12: 32679288-32679288 on Build GRCh38
rs772176772 Chr.12: 32679308-32679308 on Build GRCh38
rs767582592 Chr.12: 32679170-32679170 on Build GRCh38
rs923729047 Chr.12: 32679171-32679171 on Build GRCh38
rs374067166 Chr.12: 32679348-32679348 on Build GRCh38
rs912124914 Chr.12: 32679103-32679103 on Build GRCh38
rs771090587 Chr.12: 32679380-32679380 on Build GRCh38
rs752555558 Chr.12: 32679240-32679240 on Build GRCh38
rs748628452 Chr.12: 32679307-32679307 on Build GRCh38
rs776650839 Chr.12: 32679313-32679313 on Build GRCh38
rs757231215 Chr.12: 32679322-32679322 on Build GRCh38
rs946913846 Chr.12: 32679144-32679144 on Build GRCh38
rs771381138 Chr.12: 32679389-32679389 on Build GRCh38
rs751536498 Chr.12: 32679272-32679272 on Build GRCh38
rs374414603 Chr.12: 32679316-32679316 on Build GRCh38
rs780828933 Chr.12: 32679283-32679283 on Build GRCh38
rs1043939623 Chr.12: 32679173-32679173 on Build GRCh38
rs934055624 Chr.12: 32679130-32679130 on Build GRCh38
rs756884077 Chr.12: 32679277-32679277 on Build GRCh38
rs770521657 Chr.12: 32679326-32679326 on Build GRCh38
rs367813907 Chr.12: 32679317-32679317 on Build GRCh38
rs753070922 Chr.12: 32679360-32679360 on Build GRCh38
rs915515186 Chr.12: 32679330-32679330 on Build GRCh38
rs548807739 Chr.12: 32679287-32679287 on Build GRCh38
rs1012182643 Chr.12: 32679256-32679256 on Build GRCh38
rs939007091 Chr.12: 32679202-32679202 on Build GRCh38
rs552273580 Chr.12: 32679114-32679114 on Build GRCh38
rs1034615027 Chr.12: 32679235-32679235 on Build GRCh38
rs748468670 Chr.12: 32679343-32679343 on Build GRCh38
rs773469192 Chr.12: 32679186-32679186 on Build GRCh38
rs114505859 Chr.12: 32679332-32679332 on Build GRCh38
rs1038181985 Chr.12: 32679281-32679281 on Build GRCh38
rs550823466 Chr.12: 32679310-32679310 on Build GRCh38
rs778486856 Chr.12: 32679359-32679359 on Build GRCh38
rs372402179 Chr.12: 32679340-32679340 on Build GRCh38
rs749680977 Chr.12: 32679299-32679299 on Build GRCh38
rs58142640 Chr.12: 32679305-32679305 on Build GRCh38
rs1011997606 Chr.12: 32679253-32679253 on Build GRCh38
rs774908821 Chr.12: 32679306-32679306 on Build GRCh38
rs960416776 Chr.12: 32679053-32679053 on Build GRCh38
rs902749463 Chr.12: 32679205-32679205 on Build GRCh38
rs370381346 Chr.12: 32679356-32679356 on Build GRCh38
rs1026430250 Chr.12: 32679070-32679070 on Build GRCh38
rs755822735 Chr.12: 32679291-32679291 on Build GRCh38
rs748892584 Chr.12: 32679338-32679338 on Build GRCh38
rs990986559 Chr.12: 32679064-32679064 on Build GRCh38
rs758069766 Chr.12: 32679242-32679242 on Build GRCh38
rs371570335 Chr.12: 32679319-32679319 on Build GRCh38
rs915421896 Chr.12: 32679123-32679123 on Build GRCh38
rs764098117 Chr.12: 32679262-32679262 on Build GRCh38
rs376208045 Chr.12: 32679282-32679282 on Build GRCh38
rs922609165 Chr.12: 32679121-32679121 on Build GRCh38
rs761652857 Chr.12: 32679352-32679352 on Build GRCh38
rs776969387 Chr.12: 32679349-32679349 on Build GRCh38
rs1050273098 Chr.12: 32679137-32679137 on Build GRCh38
rs549864270 Chr.12: 32679086-32679086 on Build GRCh38
rs745921568 Chr.12: 32679391-32679391 on Build GRCh38
rs761707963 Chr.12: 32679416-32679416 on Build GRCh38
rs563411738 Chr.12: 32679091-32679091 on Build GRCh38
rs760693266 Chr.12: 32679309-32679309 on Build GRCh38
rs767523355 Chr.12: 32679325-32679325 on Build GRCh38
rs755525061 Chr.12: 32679329-32679329 on Build GRCh38
rs566391815 Chr.12: 32679166-32679166 on Build GRCh38
rs950678646 Chr.12: 32679090-32679090 on Build GRCh38
rs886049282 Chr.12: 32679314-32679314 on Build GRCh38
rs775798240 Chr.12: 32679414-32679414 on Build GRCh38
rs535003366 Chr.12: 32679233-32679233 on Build GRCh38
rs879382232 Chr.12: 32679147-32679147 on Build GRCh38
rs747457741 Chr.12: 32679372-32679372 on Build GRCh38
rs773648742 Chr.12: 32679346-32679346 on Build GRCh38
rs748926212 Chr.12: 32679108-32679108 on Build GRCh38

View Full Product Details