Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 74537684-74538195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L3B
Entrez Gene ID
552889
Gene Name
ataxin 7 like 3B
Gene Aliases
lnc-SCA7
UniGene
Hs.744849
Gene Chromosome Location
Chr.12: 74537771-74541452 on Build GRCh38
Transcripts
NM_001136262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4974024 c.4G>C p.E2Q Chr.12: 74538116-74538116 on Build GRCh38
943403 c.81G>A p.E27E Chr.12: 74538193-74538193 on Build GRCh38
4211208 c.70G>T p.D24Y Chr.12: 74538182-74538182 on Build GRCh38
6293954 c.1A>G p.M1V Chr.12: 74538113-74538113 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs949558180 Chr.12: 74537963-74537963 on Build GRCh38
rs754408882 Chr.12: 74537778-74537778 on Build GRCh38
rs556295109 Chr.12: 74537765-74537765 on Build GRCh38
rs1004894045 Chr.12: 74537935-74537935 on Build GRCh38
rs149090705 Chr.12: 74537871-74537871 on Build GRCh38
rs755409981 Chr.12: 74537789-74537789 on Build GRCh38
rs917094940 Chr.12: 74538028-74538028 on Build GRCh38
rs886508208 Chr.12: 74537844-74537844 on Build GRCh38
rs1029794168 Chr.12: 74537749-74537749 on Build GRCh38
rs547208470 Chr.12: 74538085-74538085 on Build GRCh38
rs1051716127 Chr.12: 74537884-74537884 on Build GRCh38
rs996448402 Chr.12: 74537742-74537742 on Build GRCh38
rs556067208 Chr.12: 74537809-74537809 on Build GRCh38
rs936930666 Chr.12: 74537831-74537831 on Build GRCh38
rs531382734 Chr.12: 74537923-74537923 on Build GRCh38
rs907994518 Chr.12: 74537990-74537990 on Build GRCh38
rs1008724897 Chr.12: 74538082-74538082 on Build GRCh38
rs766431109 Chr.12: 74537758-74537758 on Build GRCh38
rs370062706 Chr.12: 74537793-74537793 on Build GRCh38
rs758961994 Chr.12: 74538147-74538147 on Build GRCh38
rs867792457 Chr.12: 74537860-74537860 on Build GRCh38
rs781336704 Chr.12: 74538167-74538167 on Build GRCh38
rs747077103 Chr.12: 74538152-74538152 on Build GRCh38
rs763836975 Chr.12: 74538123-74538123 on Build GRCh38
rs748199432 Chr.12: 74537731-74537731 on Build GRCh38
rs572762527 Chr.12: 74537813-74537813 on Build GRCh38
rs745505107 Chr.12: 74537902-74537902 on Build GRCh38
rs940693278 Chr.12: 74537847-74537847 on Build GRCh38
rs951738388 Chr.12: 74537767-74537767 on Build GRCh38
rs925565327 Chr.12: 74537817-74537817 on Build GRCh38
rs1046486708 Chr.12: 74537841-74537841 on Build GRCh38
rs967098594 Chr.12: 74537880-74537880 on Build GRCh38
rs564642738 Chr.12: 74537853-74537853 on Build GRCh38
rs904510916 Chr.12: 74537722-74537722 on Build GRCh38
rs768787523 Chr.12: 74538166-74538166 on Build GRCh38
rs925881122 Chr.12: 74538057-74538057 on Build GRCh38
rs539515960 Chr.12: 74537746-74537746 on Build GRCh38
rs916714528 Chr.12: 74537834-74537834 on Build GRCh38
rs994510261 Chr.12: 74537957-74537957 on Build GRCh38
rs541856797 Chr.12: 74537808-74537808 on Build GRCh38
rs768585533 Chr.12: 74538091-74538091 on Build GRCh38
rs770729351 Chr.12: 74538079-74538079 on Build GRCh38
rs963019199 Chr.12: 74537818-74537818 on Build GRCh38
rs1050394759 Chr.12: 74538033-74538033 on Build GRCh38
rs940724233 Chr.12: 74537991-74537991 on Build GRCh38
rs541674359 Chr.12: 74537850-74537850 on Build GRCh38
rs547865714 Chr.12: 74537947-74537947 on Build GRCh38
rs779168712 Chr.12: 74537840-74537840 on Build GRCh38
rs899088773 Chr.12: 74537851-74537851 on Build GRCh38
rs779423791 Chr.12: 74538143-74538143 on Build GRCh38
rs982256371 Chr.12: 74537964-74537964 on Build GRCh38
rs1006968962 Chr.12: 74537854-74537869 on Build GRCh38
rs146565249 Chr.12: 74537859-74537859 on Build GRCh38
rs539551666 Chr.12: 74538099-74538099 on Build GRCh38
rs757635962 Chr.12: 74538137-74538137 on Build GRCh38
rs1055709427 Chr.12: 74537832-74537832 on Build GRCh38
rs1042128016 Chr.12: 74537720-74537720 on Build GRCh38
rs541609461 Chr.12: 74537849-74537849 on Build GRCh38
rs1016216411 Chr.12: 74537724-74537724 on Build GRCh38
rs545874461 Chr.12: 74537861-74537861 on Build GRCh38
rs753600487 Chr.12: 74538130-74538130 on Build GRCh38
rs144298499 Chr.12: 74537717-74537717 on Build GRCh38
rs1038110888 Chr.12: 74538013-74538013 on Build GRCh38
rs1020015427 Chr.12: 74537875-74537875 on Build GRCh38
rs978431484 Chr.12: 74537883-74537883 on Build GRCh38
rs528973117 Chr.12: 74537737-74537737 on Build GRCh38
rs997495353 Chr.12: 74538009-74538009 on Build GRCh38
rs570274148 Chr.12: 74538087-74538087 on Build GRCh38
rs949516847 Chr.12: 74537838-74537838 on Build GRCh38
rs1032644564 Chr.12: 74537891-74537891 on Build GRCh38
rs568376020 Chr.12: 74537975-74537975 on Build GRCh38
rs899143264 Chr.12: 74538014-74538014 on Build GRCh38
rs963326218 Chr.12: 74537736-74537736 on Build GRCh38
rs931906383 Chr.12: 74538016-74538016 on Build GRCh38
rs760144320 Chr.12: 74538107-74538107 on Build GRCh38
rs992560180 Chr.12: 74538017-74538017 on Build GRCh38
rs900180644 Chr.12: 74537821-74537821 on Build GRCh38
rs750854810 Chr.12: 74538145-74538145 on Build GRCh38
rs890350952 Chr.12: 74538053-74538053 on Build GRCh38
rs527558664 Chr.12: 74538039-74538039 on Build GRCh38
rs780471674 Chr.12: 74538148-74538148 on Build GRCh38
rs777975360 Chr.12: 74537955-74537955 on Build GRCh38
rs967236591 Chr.12: 74538032-74538032 on Build GRCh38

View Full Product Details