Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119667865-119668365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKAB1
Entrez Gene ID
5564
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 1
Gene Aliases
AMPK, HAMPKb
UniGene
Hs.741184
Gene Chromosome Location
Chr.12: 119667864-119681624 on Build GRCh38
Transcripts
NM_006253.4 XM_005253909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1578177 c.38G>T p.R13L Chr.12: 119668282-119668282 on Build GRCh38
7281647 c.99G>A p.R33R Chr.12: 119668343-119668343 on Build GRCh38
546780 c.111G>A p.L37L Chr.12: 119668355-119668355 on Build GRCh38
295295 c.74_75insG p.T28fs*39 Chr.12: 119668318-119668319 on Build GRCh38
1221991 c.107T>A p.I36N Chr.12: 119668351-119668351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778255982 Chr.12: 119668274-119668274 on Build GRCh38
rs983401785 Chr.12: 119668030-119668030 on Build GRCh38
rs771224582 Chr.12: 119668281-119668281 on Build GRCh38
rs6490265 Chr.12: 119667902-119667902 on Build GRCh38
rs774447370 Chr.12: 119668224-119668224 on Build GRCh38
rs762274137 Chr.12: 119668318-119668318 on Build GRCh38
rs199820598 Chr.12: 119668000-119668000 on Build GRCh38
rs78433950 Chr.12: 119668317-119668317 on Build GRCh38
rs768381257 Chr.12: 119668242-119668243 on Build GRCh38
rs761626973 Chr.12: 119668323-119668323 on Build GRCh38
rs191154896 Chr.12: 119668235-119668235 on Build GRCh38
rs375269404 Chr.12: 119668038-119668038 on Build GRCh38
rs1049991491 Chr.12: 119668071-119668071 on Build GRCh38
rs116752243 Chr.12: 119668331-119668331 on Build GRCh38
rs750954971 Chr.12: 119668344-119668364 on Build GRCh38
rs1004075103 Chr.12: 119668170-119668170 on Build GRCh38
rs764188990 Chr.12: 119668242-119668242 on Build GRCh38
rs761521273 Chr.12: 119668249-119668249 on Build GRCh38
rs921729118 Chr.12: 119668052-119668052 on Build GRCh38
rs773552804 Chr.12: 119668209-119668209 on Build GRCh38
rs935834370 Chr.12: 119668053-119668053 on Build GRCh38
rs757108077 Chr.12: 119667926-119667926 on Build GRCh38
rs3182909 Chr.12: 119668108-119668108 on Build GRCh38
rs895294802 Chr.12: 119668150-119668150 on Build GRCh38
rs983592604 Chr.12: 119668222-119668222 on Build GRCh38
rs532572076 Chr.12: 119668238-119668238 on Build GRCh38
rs544379312 Chr.12: 119668168-119668168 on Build GRCh38
rs779157494 Chr.12: 119668282-119668282 on Build GRCh38
rs373617259 Chr.12: 119668271-119668271 on Build GRCh38
rs767094765 Chr.12: 119667894-119667894 on Build GRCh38
rs6490266 Chr.12: 119667935-119667935 on Build GRCh38
rs1054184364 Chr.12: 119668061-119668061 on Build GRCh38
rs760667193 Chr.12: 119668240-119668240 on Build GRCh38
rs956361157 Chr.12: 119667964-119667964 on Build GRCh38
rs945808882 Chr.12: 119668074-119668074 on Build GRCh38
rs950826316 Chr.12: 119668009-119668009 on Build GRCh38
rs544140980 Chr.12: 119668101-119668101 on Build GRCh38
rs772077116 Chr.12: 119668236-119668236 on Build GRCh38
rs373680495 Chr.12: 119668340-119668340 on Build GRCh38
rs1027806748 Chr.12: 119668188-119668188 on Build GRCh38
rs1003833718 Chr.12: 119667910-119667910 on Build GRCh38
rs149171538 Chr.12: 119668025-119668025 on Build GRCh38
rs776725274 Chr.12: 119668320-119668320 on Build GRCh38
rs772385438 Chr.12: 119668294-119668294 on Build GRCh38
rs1043352 Chr.12: 119668268-119668268 on Build GRCh38
rs759614211 Chr.12: 119668231-119668231 on Build GRCh38
rs541199406 Chr.12: 119667984-119667984 on Build GRCh38
rs1043350 Chr.12: 119668265-119668265 on Build GRCh38
rs968390572 Chr.12: 119667929-119667929 on Build GRCh38
rs760209495 Chr.12: 119668218-119668218 on Build GRCh38
rs758106654 Chr.12: 119668267-119668267 on Build GRCh38
rs775753265 Chr.12: 119668237-119668237 on Build GRCh38
rs1045803522 Chr.12: 119668076-119668076 on Build GRCh38
rs777079156 Chr.12: 119668305-119668317 on Build GRCh38
rs1022618315 Chr.12: 119667917-119667917 on Build GRCh38
rs1033948696 Chr.12: 119668111-119668111 on Build GRCh38
rs749718531 Chr.12: 119668210-119668210 on Build GRCh38
rs950793348 Chr.12: 119668190-119668190 on Build GRCh38
rs766087073 Chr.12: 119668335-119668335 on Build GRCh38
rs994946202 Chr.12: 119668182-119668182 on Build GRCh38
rs574994287 Chr.12: 119668049-119668049 on Build GRCh38
rs572621920 Chr.12: 119667983-119667983 on Build GRCh38
rs61756440 Chr.12: 119668309-119668309 on Build GRCh38
rs1023996478 Chr.12: 119667892-119667892 on Build GRCh38
rs765766033 Chr.12: 119668318-119668319 on Build GRCh38
rs1043354 Chr.12: 119668302-119668302 on Build GRCh38
rs1056958867 Chr.12: 119668171-119668171 on Build GRCh38
rs909864646 Chr.12: 119668072-119668072 on Build GRCh38
rs771116896 Chr.12: 119668221-119668221 on Build GRCh38
rs944616273 Chr.12: 119668040-119668040 on Build GRCh38
rs768651620 Chr.12: 119668319-119668319 on Build GRCh38
rs61748075 Chr.12: 119668324-119668324 on Build GRCh38
rs749595403 Chr.12: 119668275-119668275 on Build GRCh38
rs1062688 Chr.12: 119668289-119668289 on Build GRCh38
rs906907609 Chr.12: 119668078-119668078 on Build GRCh38
rs375442599 Chr.12: 119668272-119668272 on Build GRCh38
rs973429977 Chr.12: 119667957-119667957 on Build GRCh38
rs913104801 Chr.12: 119668333-119668333 on Build GRCh38
rs976942269 Chr.12: 119667975-119667975 on Build GRCh38
rs764888871 Chr.12: 119668259-119668259 on Build GRCh38
rs186775330 Chr.12: 119667931-119667931 on Build GRCh38
rs746047315 Chr.12: 119668287-119668287 on Build GRCh38
rs34062789 Chr.12: 119667924-119667924 on Build GRCh38
rs891534901 Chr.12: 119668069-119668069 on Build GRCh38

View Full Product Details