Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120321732-120322273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G1B
Entrez Gene ID
5319
Gene Name
phospholipase A2 group IB
Gene Aliases
PLA2, PLA2A, PPLA2
UniGene
Hs.992
Gene Chromosome Location
Chr.12: 120322111-120327789 on Build GRCh38
Transcripts
NM_000928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1220838 c.401A>C p.N134T Chr.12: 120322239-120322239 on Build GRCh38
5560731 c.416A>T p.N139I Chr.12: 120322224-120322224 on Build GRCh38
6747678 c.406G>A p.A136T Chr.12: 120322234-120322234 on Build GRCh38
3416549 c.418C>T p.L140L Chr.12: 120322222-120322222 on Build GRCh38
3457035 c.421G>A p.D141N Chr.12: 120322219-120322219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906544415 Chr.12: 120321965-120321965 on Build GRCh38
rs1053618773 Chr.12: 120321939-120321939 on Build GRCh38
rs760255034 Chr.12: 120321802-120321802 on Build GRCh38
rs750816394 Chr.12: 120322244-120322244 on Build GRCh38
rs201478589 Chr.12: 120322209-120322209 on Build GRCh38
rs754918548 Chr.12: 120322161-120322161 on Build GRCh38
rs562014823 Chr.12: 120321995-120321995 on Build GRCh38
rs1004860438 Chr.12: 120321970-120321970 on Build GRCh38
rs935190746 Chr.12: 120321902-120321902 on Build GRCh38
rs572752895 Chr.12: 120321854-120321854 on Build GRCh38
rs772864413 Chr.12: 120322202-120322202 on Build GRCh38
rs1029161456 Chr.12: 120322086-120322086 on Build GRCh38
rs774996738 Chr.12: 120321786-120321786 on Build GRCh38
rs377217390 Chr.12: 120322152-120322152 on Build GRCh38
rs912212572 Chr.12: 120321893-120321893 on Build GRCh38
rs902342945 Chr.12: 120321756-120321756 on Build GRCh38
rs1054131337 Chr.12: 120321760-120321760 on Build GRCh38
rs936431935 Chr.12: 120322204-120322204 on Build GRCh38
rs933016934 Chr.12: 120321941-120321941 on Build GRCh38
rs979398960 Chr.12: 120321878-120321878 on Build GRCh38
rs922188556 Chr.12: 120321855-120321855 on Build GRCh38
rs968769844 Chr.12: 120322182-120322182 on Build GRCh38
rs923670030 Chr.12: 120321929-120321929 on Build GRCh38
rs765635237 Chr.12: 120322243-120322243 on Build GRCh38
rs752179059 Chr.12: 120322192-120322192 on Build GRCh38
rs570949748 Chr.12: 120322140-120322140 on Build GRCh38
rs9300310 Chr.12: 120321793-120321793 on Build GRCh38
rs780067104 Chr.12: 120322153-120322153 on Build GRCh38
rs748871554 Chr.12: 120322208-120322208 on Build GRCh38
rs879272753 Chr.12: 120321889-120321889 on Build GRCh38
rs377237459 Chr.12: 120322233-120322233 on Build GRCh38
rs1042133856 Chr.12: 120321895-120321895 on Build GRCh38
rs1004956763 Chr.12: 120321800-120321800 on Build GRCh38
rs1035789206 Chr.12: 120322100-120322100 on Build GRCh38
rs758559977 Chr.12: 120322147-120322147 on Build GRCh38
rs1012146604 Chr.12: 120321959-120321959 on Build GRCh38
rs35842522 Chr.12: 120321871-120321871 on Build GRCh38
rs369669070 Chr.12: 120322170-120322170 on Build GRCh38
rs771672801 Chr.12: 120321764-120321764 on Build GRCh38
rs183612258 Chr.12: 120322157-120322157 on Build GRCh38
rs139911957 Chr.12: 120322094-120322094 on Build GRCh38
rs1000675828 Chr.12: 120321759-120321759 on Build GRCh38
rs559841330 Chr.12: 120321888-120321888 on Build GRCh38
rs192443232 Chr.12: 120321792-120321792 on Build GRCh38
rs987670816 Chr.12: 120322125-120322125 on Build GRCh38
rs965032731 Chr.12: 120322145-120322145 on Build GRCh38
rs566373968 Chr.12: 120321992-120321992 on Build GRCh38
rs763768506 Chr.12: 120321845-120321845 on Build GRCh38
rs1052259645 Chr.12: 120321924-120321924 on Build GRCh38
rs2066539 Chr.12: 120322126-120322126 on Build GRCh38
rs544894226 Chr.12: 120322048-120322048 on Build GRCh38
rs528842150 Chr.12: 120321963-120321963 on Build GRCh38
rs888758834 Chr.12: 120322011-120322011 on Build GRCh38
rs943819645 Chr.12: 120321894-120321894 on Build GRCh38
rs995450489 Chr.12: 120322012-120322012 on Build GRCh38
rs373127940 Chr.12: 120322211-120322211 on Build GRCh38
rs766767485 Chr.12: 120322172-120322172 on Build GRCh38
rs763427130 Chr.12: 120322029-120322029 on Build GRCh38
rs750741390 Chr.12: 120321931-120321931 on Build GRCh38
rs970824617 Chr.12: 120321799-120321799 on Build GRCh38
rs895093864 Chr.12: 120321949-120321949 on Build GRCh38
rs764541305 Chr.12: 120322217-120322217 on Build GRCh38
rs777230285 Chr.12: 120322223-120322223 on Build GRCh38
rs546759136 Chr.12: 120322166-120322166 on Build GRCh38
rs762286318 Chr.12: 120322242-120322242 on Build GRCh38
rs113744270 Chr.12: 120322068-120322068 on Build GRCh38
rs923720236 Chr.12: 120322194-120322194 on Build GRCh38
rs1016540526 Chr.12: 120322030-120322030 on Build GRCh38

View Full Product Details