Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121793354-121793857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOF
Entrez Gene ID
54509
Gene Name
ras homolog family member F, filopodia associated
Gene Aliases
ARHF, RIF
UniGene
Hs.707579
Gene Chromosome Location
Chr.12: 121777754-121793688 on Build GRCh38
Transcripts
NM_019034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4957958 c.48C>A p.G16G Chr.12: 121793586-121793586 on Build GRCh38
6206712 c.114G>A p.V38V Chr.12: 121793520-121793520 on Build GRCh38
4643660 c.54G>T p.K18N Chr.12: 121793580-121793580 on Build GRCh38
6391397 c.101C>A p.S34* Chr.12: 121793533-121793533 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045406479 Chr.12: 121793816-121793816 on Build GRCh38
rs866165056 Chr.12: 121793611-121793611 on Build GRCh38
rs1028270524 Chr.12: 121793671-121793671 on Build GRCh38
rs919711911 Chr.12: 121793815-121793815 on Build GRCh38
rs1041890795 Chr.12: 121793487-121793487 on Build GRCh38
rs762475526 Chr.12: 121793549-121793549 on Build GRCh38
rs906042505 Chr.12: 121793708-121793708 on Build GRCh38
rs936196811 Chr.12: 121793750-121793750 on Build GRCh38
rs773839153 Chr.12: 121793684-121793684 on Build GRCh38
rs761590215 Chr.12: 121793535-121793535 on Build GRCh38
rs780256764 Chr.12: 121793627-121793627 on Build GRCh38
rs1046675690 Chr.12: 121793835-121793835 on Build GRCh38
rs538768246 Chr.12: 121793677-121793677 on Build GRCh38
rs886659813 Chr.12: 121793695-121793695 on Build GRCh38
rs747021964 Chr.12: 121793638-121793638 on Build GRCh38
rs1020303491 Chr.12: 121793772-121793772 on Build GRCh38
rs929736870 Chr.12: 121793821-121793821 on Build GRCh38
rs963935695 Chr.12: 121793814-121793814 on Build GRCh38
rs372836309 Chr.12: 121793556-121793556 on Build GRCh38
rs769533226 Chr.12: 121793663-121793663 on Build GRCh38
rs772797329 Chr.12: 121793544-121793544 on Build GRCh38
rs1055875713 Chr.12: 121793758-121793758 on Build GRCh38
rs865802827 Chr.12: 121793579-121793579 on Build GRCh38
rs1010274402 Chr.12: 121793764-121793764 on Build GRCh38
rs955428209 Chr.12: 121793778-121793779 on Build GRCh38
rs957384959 Chr.12: 121793470-121793470 on Build GRCh38
rs921177092 Chr.12: 121793429-121793429 on Build GRCh38
rs776274227 Chr.12: 121793528-121793528 on Build GRCh38
rs772229293 Chr.12: 121793472-121793472 on Build GRCh38
rs926034892 Chr.12: 121793730-121793730 on Build GRCh38
rs988707855 Chr.12: 121793489-121793489 on Build GRCh38
rs1056011524 Chr.12: 121793589-121793589 on Build GRCh38
rs2707067 Chr.12: 121793462-121793462 on Build GRCh38
rs977941386 Chr.12: 121793718-121793718 on Build GRCh38
rs911839561 Chr.12: 121793689-121793689 on Build GRCh38
rs747912966 Chr.12: 121793662-121793662 on Build GRCh38
rs1033116539 Chr.12: 121793715-121793715 on Build GRCh38
rs866721189 Chr.12: 121793557-121793557 on Build GRCh38
rs946097709 Chr.12: 121793478-121793478 on Build GRCh38
rs764579464 Chr.12: 121793600-121793600 on Build GRCh38
rs754489189 Chr.12: 121793509-121793509 on Build GRCh38
rs778327921 Chr.12: 121793626-121793626 on Build GRCh38
rs1013949036 Chr.12: 121793620-121793620 on Build GRCh38
rs867910062 Chr.12: 121793590-121793590 on Build GRCh38
rs974228853 Chr.12: 121793814-121793814 on Build GRCh38
rs754149985 Chr.12: 121793603-121793603 on Build GRCh38
rs776536269 Chr.12: 121793595-121793595 on Build GRCh38
rs868826164 Chr.12: 121793571-121793571 on Build GRCh38
rs932011868 Chr.12: 121793448-121793448 on Build GRCh38
rs772886812 Chr.12: 121793665-121793665 on Build GRCh38
rs757583668 Chr.12: 121793613-121793613 on Build GRCh38
rs556990730 Chr.12: 121793788-121793788 on Build GRCh38
rs986753523 Chr.12: 121793810-121793810 on Build GRCh38
rs1002009725 Chr.12: 121793713-121793713 on Build GRCh38
rs987004474 Chr.12: 121793607-121793607 on Build GRCh38
rs775591084 Chr.12: 121793427-121793427 on Build GRCh38
rs916175995 Chr.12: 121793760-121793760 on Build GRCh38
rs2707066 Chr.12: 121793667-121793667 on Build GRCh38
rs780170687 Chr.12: 121793501-121793501 on Build GRCh38
rs200285606 Chr.12: 121793459-121793459 on Build GRCh38
rs768469622 Chr.12: 121793643-121793643 on Build GRCh38
rs530694396 Chr.12: 121793436-121793436 on Build GRCh38
rs768542510 Chr.12: 121793515-121793515 on Build GRCh38
rs886881447 Chr.12: 121793630-121793630 on Build GRCh38
rs911033334 Chr.12: 121793811-121793811 on Build GRCh38
rs1046298026 Chr.12: 121793704-121793708 on Build GRCh38
rs950324180 Chr.12: 121793806-121793806 on Build GRCh38
rs779299932 Chr.12: 121793615-121793615 on Build GRCh38
rs1014513872 Chr.12: 121793699-121793699 on Build GRCh38
rs777970145 Chr.12: 121793458-121793458 on Build GRCh38
rs769331970 Chr.12: 121793543-121793543 on Build GRCh38
rs567549599 Chr.12: 121793461-121793461 on Build GRCh38
rs1024486309 Chr.12: 121793416-121793416 on Build GRCh38
rs776923916 Chr.12: 121793593-121793619 on Build GRCh38
rs1016776065 Chr.12: 121793646-121793646 on Build GRCh38
rs746910314 Chr.12: 121793508-121793508 on Build GRCh38
rs750632490 Chr.12: 121793621-121793621 on Build GRCh38
rs967495480 Chr.12: 121793697-121793697 on Build GRCh38
rs781113978 Chr.12: 121793654-121793654 on Build GRCh38
rs773704722 Chr.12: 121793465-121793465 on Build GRCh38
rs761065704 Chr.12: 121793599-121793599 on Build GRCh38

View Full Product Details