Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122854970-122855471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1R
Entrez Gene ID
9026
Gene Name
huntingtin interacting protein 1 related
Gene Aliases
HIP12, HIP3, ILWEQ
UniGene
Hs.524815
Gene Chromosome Location
Chr.12: 122834498-122862961 on Build GRCh38
Transcripts
NM_001303097.1 NM_001303099.1 NM_003959.2 XM_011538963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398476 c.979G>A p.A327T Chr.12: 122855391-122855391 on Build GRCh38
2225431 c.923A>C p.E308A Chr.12: 122855335-122855335 on Build GRCh38
5643686 c.823C>T p.R275W Chr.12: 122855099-122855099 on Build GRCh38
6553748 c.848C>T p.P283L Chr.12: 122855124-122855124 on Build GRCh38
240111 c.943C>T p.P315S Chr.12: 122855355-122855355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202129877 Chr.12: 122855342-122855342 on Build GRCh38
rs763121106 Chr.12: 122855092-122855092 on Build GRCh38
rs747842648 Chr.12: 122854997-122854997 on Build GRCh38
rs377744238 Chr.12: 122855152-122855152 on Build GRCh38
rs761059259 Chr.12: 122855223-122855223 on Build GRCh38
rs772380417 Chr.12: 122855304-122855304 on Build GRCh38
rs773944111 Chr.12: 122854995-122854995 on Build GRCh38
rs761161357 Chr.12: 122855314-122855314 on Build GRCh38
rs767873296 Chr.12: 122855110-122855110 on Build GRCh38
rs773113409 Chr.12: 122855071-122855071 on Build GRCh38
rs373133716 Chr.12: 122855418-122855418 on Build GRCh38
rs994255657 Chr.12: 122855098-122855098 on Build GRCh38
rs748817714 Chr.12: 122855053-122855053 on Build GRCh38
rs756990237 Chr.12: 122855255-122855255 on Build GRCh38
rs746799828 Chr.12: 122855135-122855135 on Build GRCh38
rs934497920 Chr.12: 122855322-122855322 on Build GRCh38
rs374440993 Chr.12: 122855126-122855126 on Build GRCh38
rs747204142 Chr.12: 122855401-122855401 on Build GRCh38
rs779717548 Chr.12: 122855363-122855363 on Build GRCh38
rs759587881 Chr.12: 122855293-122855296 on Build GRCh38
rs771885860 Chr.12: 122855069-122855069 on Build GRCh38
rs776752371 Chr.12: 122855010-122855010 on Build GRCh38
rs879238989 Chr.12: 122855399-122855399 on Build GRCh38
rs373872066 Chr.12: 122855028-122855028 on Build GRCh38
rs749175357 Chr.12: 122855366-122855366 on Build GRCh38
rs758416618 Chr.12: 122855191-122855191 on Build GRCh38
rs751899254 Chr.12: 122855105-122855105 on Build GRCh38
rs2292138 Chr.12: 122855165-122855165 on Build GRCh38
rs960758328 Chr.12: 122855019-122855019 on Build GRCh38
rs897291537 Chr.12: 122855424-122855424 on Build GRCh38
rs774437296 Chr.12: 122855299-122855299 on Build GRCh38
rs1017046874 Chr.12: 122855193-122855193 on Build GRCh38
rs575758377 Chr.12: 122855131-122855131 on Build GRCh38
rs778934790 Chr.12: 122855385-122855385 on Build GRCh38
rs960958507 Chr.12: 122855267-122855267 on Build GRCh38
rs756003322 Chr.12: 122855357-122855357 on Build GRCh38
rs769798559 Chr.12: 122855150-122855150 on Build GRCh38
rs199750482 Chr.12: 122855133-122855133 on Build GRCh38
rs772173203 Chr.12: 122855391-122855391 on Build GRCh38
rs775959948 Chr.12: 122855169-122855169 on Build GRCh38
rs766795987 Chr.12: 122855315-122855315 on Build GRCh38
rs934550383 Chr.12: 122855117-122855117 on Build GRCh38
rs771935175 Chr.12: 122854999-122854999 on Build GRCh38
rs530434049 Chr.12: 122855425-122855425 on Build GRCh38
rs767072670 Chr.12: 122855435-122855435 on Build GRCh38
rs1018277361 Chr.12: 122855392-122855392 on Build GRCh38
rs758052653 Chr.12: 122855248-122855248 on Build GRCh38
rs757728991 Chr.12: 122855030-122855030 on Build GRCh38
rs773429443 Chr.12: 122855310-122855310 on Build GRCh38
rs201464595 Chr.12: 122855031-122855031 on Build GRCh38
rs200261764 Chr.12: 122855125-122855125 on Build GRCh38
rs773554395 Chr.12: 122855398-122855398 on Build GRCh38
rs116483556 Chr.12: 122855254-122855254 on Build GRCh38
rs756825361 Chr.12: 122855341-122855341 on Build GRCh38
rs768664886 Chr.12: 122855297-122855297 on Build GRCh38
rs761984492 Chr.12: 122855303-122855303 on Build GRCh38
rs139745552 Chr.12: 122855330-122855330 on Build GRCh38
rs763067680 Chr.12: 122855161-122855161 on Build GRCh38
rs745714905 Chr.12: 122855258-122855258 on Build GRCh38
rs776495362 Chr.12: 122855084-122855084 on Build GRCh38
rs956529655 Chr.12: 122855443-122855443 on Build GRCh38
rs768852610 Chr.12: 122855162-122855162 on Build GRCh38
rs766961864 Chr.12: 122855228-122855228 on Build GRCh38
rs752245775 Chr.12: 122855245-122855245 on Build GRCh38
rs773974604 Chr.12: 122855426-122855426 on Build GRCh38
rs778500564 Chr.12: 122855059-122855059 on Build GRCh38
rs769594202 Chr.12: 122855264-122855264 on Build GRCh38
rs759518295 Chr.12: 122855085-122855085 on Build GRCh38
rs756668419 Chr.12: 122855118-122855118 on Build GRCh38
rs765488425 Chr.12: 122855012-122855012 on Build GRCh38
rs553363533 Chr.12: 122855004-122855004 on Build GRCh38
rs766464581 Chr.12: 122855005-122855005 on Build GRCh38
rs760129648 Chr.12: 122855327-122855327 on Build GRCh38
rs367922574 Chr.12: 122855356-122855356 on Build GRCh38
rs371091485 Chr.12: 122855256-122855256 on Build GRCh38
rs748278918 Chr.12: 122855390-122855390 on Build GRCh38
rs752271388 Chr.12: 122855371-122855371 on Build GRCh38
rs370429031 Chr.12: 122855015-122855015 on Build GRCh38
rs780096407 Chr.12: 122855279-122855279 on Build GRCh38
rs764390540 Chr.12: 122855024-122855024 on Build GRCh38
rs770779949 Chr.12: 122855138-122855138 on Build GRCh38
rs375196602 Chr.12: 122855215-122855215 on Build GRCh38
rs373603682 Chr.12: 122855430-122855430 on Build GRCh38
rs561368690 Chr.12: 122855139-122855139 on Build GRCh38
rs202162558 Chr.12: 122855018-122855018 on Build GRCh38
rs926314031 Chr.12: 122855274-122855274 on Build GRCh38
rs374098037 Chr.12: 122855331-122855331 on Build GRCh38
rs777423116 Chr.12: 122855249-122855249 on Build GRCh38
rs777153003 Chr.12: 122855412-122855412 on Build GRCh38
rs777216088 Chr.12: 122855318-122855318 on Build GRCh38
rs1053036650 Chr.12: 122855134-122855134 on Build GRCh38
rs1005569730 Chr.12: 122855145-122855145 on Build GRCh38
rs749364683 Chr.12: 122855284-122855284 on Build GRCh38
rs765170780 Chr.12: 122855087-122855087 on Build GRCh38
rs770939437 Chr.12: 122855131-122855173 on Build GRCh38
rs375709915 Chr.12: 122855080-122855080 on Build GRCh38
rs763513962 Chr.12: 122855173-122855173 on Build GRCh38
rs994751614 Chr.12: 122855337-122855337 on Build GRCh38
rs200483656 Chr.12: 122855002-122855002 on Build GRCh38
rs766909159 Chr.12: 122855349-122855351 on Build GRCh38
rs998112715 Chr.12: 122855420-122855420 on Build GRCh38
rs775658240 Chr.12: 122855088-122855088 on Build GRCh38
rs370031826 Chr.12: 122855109-122855109 on Build GRCh38
rs767725095 Chr.12: 122855178-122855178 on Build GRCh38
rs541234984 Chr.12: 122855198-122855198 on Build GRCh38
rs770982695 Chr.12: 122855081-122855081 on Build GRCh38
rs184197853 Chr.12: 122855368-122855368 on Build GRCh38
rs771266003 Chr.12: 122855411-122855411 on Build GRCh38
rs987871557 Chr.12: 122855213-122855213 on Build GRCh38
rs747708374 Chr.12: 122855062-122855062 on Build GRCh38
rs768331765 Chr.12: 122854994-122854994 on Build GRCh38
rs759596325 Chr.12: 122855011-122855011 on Build GRCh38
rs754379342 Chr.12: 122855054-122855054 on Build GRCh38
rs941087386 Chr.12: 122855397-122855397 on Build GRCh38
rs750838323 Chr.12: 122855111-122855111 on Build GRCh38
rs774146563 Chr.12: 122855147-122855165 on Build GRCh38
rs762037321 Chr.12: 122855168-122855168 on Build GRCh38

View Full Product Details