Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21001600-21002106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A2
Entrez Gene ID
29986
Gene Name
solute carrier family 39 member 2
Gene Aliases
6A1, ETI-1, ZIP-2, ZIP2
UniGene
Hs.175783
Gene Chromosome Location
Chr.14: 20999255-21001875 on Build GRCh38
Transcripts
NM_001256588.1 NM_014579.3

SNP Detail

SNP ID Chromosome Location
rs996395854 Chr.14: 21001969-21001969 on Build GRCh38
rs971816418 Chr.14: 21002052-21002052 on Build GRCh38
rs1020276664 Chr.14: 21001710-21001710 on Build GRCh38
rs756074753 Chr.14: 21002006-21002006 on Build GRCh38
rs750284942 Chr.14: 21001959-21001959 on Build GRCh38
rs547272051 Chr.14: 21002062-21002062 on Build GRCh38
rs898882723 Chr.14: 21001878-21001878 on Build GRCh38
rs995032607 Chr.14: 21001884-21001884 on Build GRCh38
rs950898072 Chr.14: 21001924-21001924 on Build GRCh38
rs927454364 Chr.14: 21001808-21001808 on Build GRCh38
rs1007142926 Chr.14: 21001833-21001833 on Build GRCh38
rs547854624 Chr.14: 21001861-21001861 on Build GRCh38
rs564744990 Chr.14: 21001739-21001739 on Build GRCh38
rs865915006 Chr.14: 21001990-21001990 on Build GRCh38
rs893048170 Chr.14: 21002010-21002010 on Build GRCh38
rs572528359 Chr.14: 21001682-21001682 on Build GRCh38
rs1051624119 Chr.14: 21001721-21001721 on Build GRCh38
rs531223778 Chr.14: 21001830-21001830 on Build GRCh38
rs374678543 Chr.14: 21001754-21001755 on Build GRCh38
rs137875877 Chr.14: 21001801-21001801 on Build GRCh38
rs1047396383 Chr.14: 21001887-21001887 on Build GRCh38
rs1020217915 Chr.14: 21002040-21002040 on Build GRCh38
rs1040542621 Chr.14: 21001926-21001926 on Build GRCh38
rs934608616 Chr.14: 21001689-21001689 on Build GRCh38
rs142380686 Chr.14: 21001922-21001922 on Build GRCh38
rs562104758 Chr.14: 21001814-21001814 on Build GRCh38
rs767399112 Chr.14: 21001782-21001782 on Build GRCh38
rs1003829824 Chr.14: 21001925-21001925 on Build GRCh38
rs1010190035 Chr.14: 21001704-21001704 on Build GRCh38
rs957308824 Chr.14: 21001679-21001679 on Build GRCh38
rs1038792082 Chr.14: 21001855-21001855 on Build GRCh38
rs183186857 Chr.14: 21001686-21001686 on Build GRCh38
rs779913837 Chr.14: 21002073-21002073 on Build GRCh38
rs374216878 Chr.14: 21001886-21001886 on Build GRCh38
rs2234640 Chr.14: 21001638-21001638 on Build GRCh38
rs757071612 Chr.14: 21001763-21001763 on Build GRCh38
rs915340593 Chr.14: 21002075-21002075 on Build GRCh38
rs947248010 Chr.14: 21001629-21001629 on Build GRCh38
rs990784970 Chr.14: 21002022-21002022 on Build GRCh38
rs974290741 Chr.14: 21001831-21001832 on Build GRCh38
rs924451673 Chr.14: 21001645-21001645 on Build GRCh38
rs186382977 Chr.14: 21001992-21001992 on Build GRCh38
rs562183286 Chr.14: 21001665-21001665 on Build GRCh38
rs533456934 Chr.14: 21001791-21001791 on Build GRCh38
rs900279165 Chr.14: 21001931-21001931 on Build GRCh38

View Full Product Details