Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19312531-19313030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PQLC2
Entrez Gene ID
54896
Gene Name
PQ loop repeat containing 2
Gene Aliases
-
UniGene
Hs.647620
Gene Chromosome Location
Chr.1: 19312246-19332611 on Build GRCh38
Transcripts
NM_001040125.1 NM_001040126.1 NM_001287531.1 NM_017765.2 NR_109848.1 XM_005245916.2 XM_006710710.2 XM_006710711.2 XM_011541634.2

SNP Detail

SNP ID Chromosome Location
rs1035116369 Chr.1: 19312987-19312987 on Build GRCh38
rs930677060 Chr.1: 19312608-19312608 on Build GRCh38
rs183381823 Chr.1: 19312808-19312808 on Build GRCh38
rs901077021 Chr.1: 19312732-19312732 on Build GRCh38
rs2231192 Chr.1: 19312815-19312815 on Build GRCh38
rs577136557 Chr.1: 19312804-19312804 on Build GRCh38
rs983484622 Chr.1: 19312570-19312570 on Build GRCh38
rs984998304 Chr.1: 19312870-19312870 on Build GRCh38
rs534120214 Chr.1: 19312709-19312709 on Build GRCh38
rs1010908783 Chr.1: 19312688-19312688 on Build GRCh38
rs907670726 Chr.1: 19312576-19312576 on Build GRCh38
rs368611758 Chr.1: 19312949-19312949 on Build GRCh38
rs975824397 Chr.1: 19312868-19312868 on Build GRCh38
rs2231194 Chr.1: 19312594-19312594 on Build GRCh38
rs930338149 Chr.1: 19312553-19312553 on Build GRCh38
rs139369121 Chr.1: 19312818-19312818 on Build GRCh38
rs1034875039 Chr.1: 19312802-19312802 on Build GRCh38
rs933874759 Chr.1: 19312950-19312950 on Build GRCh38
rs940800364 Chr.1: 19312901-19312901 on Build GRCh38
rs916717588 Chr.1: 19312861-19312861 on Build GRCh38
rs868860317 Chr.1: 19312780-19312780 on Build GRCh38
rs1050025238 Chr.1: 19312658-19312658 on Build GRCh38
rs2231191 Chr.1: 19312995-19312995 on Build GRCh38
rs41264063 Chr.1: 19312975-19312975 on Build GRCh38
rs11558391 Chr.1: 19312779-19312779 on Build GRCh38
rs944485605 Chr.1: 19312866-19312866 on Build GRCh38
rs192385011 Chr.1: 19312604-19312604 on Build GRCh38
rs1049624052 Chr.1: 19312964-19312964 on Build GRCh38
rs940700132 Chr.1: 19312706-19312706 on Build GRCh38
rs568593681 Chr.1: 19312830-19312830 on Build GRCh38
rs932790396 Chr.1: 19312620-19312620 on Build GRCh38
rs2231193 Chr.1: 19312645-19312645 on Build GRCh38
rs1050309767 Chr.1: 19312944-19312944 on Build GRCh38
rs889623366 Chr.1: 19312792-19312792 on Build GRCh38
rs538330094 Chr.1: 19312807-19312807 on Build GRCh38
rs754139057 Chr.1: 19312670-19312670 on Build GRCh38
rs554407192 Chr.1: 19312875-19312875 on Build GRCh38
rs998076817 Chr.1: 19312753-19312753 on Build GRCh38
rs1041226525 Chr.1: 19312596-19312596 on Build GRCh38
rs558779341 Chr.1: 19312786-19312786 on Build GRCh38
rs548416558 Chr.1: 19312684-19312684 on Build GRCh38
rs11585809 Chr.1: 19312738-19312738 on Build GRCh38
rs895186797 Chr.1: 19312993-19312993 on Build GRCh38
rs111843204 Chr.1: 19312817-19312817 on Build GRCh38
rs566661563 Chr.1: 19312704-19312704 on Build GRCh38
rs1042439925 Chr.1: 19312973-19312973 on Build GRCh38
rs546135880 Chr.1: 19312989-19312989 on Build GRCh38
rs1003520773 Chr.1: 19312982-19312982 on Build GRCh38
rs946949250 Chr.1: 19312962-19312962 on Build GRCh38
rs991074726 Chr.1: 19312854-19312854 on Build GRCh38
rs921659234 Chr.1: 19312871-19312871 on Build GRCh38
rs928217360 Chr.1: 19312565-19312565 on Build GRCh38
rs889738224 Chr.1: 19312970-19312970 on Build GRCh38
rs1003615690 Chr.1: 19312797-19312797 on Build GRCh38

View Full Product Details