Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41564106-41564612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYRO3
Entrez Gene ID
7301
Gene Name
TYRO3 protein tyrosine kinase
Gene Aliases
BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif
UniGene
Hs.381282
Gene Chromosome Location
Chr.15: 41559022-41579338 on Build GRCh38
Transcripts
NM_006293.3 XM_017022542.1 XM_017022543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6814696 c.625A>G p.K209E Chr.15: 41564228-41564228 on Build GRCh38
7146955 c.622C>A p.L208I Chr.15: 41564225-41564225 on Build GRCh38
961464 c.644G>A p.R215H Chr.15: 41564247-41564247 on Build GRCh38
4738539 c.654T>G p.T218T Chr.15: 41564257-41564257 on Build GRCh38
7169888 c.667+1G>C p.? Chr.15: 41564271-41564271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375507876 Chr.15: 41564202-41564202 on Build GRCh38
rs900174955 Chr.15: 41564197-41564197 on Build GRCh38
rs763708051 Chr.15: 41564138-41564138 on Build GRCh38
rs76520897 Chr.15: 41564226-41564226 on Build GRCh38
rs766022527 Chr.15: 41564405-41564405 on Build GRCh38
rs756213024 Chr.15: 41564317-41564317 on Build GRCh38
rs376621156 Chr.15: 41564139-41564139 on Build GRCh38
rs1013487428 Chr.15: 41564367-41564367 on Build GRCh38
rs868617296 Chr.15: 41564535-41564535 on Build GRCh38
rs997712401 Chr.15: 41564134-41564134 on Build GRCh38
rs369311519 Chr.15: 41564219-41564219 on Build GRCh38
rs776248558 Chr.15: 41564248-41564248 on Build GRCh38
rs764190736 Chr.15: 41564246-41564246 on Build GRCh38
rs765020875 Chr.15: 41564150-41564150 on Build GRCh38
rs1053265881 Chr.15: 41564144-41564144 on Build GRCh38
rs925960371 Chr.15: 41564426-41564426 on Build GRCh38
rs1014732098 Chr.15: 41564344-41564344 on Build GRCh38
rs752754184 Chr.15: 41564312-41564312 on Build GRCh38
rs117224665 Chr.15: 41564152-41564152 on Build GRCh38
rs756961845 Chr.15: 41564147-41564147 on Build GRCh38
rs557270589 Chr.15: 41564258-41564258 on Build GRCh38
rs765202263 Chr.15: 41564323-41564323 on Build GRCh38
rs932239226 Chr.15: 41564565-41564565 on Build GRCh38
rs1011618591 Chr.15: 41564509-41564524 on Build GRCh38
rs572480863 Chr.15: 41564365-41564365 on Build GRCh38
rs763161374 Chr.15: 41564274-41564274 on Build GRCh38
rs544721085 Chr.15: 41564494-41564494 on Build GRCh38
rs748295108 Chr.15: 41564193-41564193 on Build GRCh38
rs573845570 Chr.15: 41564335-41564335 on Build GRCh38
rs1048687683 Chr.15: 41564569-41564569 on Build GRCh38
rs957411147 Chr.15: 41564468-41564468 on Build GRCh38
rs558785782 Chr.15: 41564548-41564548 on Build GRCh38
rs537220234 Chr.15: 41564378-41564378 on Build GRCh38
rs980058427 Chr.15: 41564411-41564411 on Build GRCh38
rs758811069 Chr.15: 41564153-41564153 on Build GRCh38
rs781235941 Chr.15: 41564186-41564186 on Build GRCh38
rs1053251179 Chr.15: 41564444-41564444 on Build GRCh38
rs575839089 Chr.15: 41564592-41564592 on Build GRCh38
rs936072062 Chr.15: 41564132-41564132 on Build GRCh38
rs755473673 Chr.15: 41564291-41564291 on Build GRCh38
rs373641420 Chr.15: 41564322-41564322 on Build GRCh38
rs749534956 Chr.15: 41564319-41564319 on Build GRCh38
rs1042712824 Chr.15: 41564528-41564528 on Build GRCh38
rs2289742 Chr.15: 41564343-41564343 on Build GRCh38
rs769976823 Chr.15: 41564201-41564201 on Build GRCh38
rs781695888 Chr.15: 41564310-41564310 on Build GRCh38
rs760643930 Chr.15: 41564241-41564241 on Build GRCh38
rs887458955 Chr.15: 41564581-41564581 on Build GRCh38
rs865927269 Chr.15: 41564536-41564536 on Build GRCh38
rs756423982 Chr.15: 41564270-41564271 on Build GRCh38
rs1040621362 Chr.15: 41564529-41564529 on Build GRCh38
rs751415750 Chr.15: 41564472-41564472 on Build GRCh38
rs564614409 Chr.15: 41564525-41564525 on Build GRCh38
rs138227584 Chr.15: 41564203-41564203 on Build GRCh38
rs1011253888 Chr.15: 41564154-41564154 on Build GRCh38
rs745604133 Chr.15: 41564215-41564215 on Build GRCh38
rs746575481 Chr.15: 41564470-41564470 on Build GRCh38
rs201165681 Chr.15: 41564178-41564178 on Build GRCh38
rs944908372 Chr.15: 41564523-41564523 on Build GRCh38
rs911441395 Chr.15: 41564364-41564364 on Build GRCh38
rs199792732 Chr.15: 41564245-41564245 on Build GRCh38
rs751929267 Chr.15: 41564284-41564284 on Build GRCh38
rs780383951 Chr.15: 41564168-41564168 on Build GRCh38
rs11070339 Chr.15: 41564345-41564345 on Build GRCh38
rs775230837 Chr.15: 41564221-41564221 on Build GRCh38
rs750120069 Chr.15: 41564268-41564268 on Build GRCh38
rs1022102399 Chr.15: 41564394-41564394 on Build GRCh38
rs1041230028 Chr.15: 41564390-41564390 on Build GRCh38
rs777822199 Chr.15: 41564318-41564318 on Build GRCh38
rs766782057 Chr.15: 41564282-41564282 on Build GRCh38
rs761457140 Chr.15: 41564249-41564249 on Build GRCh38
rs918584631 Chr.15: 41564336-41564336 on Build GRCh38

View Full Product Details