Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41815181-41815689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPKBP1
Entrez Gene ID
23005
Gene Name
mitogen-activated protein kinase binding protein 1
Gene Aliases
JNKBP-1, JNKBP1
UniGene
Hs.513661
Gene Chromosome Location
Chr.15: 41774434-41827855 on Build GRCh38
Transcripts
NM_001128608.1 NM_001265611.1 NM_014994.2 NR_049761.1 NR_049762.1 XM_006720438.1 XM_011521383.1 XM_011521384.2 XM_017022017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7265845 c.1210C>T p.P404S Chr.15: 41815298-41815298 on Build GRCh38
4462071 c.1231C>T p.P411S Chr.15: 41815301-41815301 on Build GRCh38
4896978 c.1265C>T p.T422I Chr.15: 41815335-41815335 on Build GRCh38
50407 c.1247C>A p.T416N Chr.15: 41815335-41815335 on Build GRCh38
3672042 c.1345G>T p.G449W Chr.15: 41815651-41815651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004773820 Chr.15: 41815463-41815463 on Build GRCh38
rs766185662 Chr.15: 41815280-41815280 on Build GRCh38
rs183633907 Chr.15: 41815221-41815221 on Build GRCh38
rs756380624 Chr.15: 41815341-41815341 on Build GRCh38
rs987485754 Chr.15: 41815210-41815210 on Build GRCh38
rs199636307 Chr.15: 41815575-41815575 on Build GRCh38
rs894889935 Chr.15: 41815337-41815337 on Build GRCh38
rs893814353 Chr.15: 41815487-41815487 on Build GRCh38
rs772108230 Chr.15: 41815247-41815247 on Build GRCh38
rs962710250 Chr.15: 41815596-41815596 on Build GRCh38
rs774793622 Chr.15: 41815249-41815249 on Build GRCh38
rs746920711 Chr.15: 41815642-41815642 on Build GRCh38
rs747788536 Chr.15: 41815411-41815411 on Build GRCh38
rs764387739 Chr.15: 41815594-41815594 on Build GRCh38
rs1043036683 Chr.15: 41815496-41815496 on Build GRCh38
rs200313290 Chr.15: 41815626-41815626 on Build GRCh38
rs1029875644 Chr.15: 41815544-41815544 on Build GRCh38
rs949888640 Chr.15: 41815387-41815387 on Build GRCh38
rs555101294 Chr.15: 41815273-41815273 on Build GRCh38
rs936137674 Chr.15: 41815267-41815267 on Build GRCh38
rs750132909 Chr.15: 41815602-41815602 on Build GRCh38
rs766748747 Chr.15: 41815450-41815450 on Build GRCh38
rs1011135910 Chr.15: 41815492-41815492 on Build GRCh38
rs767581780 Chr.15: 41815612-41815612 on Build GRCh38
rs759760650 Chr.15: 41815591-41815591 on Build GRCh38
rs200396823 Chr.15: 41815431-41815431 on Build GRCh38
rs902650704 Chr.15: 41815499-41815499 on Build GRCh38
rs1029427174 Chr.15: 41815226-41815226 on Build GRCh38
rs143147781 Chr.15: 41815364-41815364 on Build GRCh38
rs998347080 Chr.15: 41815530-41815530 on Build GRCh38
rs752835842 Chr.15: 41815588-41815588 on Build GRCh38
rs760081154 Chr.15: 41815580-41815580 on Build GRCh38
rs763363819 Chr.15: 41815301-41815301 on Build GRCh38
rs1021095816 Chr.15: 41815206-41815206 on Build GRCh38
rs267604190 Chr.15: 41815308-41815308 on Build GRCh38
rs994620700 Chr.15: 41815550-41815550 on Build GRCh38
rs756184203 Chr.15: 41815630-41815630 on Build GRCh38
rs151201972 Chr.15: 41815380-41815380 on Build GRCh38
rs773683081 Chr.15: 41815288-41815288 on Build GRCh38
rs199681928 Chr.15: 41815656-41815656 on Build GRCh38
rs909563190 Chr.15: 41815345-41815345 on Build GRCh38
rs200746378 Chr.15: 41815412-41815412 on Build GRCh38
rs755072633 Chr.15: 41815317-41815317 on Build GRCh38
rs1029531002 Chr.15: 41815560-41815560 on Build GRCh38
rs941274600 Chr.15: 41815375-41815375 on Build GRCh38
rs374321273 Chr.15: 41815643-41815643 on Build GRCh38
rs766915233 Chr.15: 41815309-41815309 on Build GRCh38
rs368296428 Chr.15: 41815664-41815664 on Build GRCh38
rs751536100 Chr.15: 41815315-41815315 on Build GRCh38
rs544587803 Chr.15: 41815276-41815276 on Build GRCh38
rs1046428790 Chr.15: 41815389-41815389 on Build GRCh38
rs774095944 Chr.15: 41815467-41815467 on Build GRCh38
rs1053976577 Chr.15: 41815293-41815293 on Build GRCh38
rs768249761 Chr.15: 41815385-41815385 on Build GRCh38
rs373370590 Chr.15: 41815264-41815264 on Build GRCh38
rs776451536 Chr.15: 41815271-41815271 on Build GRCh38
rs113020718 Chr.15: 41815373-41815373 on Build GRCh38
rs778561350 Chr.15: 41815359-41815359 on Build GRCh38
rs749034899 Chr.15: 41815214-41815214 on Build GRCh38
rs752954086 Chr.15: 41815614-41815614 on Build GRCh38
rs377392952 Chr.15: 41815318-41815318 on Build GRCh38
rs760272477 Chr.15: 41815251-41815251 on Build GRCh38
rs202001274 Chr.15: 41815388-41815388 on Build GRCh38
rs752889005 Chr.15: 41815340-41815340 on Build GRCh38
rs1055122974 Chr.15: 41815466-41815466 on Build GRCh38
rs953490863 Chr.15: 41815562-41815562 on Build GRCh38
rs879244230 Chr.15: 41815352-41815352 on Build GRCh38
rs745773925 Chr.15: 41815236-41815236 on Build GRCh38
rs778812927 Chr.15: 41815222-41815222 on Build GRCh38
rs140264064 Chr.15: 41815669-41815669 on Build GRCh38
rs779750168 Chr.15: 41815637-41815637 on Build GRCh38
rs767566089 Chr.15: 41815581-41815581 on Build GRCh38

View Full Product Details