Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74181103-74181594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRA6
Entrez Gene ID
64220
Gene Name
stimulated by retinoic acid 6
Gene Aliases
MCOPCB8, MCOPS9, PP14296
UniGene
Hs.24553
Gene Chromosome Location
Chr.15: 74179466-74212267 on Build GRCh38
Transcripts
NM_001142617.1 NM_001142618.1 NM_001142619.1 NM_001199040.1 NM_001199041.1 NM_001199042.1 NM_022369.3 XM_011521883.1 XM_011521884.1 XM_017022478.1 XM_017022479.1 XM_017022480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4904010 c.1640G>A p.R547Q Chr.15: 74181456-74181456 on Build GRCh38
6586951 c.1690G>T p.G564C Chr.15: 74181406-74181406 on Build GRCh38
147950 c.1581G>A p.M527I Chr.15: 74181398-74181398 on Build GRCh38
315667 c.1669G>A p.A557T Chr.15: 74181310-74181310 on Build GRCh38
6586952 c.1573G>T p.G525C Chr.15: 74181406-74181406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757313431 Chr.15: 74181480-74181480 on Build GRCh38
rs961317341 Chr.15: 74181210-74181210 on Build GRCh38
rs779753862 Chr.15: 74181349-74181349 on Build GRCh38
rs777759255 Chr.15: 74181464-74181464 on Build GRCh38
rs964727708 Chr.15: 74181351-74181351 on Build GRCh38
rs901979311 Chr.15: 74181395-74181395 on Build GRCh38
rs766874761 Chr.15: 74181337-74181337 on Build GRCh38
rs3743213 Chr.15: 74181155-74181155 on Build GRCh38
rs952585823 Chr.15: 74181232-74181232 on Build GRCh38
rs765784282 Chr.15: 74181319-74181319 on Build GRCh38
rs397518484 Chr.15: 74181459-74181459 on Build GRCh38
rs748286024 Chr.15: 74181309-74181309 on Build GRCh38
rs532103784 Chr.15: 74181217-74181217 on Build GRCh38
rs76890765 Chr.15: 74181275-74181275 on Build GRCh38
rs765716879 Chr.15: 74181276-74181276 on Build GRCh38
rs572207002 Chr.15: 74181563-74181563 on Build GRCh38
rs755780472 Chr.15: 74181346-74181346 on Build GRCh38
rs1017713443 Chr.15: 74181129-74181129 on Build GRCh38
rs756741849 Chr.15: 74181279-74181279 on Build GRCh38
rs769084855 Chr.15: 74181509-74181509 on Build GRCh38
rs201582612 Chr.15: 74181302-74181302 on Build GRCh38
rs910950973 Chr.15: 74181295-74181295 on Build GRCh38
rs759887263 Chr.15: 74181318-74181318 on Build GRCh38
rs753156796 Chr.15: 74181330-74181330 on Build GRCh38
rs776915144 Chr.15: 74181317-74181317 on Build GRCh38
rs368883595 Chr.15: 74181310-74181310 on Build GRCh38
rs141004067 Chr.15: 74181374-74181374 on Build GRCh38
rs747096035 Chr.15: 74181479-74181479 on Build GRCh38
rs779656502 Chr.15: 74181296-74181296 on Build GRCh38
rs781427025 Chr.15: 74181484-74181484 on Build GRCh38
rs372149672 Chr.15: 74181311-74181311 on Build GRCh38
rs770108092 Chr.15: 74181402-74181402 on Build GRCh38
rs186512506 Chr.15: 74181167-74181167 on Build GRCh38
rs1027249394 Chr.15: 74181496-74181496 on Build GRCh38
rs771122659 Chr.15: 74181394-74181394 on Build GRCh38
rs778960900 Chr.15: 74181304-74181304 on Build GRCh38
rs766612045 Chr.15: 74181259-74181259 on Build GRCh38
rs753490176 Chr.15: 74181438-74181438 on Build GRCh38
rs761122376 Chr.15: 74181246-74181246 on Build GRCh38
rs994411503 Chr.15: 74181216-74181216 on Build GRCh38
rs985188018 Chr.15: 74181266-74181266 on Build GRCh38
rs570214336 Chr.15: 74181385-74181385 on Build GRCh38
rs965095410 Chr.15: 74181545-74181545 on Build GRCh38
rs752691134 Chr.15: 74181452-74181452 on Build GRCh38
rs372011272 Chr.15: 74181377-74181377 on Build GRCh38
rs776814085 Chr.15: 74181397-74181397 on Build GRCh38
rs750103700 Chr.15: 74181341-74181341 on Build GRCh38
rs775984886 Chr.15: 74181403-74181403 on Build GRCh38
rs375362287 Chr.15: 74181358-74181358 on Build GRCh38
rs755941898 Chr.15: 74181294-74181294 on Build GRCh38
rs754849860 Chr.15: 74181447-74181447 on Build GRCh38
rs770161440 Chr.15: 74181490-74181490 on Build GRCh38
rs765057513 Chr.15: 74181449-74181449 on Build GRCh38
rs1036328200 Chr.15: 74181412-74181412 on Build GRCh38
rs1017612903 Chr.15: 74181336-74181336 on Build GRCh38
rs758744030 Chr.15: 74181376-74181376 on Build GRCh38
rs190173343 Chr.15: 74181193-74181193 on Build GRCh38
rs202138009 Chr.15: 74181280-74181280 on Build GRCh38
rs369050725 Chr.15: 74181421-74181421 on Build GRCh38
rs375230883 Chr.15: 74181519-74181519 on Build GRCh38
rs559042530 Chr.15: 74181414-74181414 on Build GRCh38
rs141168559 Chr.15: 74181355-74181355 on Build GRCh38
rs371301233 Chr.15: 74181435-74181435 on Build GRCh38
rs145465608 Chr.15: 74181315-74181315 on Build GRCh38
rs966277279 Chr.15: 74181146-74181146 on Build GRCh38
rs1006663617 Chr.15: 74181502-74181502 on Build GRCh38
rs772666646 Chr.15: 74181423-74181423 on Build GRCh38
rs879868238 Chr.15: 74181570-74181570 on Build GRCh38
rs751112020 Chr.15: 74181277-74181277 on Build GRCh38
rs11545567 Chr.15: 74181430-74181430 on Build GRCh38
rs748057183 Chr.15: 74181367-74181367 on Build GRCh38
rs150151099 Chr.15: 74181489-74181489 on Build GRCh38
rs886051474 Chr.15: 74181413-74181413 on Build GRCh38
rs763738075 Chr.15: 74181156-74181156 on Build GRCh38
rs947109228 Chr.15: 74181321-74181321 on Build GRCh38
rs560976868 Chr.15: 74181128-74181128 on Build GRCh38
rs553473245 Chr.15: 74181533-74181533 on Build GRCh38
rs397514638 Chr.15: 74181301-74181301 on Build GRCh38
rs758321198 Chr.15: 74181456-74181456 on Build GRCh38
rs736118 Chr.15: 74181398-74181398 on Build GRCh38
rs775890826 Chr.15: 74181495-74181495 on Build GRCh38
rs535135524 Chr.15: 74181503-74181503 on Build GRCh38
rs760204278 Chr.15: 74181426-74181426 on Build GRCh38
rs761346851 Chr.15: 74181335-74181335 on Build GRCh38
rs754370300 Chr.15: 74181265-74181265 on Build GRCh38
rs201175968 Chr.15: 74181393-74181393 on Build GRCh38
rs146956561 Chr.15: 74181359-74181359 on Build GRCh38
rs867383574 Chr.15: 74181338-74181338 on Build GRCh38
rs888301090 Chr.15: 74181498-74181498 on Build GRCh38
rs765995514 Chr.15: 74181437-74181437 on Build GRCh38

View Full Product Details