Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78266138-78266544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA4
Entrez Gene ID
55466
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
Gene Aliases
MST104, MSTP104, PRO1472
UniGene
Hs.513053
Gene Chromosome Location
Chr.15: 78264145-78282196 on Build GRCh38
Transcripts
NM_001130182.1 NM_001130183.1 NM_018602.3 XM_005254535.3 XM_006720601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4602236 c.43C>T p.L15F Chr.15: 78266285-78266285 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758817590 Chr.15: 78266335-78266335 on Build GRCh38
rs142025971 Chr.15: 78266243-78266244 on Build GRCh38
rs748114839 Chr.15: 78266319-78266319 on Build GRCh38
rs1048723713 Chr.15: 78266422-78266422 on Build GRCh38
rs887442031 Chr.15: 78266430-78266430 on Build GRCh38
rs774463559 Chr.15: 78266276-78266276 on Build GRCh38
rs962903968 Chr.15: 78266452-78266452 on Build GRCh38
rs762981880 Chr.15: 78266230-78266230 on Build GRCh38
rs10152846 Chr.15: 78266515-78266515 on Build GRCh38
rs755838209 Chr.15: 78266377-78266377 on Build GRCh38
rs747945944 Chr.15: 78266215-78266215 on Build GRCh38
rs900251452 Chr.15: 78266289-78266289 on Build GRCh38
rs995636045 Chr.15: 78266497-78266497 on Build GRCh38
rs777567475 Chr.15: 78266343-78266343 on Build GRCh38
rs754466028 Chr.15: 78266306-78266306 on Build GRCh38
rs778759370 Chr.15: 78266315-78266315 on Build GRCh38
rs936503865 Chr.15: 78266211-78266211 on Build GRCh38
rs1012456523 Chr.15: 78266513-78266513 on Build GRCh38
rs1035165837 Chr.15: 78266504-78266504 on Build GRCh38
rs776786246 Chr.15: 78266242-78266242 on Build GRCh38
rs764184013 Chr.15: 78266254-78266254 on Build GRCh38
rs879128262 Chr.15: 78266379-78266379 on Build GRCh38
rs760480186 Chr.15: 78266242-78266243 on Build GRCh38
rs771148304 Chr.15: 78266237-78266237 on Build GRCh38
rs750053705 Chr.15: 78266283-78266283 on Build GRCh38
rs921820379 Chr.15: 78266353-78266353 on Build GRCh38
rs7179906 Chr.15: 78266288-78266288 on Build GRCh38
rs530534531 Chr.15: 78266296-78266296 on Build GRCh38
rs763291658 Chr.15: 78266278-78266278 on Build GRCh38
rs886143132 Chr.15: 78266340-78266340 on Build GRCh38
rs750803205 Chr.15: 78266300-78266306 on Build GRCh38
rs758186324 Chr.15: 78266338-78266338 on Build GRCh38
rs1040253278 Chr.15: 78266347-78266347 on Build GRCh38
rs771871312 Chr.15: 78266225-78266225 on Build GRCh38
rs768537664 Chr.15: 78266206-78266206 on Build GRCh38
rs1051941242 Chr.15: 78266308-78266308 on Build GRCh38
rs906972509 Chr.15: 78266462-78266462 on Build GRCh38
rs541929187 Chr.15: 78266255-78266255 on Build GRCh38
rs753300073 Chr.15: 78266304-78266304 on Build GRCh38
rs372756252 Chr.15: 78266202-78266202 on Build GRCh38
rs1004992051 Chr.15: 78266444-78266444 on Build GRCh38
rs753215584 Chr.15: 78266270-78266270 on Build GRCh38
rs775583047 Chr.15: 78266171-78266171 on Build GRCh38
rs778705789 Chr.15: 78266212-78266212 on Build GRCh38
rs112449658 Chr.15: 78266411-78266411 on Build GRCh38
rs978197778 Chr.15: 78266286-78266286 on Build GRCh38
rs565286600 Chr.15: 78266463-78266463 on Build GRCh38
rs764503943 Chr.15: 78266282-78266282 on Build GRCh38
rs1003691525 Chr.15: 78266496-78266496 on Build GRCh38
rs959581686 Chr.15: 78266511-78266511 on Build GRCh38
rs2037347 Chr.15: 78266167-78266167 on Build GRCh38
rs990752919 Chr.15: 78266231-78266231 on Build GRCh38
rs760141748 Chr.15: 78266244-78266244 on Build GRCh38
rs543713177 Chr.15: 78266367-78266367 on Build GRCh38
rs767974932 Chr.15: 78266394-78266394 on Build GRCh38
rs968498523 Chr.15: 78266279-78266279 on Build GRCh38
rs779478448 Chr.15: 78266194-78266194 on Build GRCh38

View Full Product Details