Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82986111-82986604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM103A1
Entrez Gene ID
83640
Gene Name
family with sequence similarity 103 member A1
Gene Aliases
C15orf18, HsT19360, RAM
UniGene
Hs.80624
Gene Chromosome Location
Chr.15: 82986203-82991057 on Build GRCh38
Transcripts
NM_031452.3 XM_005272449.3 XM_006720776.3

Gene Symbol
HOMER2
Entrez Gene ID
9455
Gene Name
homer scaffolding protein 2
Gene Aliases
ACPD, CPD, DFNA68, HOMER-2, VESL-2
UniGene
Hs.578443
Gene Chromosome Location
Chr.15: 82836946-82986176 on Build GRCh38
Transcripts
NM_031452.3 XM_005272449.3 XM_006720776.3

SNP Detail

SNP ID Chromosome Location
rs1021099701 Chr.15: 82986149-82986149 on Build GRCh38
rs776808533 Chr.15: 82986198-82986198 on Build GRCh38
rs557321190 Chr.15: 82986188-82986188 on Build GRCh38
rs766669661 Chr.15: 82986193-82986193 on Build GRCh38
rs933940901 Chr.15: 82986215-82986215 on Build GRCh38
rs1042060430 Chr.15: 82986401-82986401 on Build GRCh38
rs577306963 Chr.15: 82986201-82986201 on Build GRCh38
rs191332845 Chr.15: 82986447-82986447 on Build GRCh38
rs943715059 Chr.15: 82986371-82986371 on Build GRCh38
rs983509246 Chr.15: 82986259-82986259 on Build GRCh38
rs376492851 Chr.15: 82986200-82986200 on Build GRCh38
rs537052435 Chr.15: 82986177-82986177 on Build GRCh38
rs11545236 Chr.15: 82986305-82986305 on Build GRCh38
rs765663672 Chr.15: 82986241-82986241 on Build GRCh38
rs959517625 Chr.15: 82986514-82986514 on Build GRCh38
rs546305021 Chr.15: 82986236-82986236 on Build GRCh38
rs906243023 Chr.15: 82986417-82986417 on Build GRCh38
rs112694444 Chr.15: 82986330-82986330 on Build GRCh38
rs763589116 Chr.15: 82986384-82986384 on Build GRCh38
rs1021115325 Chr.15: 82986192-82986192 on Build GRCh38
rs1004083104 Chr.15: 82986209-82986209 on Build GRCh38
rs920898967 Chr.15: 82986214-82986214 on Build GRCh38
rs553631878 Chr.15: 82986237-82986237 on Build GRCh38
rs543909335 Chr.15: 82986256-82986256 on Build GRCh38
rs1033820166 Chr.15: 82986234-82986234 on Build GRCh38
rs994999952 Chr.15: 82986529-82986529 on Build GRCh38
rs896456461 Chr.15: 82986405-82986405 on Build GRCh38
rs966886728 Chr.15: 82986578-82986578 on Build GRCh38
rs1027857192 Chr.15: 82986566-82986566 on Build GRCh38
rs1021130766 Chr.15: 82986553-82986553 on Build GRCh38
rs192869782 Chr.15: 82986134-82986134 on Build GRCh38
rs960975809 Chr.15: 82986137-82986137 on Build GRCh38
rs939827798 Chr.15: 82986266-82986266 on Build GRCh38
rs921116583 Chr.15: 82986323-82986323 on Build GRCh38
rs542409633 Chr.15: 82986249-82986249 on Build GRCh38
rs532889813 Chr.15: 82986291-82986291 on Build GRCh38
rs573501011 Chr.15: 82986248-82986248 on Build GRCh38
rs564091221 Chr.15: 82986260-82986260 on Build GRCh38
rs1013796837 Chr.15: 82986526-82986526 on Build GRCh38
rs908382869 Chr.15: 82986239-82986239 on Build GRCh38
rs1009644527 Chr.15: 82986360-82986360 on Build GRCh38
rs1033787645 Chr.15: 82986477-82986477 on Build GRCh38
rs1014227712 Chr.15: 82986144-82986144 on Build GRCh38
rs377668077 Chr.15: 82986203-82986203 on Build GRCh38
rs546308731 Chr.15: 82986324-82986324 on Build GRCh38
rs188655962 Chr.15: 82986326-82986326 on Build GRCh38
rs760772904 Chr.15: 82986150-82986150 on Build GRCh38
rs562257631 Chr.15: 82986250-82986250 on Build GRCh38
rs977151171 Chr.15: 82986168-82986168 on Build GRCh38
rs569103687 Chr.15: 82986562-82986562 on Build GRCh38
rs944816011 Chr.15: 82986289-82986289 on Build GRCh38
rs575851232 Chr.15: 82986254-82986254 on Build GRCh38
rs185792449 Chr.15: 82986135-82986135 on Build GRCh38
rs74030863 Chr.15: 82986352-82986352 on Build GRCh38
rs893458655 Chr.15: 82986153-82986153 on Build GRCh38
rs371044787 Chr.15: 82986283-82986283 on Build GRCh38
rs569092916 Chr.15: 82986141-82986141 on Build GRCh38
rs950766128 Chr.15: 82986251-82986251 on Build GRCh38
rs369909647 Chr.15: 82986441-82986441 on Build GRCh38
rs749764149 Chr.15: 82986498-82986498 on Build GRCh38

View Full Product Details